Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 27184644-27184885 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXA11
Entrez Gene ID
3207
Gene Name
homeobox A11
Gene Aliases
HOX1, HOX1I, RUSAT1
UniGene
Hs.249171
Gene Chromosome Location
Chr.7: 27181157-27185216 on Build GRCh38
Transcripts
NM_005523.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7198310 c.340G>A p.V114I Chr.7: 27184805-27184805 on Build GRCh38
6177729 c.300C>T p.A100A Chr.7: 27184845-27184845 on Build GRCh38
1088884 c.412C>T p.Q138* Chr.7: 27184733-27184733 on Build GRCh38
7139021 c.378C>G p.F126L Chr.7: 27184767-27184767 on Build GRCh38
1088885 c.378C>A p.F126L Chr.7: 27184767-27184767 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758601680 Chr.7: 27184776-27184776 on Build GRCh38
rs776561756 Chr.7: 27184686-27184686 on Build GRCh38
rs995097601 Chr.7: 27184737-27184737 on Build GRCh38
rs756117422 Chr.7: 27184839-27184839 on Build GRCh38
rs577175554 Chr.7: 27184848-27184856 on Build GRCh38
rs199544203 Chr.7: 27184784-27184784 on Build GRCh38
rs544898158 Chr.7: 27184662-27184662 on Build GRCh38
rs202065784 Chr.7: 27184729-27184729 on Build GRCh38
rs967315565 Chr.7: 27184668-27184668 on Build GRCh38
rs745804777 Chr.7: 27184712-27184712 on Build GRCh38
rs200384070 Chr.7: 27184859-27184859 on Build GRCh38
rs755029079 Chr.7: 27184664-27184664 on Build GRCh38
rs557720238 Chr.7: 27184683-27184683 on Build GRCh38
rs146591552 Chr.7: 27184835-27184835 on Build GRCh38
rs143812636 Chr.7: 27184749-27184749 on Build GRCh38
rs371593760 Chr.7: 27184763-27184763 on Build GRCh38
rs776902959 Chr.7: 27184817-27184817 on Build GRCh38
rs926107951 Chr.7: 27184813-27184813 on Build GRCh38
rs778600905 Chr.7: 27184844-27184844 on Build GRCh38
rs778031419 Chr.7: 27184779-27184779 on Build GRCh38
rs369491882 Chr.7: 27184832-27184832 on Build GRCh38
rs138305707 Chr.7: 27184785-27184785 on Build GRCh38
rs758663019 Chr.7: 27184681-27184681 on Build GRCh38
rs762951157 Chr.7: 27184829-27184829 on Build GRCh38
rs752827520 Chr.7: 27184775-27184775 on Build GRCh38
rs764169770 Chr.7: 27184831-27184831 on Build GRCh38
rs369311259 Chr.7: 27184786-27184786 on Build GRCh38
rs752764603 Chr.7: 27184822-27184822 on Build GRCh38
rs200850004 Chr.7: 27184724-27184724 on Build GRCh38
rs775170368 Chr.7: 27184730-27184730 on Build GRCh38
rs760069787 Chr.7: 27184820-27184820 on Build GRCh38
rs761396527 Chr.7: 27184811-27184811 on Build GRCh38
rs952479268 Chr.7: 27184777-27184777 on Build GRCh38
rs778884222 Chr.7: 27184665-27184665 on Build GRCh38
rs777044854 Chr.7: 27184851-27184851 on Build GRCh38
rs751732678 Chr.7: 27184780-27184780 on Build GRCh38
rs527502075 Chr.7: 27184746-27184746 on Build GRCh38
rs976975849 Chr.7: 27184802-27184802 on Build GRCh38
rs375344699 Chr.7: 27184677-27184677 on Build GRCh38
rs200352994 Chr.7: 27184845-27184845 on Build GRCh38
rs777519049 Chr.7: 27184682-27184682 on Build GRCh38
rs377286501 Chr.7: 27184815-27184815 on Build GRCh38
rs757449754 Chr.7: 27184833-27184833 on Build GRCh38
rs372937744 Chr.7: 27184797-27184797 on Build GRCh38
rs761036303 Chr.7: 27184751-27184751 on Build GRCh38
rs766668972 Chr.7: 27184752-27184752 on Build GRCh38
rs771674623 Chr.7: 27184850-27184850 on Build GRCh38
rs868398286 Chr.7: 27184732-27184732 on Build GRCh38
rs759995027 Chr.7: 27184772-27184772 on Build GRCh38
rs774090664 Chr.7: 27184809-27184809 on Build GRCh38
rs201074014 Chr.7: 27184836-27184836 on Build GRCh38
rs765788044 Chr.7: 27184821-27184821 on Build GRCh38
rs779688027 Chr.7: 27184806-27184806 on Build GRCh38
rs768233475 Chr.7: 27184842-27184842 on Build GRCh38
rs543102978 Chr.7: 27184688-27184688 on Build GRCh38
rs780070206 Chr.7: 27184841-27184841 on Build GRCh38
rs768286488 Chr.7: 27184807-27184807 on Build GRCh38

View Full Product Details