Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 85107283-85107518 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRCC1
Entrez Gene ID
85444
Gene Name
leucine rich repeat and coiled-coil centrosomal protein 1
Gene Aliases
CLERC, SAP2, VFL1
UniGene
Hs.193115
Gene Chromosome Location
Chr.8: 85107088-85146080 on Build GRCh38
Transcripts
NM_033402.4 XM_005251318.3 XM_005251319.4 XM_005251320.3 XM_005251322.3 XM_017013920.1 XM_017013921.1 XM_017013922.1 XM_017013923.1 XM_017013924.1 XM_017013926.1 XM_017013927.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4792763 c.14C>A p.A5E Chr.8: 85107309-85107309 on Build GRCh38
1458400 c.17C>T p.A6V Chr.8: 85107312-85107312 on Build GRCh38
6704120 c.93A>G p.K31K Chr.8: 85107388-85107388 on Build GRCh38
486701 c.42G>T p.V14V Chr.8: 85107337-85107337 on Build GRCh38
5680991 c.78A>G p.V26V Chr.8: 85107373-85107373 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547117503 Chr.8: 85107382-85107382 on Build GRCh38
rs1041138548 Chr.8: 85107372-85107372 on Build GRCh38
rs771541384 Chr.8: 85107341-85107341 on Build GRCh38
rs745850491 Chr.8: 85107408-85107408 on Build GRCh38
rs773342260 Chr.8: 85107310-85107315 on Build GRCh38
rs779001897 Chr.8: 85107326-85107326 on Build GRCh38
rs772121565 Chr.8: 85107300-85107301 on Build GRCh38
rs755913046 Chr.8: 85107310-85107310 on Build GRCh38
rs537140038 Chr.8: 85107349-85107349 on Build GRCh38
rs757206537 Chr.8: 85107321-85107321 on Build GRCh38
rs78932083 Chr.8: 85107397-85107397 on Build GRCh38
rs1002052618 Chr.8: 85107307-85107318 on Build GRCh38
rs776269737 Chr.8: 85107426-85107426 on Build GRCh38
rs369922208 Chr.8: 85107431-85107431 on Build GRCh38
rs908250756 Chr.8: 85107455-85107455 on Build GRCh38
rs983867959 Chr.8: 85107447-85107447 on Build GRCh38
rs371924871 Chr.8: 85107306-85107306 on Build GRCh38
rs756247543 Chr.8: 85107389-85107389 on Build GRCh38
rs759998882 Chr.8: 85107435-85107435 on Build GRCh38
rs749120809 Chr.8: 85107315-85107315 on Build GRCh38
rs766803830 Chr.8: 85107360-85107360 on Build GRCh38
rs775187457 Chr.8: 85107347-85107347 on Build GRCh38
rs777466741 Chr.8: 85107313-85107313 on Build GRCh38
rs567279699 Chr.8: 85107351-85107351 on Build GRCh38
rs753623083 Chr.8: 85107396-85107396 on Build GRCh38
rs939681818 Chr.8: 85107456-85107456 on Build GRCh38
rs927608812 Chr.8: 85107469-85107469 on Build GRCh38
rs745372497 Chr.8: 85107328-85107328 on Build GRCh38
rs766588880 Chr.8: 85107315-85107316 on Build GRCh38
rs371725003 Chr.8: 85107411-85107411 on Build GRCh38
rs369190714 Chr.8: 85107467-85107467 on Build GRCh38
rs558438806 Chr.8: 85107415-85107415 on Build GRCh38
rs200508775 Chr.8: 85107373-85107373 on Build GRCh38
rs773514193 Chr.8: 85107355-85107355 on Build GRCh38
rs969792343 Chr.8: 85107466-85107466 on Build GRCh38
rs376358358 Chr.8: 85107430-85107430 on Build GRCh38
rs1049570706 Chr.8: 85107353-85107353 on Build GRCh38
rs1011709095 Chr.8: 85107318-85107319 on Build GRCh38
rs774905053 Chr.8: 85107362-85107362 on Build GRCh38
rs749648742 Chr.8: 85107429-85107429 on Build GRCh38
rs781467429 Chr.8: 85107303-85107303 on Build GRCh38
rs746682817 Chr.8: 85107350-85107350 on Build GRCh38
rs757896086 Chr.8: 85107409-85107409 on Build GRCh38
rs760878712 Chr.8: 85107312-85107313 on Build GRCh38
rs902499620 Chr.8: 85107386-85107386 on Build GRCh38
rs369075882 Chr.8: 85107383-85107383 on Build GRCh38
rs759644779 Chr.8: 85107365-85107365 on Build GRCh38
rs200933031 Chr.8: 85107309-85107309 on Build GRCh38
rs893626987 Chr.8: 85107314-85107314 on Build GRCh38
rs965628193 Chr.8: 85107419-85107419 on Build GRCh38
rs762841704 Chr.8: 85107301-85107303 on Build GRCh38
rs778985291 Chr.8: 85107407-85107407 on Build GRCh38
rs764427810 Chr.8: 85107393-85107393 on Build GRCh38

View Full Product Details