Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49831887-49832140 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MED25
Entrez Gene ID
81857
Gene Name
mediator complex subunit 25
Gene Aliases
ACID1, ARC92, BVSYS, CMT2B2, P78, PTOV2, TCBAP0758
UniGene
Hs.656639
Gene Chromosome Location
Chr.19: 49818279-49840384 on Build GRCh38
Transcripts
NM_030973.3 XM_011527353.1 NR_106858.1

Gene Symbol
MIR6800
Entrez Gene ID
102465480
Gene Name
microRNA 6800
Gene Aliases
hsa-mir-6800
UniGene
Gene Chromosome Location
Chr.19: 49832018-49832099 on Build GRCh38
Transcripts
NM_030973.3 XM_011527353.1 NR_106858.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4080216 c.1326G>A p.E442E Chr.19: 49832109-49832109 on Build GRCh38
4462453 c.1247C>T p.S416L Chr.19: 49831952-49831952 on Build GRCh38
5794189 c.1231-3C>T p.? Chr.19: 49831933-49831933 on Build GRCh38
7088173 c.1320G>T p.K440N Chr.19: 49832103-49832103 on Build GRCh38
1481347 c.1271C>T p.T424M Chr.19: 49831976-49831976 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763995731 Chr.19: 49831984-49831984 on Build GRCh38
rs775829724 Chr.19: 49832042-49832042 on Build GRCh38
rs921015866 Chr.19: 49832069-49832069 on Build GRCh38
rs149030211 Chr.19: 49832019-49832019 on Build GRCh38
rs751247324 Chr.19: 49832088-49832088 on Build GRCh38
rs201845985 Chr.19: 49831927-49831927 on Build GRCh38
rs74182073 Chr.19: 49832060-49832061 on Build GRCh38
rs367774572 Chr.19: 49832097-49832097 on Build GRCh38
rs759682535 Chr.19: 49831944-49831944 on Build GRCh38
rs778873937 Chr.19: 49832039-49832039 on Build GRCh38
rs749843673 Chr.19: 49832029-49832029 on Build GRCh38
rs759416367 Chr.19: 49832074-49832075 on Build GRCh38
rs74182072 Chr.19: 49832049-49832050 on Build GRCh38
rs750031901 Chr.19: 49832013-49832013 on Build GRCh38
rs761751386 Chr.19: 49832065-49832065 on Build GRCh38
rs201263206 Chr.19: 49832072-49832072 on Build GRCh38
rs912091485 Chr.19: 49832081-49832081 on Build GRCh38
rs887032543 Chr.19: 49832035-49832035 on Build GRCh38
rs753794673 Chr.19: 49832000-49832000 on Build GRCh38
rs757574065 Chr.19: 49832045-49832046 on Build GRCh38
rs778157941 Chr.19: 49831926-49831927 on Build GRCh38
rs924251754 Chr.19: 49831962-49831962 on Build GRCh38
rs779825460 Chr.19: 49832016-49832016 on Build GRCh38
rs757971451 Chr.19: 49832014-49832014 on Build GRCh38
rs375259879 Chr.19: 49832051-49832051 on Build GRCh38
rs768470202 Chr.19: 49832046-49832046 on Build GRCh38
rs764727431 Chr.19: 49832004-49832004 on Build GRCh38
rs897801306 Chr.19: 49832093-49832093 on Build GRCh38
rs749360253 Chr.19: 49831930-49831930 on Build GRCh38
rs747395243 Chr.19: 49832000-49832001 on Build GRCh38
rs986697858 Chr.19: 49832076-49832076 on Build GRCh38
rs765215439 Chr.19: 49832068-49832068 on Build GRCh38
rs200594031 Chr.19: 49831928-49831928 on Build GRCh38
rs778380162 Chr.19: 49832026-49832026 on Build GRCh38
rs370255278 Chr.19: 49832091-49832091 on Build GRCh38
rs1025557662 Chr.19: 49832025-49832025 on Build GRCh38
rs1036590547 Chr.19: 49832092-49832092 on Build GRCh38
rs774675537 Chr.19: 49831938-49831938 on Build GRCh38
rs956950038 Chr.19: 49832045-49832045 on Build GRCh38
rs757340805 Chr.19: 49832030-49832030 on Build GRCh38
rs566927009 Chr.19: 49832040-49832040 on Build GRCh38
rs113483619 Chr.19: 49831933-49831933 on Build GRCh38
rs765926397 Chr.19: 49832082-49832082 on Build GRCh38
rs760556318 Chr.19: 49831969-49831969 on Build GRCh38
rs748829394 Chr.19: 49832083-49832083 on Build GRCh38
rs533964778 Chr.19: 49832041-49832041 on Build GRCh38
rs370984356 Chr.19: 49832044-49832044 on Build GRCh38
rs771182615 Chr.19: 49831925-49831925 on Build GRCh38
rs372191174 Chr.19: 49832118-49832118 on Build GRCh38
rs570852452 Chr.19: 49832078-49832078 on Build GRCh38
rs775141134 Chr.19: 49831949-49831949 on Build GRCh38
rs754342751 Chr.19: 49832105-49832105 on Build GRCh38
rs771844561 Chr.19: 49831948-49831948 on Build GRCh38
rs532438353 Chr.19: 49831908-49831908 on Build GRCh38
rs748601344 Chr.19: 49831926-49831926 on Build GRCh38
rs529873708 Chr.19: 49831968-49831968 on Build GRCh38
rs761268819 Chr.19: 49832001-49832001 on Build GRCh38

View Full Product Details