Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 155138045-155138286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MME
Entrez Gene ID
4311
Gene Name
membrane metallo-endopeptidase
Gene Aliases
CALLA, CD10, CMT2T, NEP, SFE
UniGene
Hs.307734
Gene Chromosome Location
Chr.3: 155024124-155183729 on Build GRCh38
Transcripts
NM_000902.3 NM_007287.2 NM_007288.2 NM_007289.2 XM_006713646.3 XM_006713647.3 XM_011512855.2 XM_011512856.2 XM_011512857.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5683752 c.731C>G p.A244G Chr.3: 155138112-155138112 on Build GRCh38
4114665 c.844G>A p.E282K Chr.3: 155138225-155138225 on Build GRCh38
1040250 c.769C>T p.R257C Chr.3: 155138150-155138150 on Build GRCh38
1693632 c.776A>T p.E259V Chr.3: 155138157-155138157 on Build GRCh38
3774663 c.819G>C p.M273I Chr.3: 155138200-155138200 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200961860 Chr.3: 155138235-155138235 on Build GRCh38
rs774720833 Chr.3: 155138250-155138250 on Build GRCh38
rs756294077 Chr.3: 155138182-155138182 on Build GRCh38
rs79211121 Chr.3: 155138106-155138106 on Build GRCh38
rs41267907 Chr.3: 155138242-155138242 on Build GRCh38
rs368924551 Chr.3: 155138099-155138099 on Build GRCh38
rs34147103 Chr.3: 155138167-155138167 on Build GRCh38
rs201238968 Chr.3: 155138086-155138086 on Build GRCh38
rs758126893 Chr.3: 155138200-155138200 on Build GRCh38
rs201809149 Chr.3: 155138138-155138138 on Build GRCh38
rs572687446 Chr.3: 155138231-155138231 on Build GRCh38
rs748346111 Chr.3: 155138098-155138098 on Build GRCh38
rs775276723 Chr.3: 155138128-155138128 on Build GRCh38
rs200298222 Chr.3: 155138076-155138076 on Build GRCh38
rs375749664 Chr.3: 155138221-155138221 on Build GRCh38
rs769401228 Chr.3: 155138240-155138240 on Build GRCh38
rs199818584 Chr.3: 155138151-155138151 on Build GRCh38
rs777873470 Chr.3: 155138186-155138186 on Build GRCh38
rs762705814 Chr.3: 155138245-155138245 on Build GRCh38
rs781288611 Chr.3: 155138088-155138088 on Build GRCh38
rs747670787 Chr.3: 155138239-155138239 on Build GRCh38
rs201045219 Chr.3: 155138255-155138255 on Build GRCh38
rs554131206 Chr.3: 155138198-155138198 on Build GRCh38
rs781570758 Chr.3: 155138173-155138173 on Build GRCh38
rs760085322 Chr.3: 155138170-155138170 on Build GRCh38
rs779557823 Chr.3: 155138087-155138088 on Build GRCh38
rs763210226 Chr.3: 155138154-155138154 on Build GRCh38
rs771175787 Chr.3: 155138246-155138246 on Build GRCh38
rs752803444 Chr.3: 155138181-155138181 on Build GRCh38
rs767535205 Chr.3: 155138174-155138174 on Build GRCh38
rs145687755 Chr.3: 155138150-155138150 on Build GRCh38
rs778731154 Chr.3: 155138102-155138102 on Build GRCh38
rs200279069 Chr.3: 155138160-155138160 on Build GRCh38
rs759995584 Chr.3: 155138251-155138251 on Build GRCh38
rs141811699 Chr.3: 155138144-155138144 on Build GRCh38
rs200845545 Chr.3: 155138134-155138134 on Build GRCh38
rs199560258 Chr.3: 155138111-155138111 on Build GRCh38
rs768253967 Chr.3: 155138140-155138140 on Build GRCh38
rs746594222 Chr.3: 155138246-155138247 on Build GRCh38
rs771987826 Chr.3: 155138108-155138108 on Build GRCh38
rs768019866 Chr.3: 155138247-155138247 on Build GRCh38
rs780037500 Chr.3: 155138209-155138209 on Build GRCh38

View Full Product Details