Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 138727246-138727519 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GVQW2
Entrez Gene ID
100507462
Gene Name
GVQW motif containing 2
Gene Aliases
-
UniGene
Hs.571054
Gene Chromosome Location
Chr.6: 138725211-138773679 on Build GRCh38
Transcripts
NM_001242740.1

SNP Detail

SNP ID Chromosome Location
rs74960102 Chr.6: 138727470-138727470 on Build GRCh38
rs774908996 Chr.6: 138727488-138727488 on Build GRCh38
rs771257058 Chr.6: 138727480-138727480 on Build GRCh38
rs142320380 Chr.6: 138727402-138727402 on Build GRCh38
rs1036836123 Chr.6: 138727313-138727313 on Build GRCh38
rs1054320058 Chr.6: 138727390-138727390 on Build GRCh38
rs778023551 Chr.6: 138727321-138727321 on Build GRCh38
rs542111392 Chr.6: 138727449-138727449 on Build GRCh38
rs982092641 Chr.6: 138727370-138727370 on Build GRCh38
rs760027999 Chr.6: 138727490-138727490 on Build GRCh38
rs1050425078 Chr.6: 138727369-138727369 on Build GRCh38
rs937996120 Chr.6: 138727385-138727385 on Build GRCh38
rs754899914 Chr.6: 138727455-138727455 on Build GRCh38
rs998645855 Chr.6: 138727440-138727440 on Build GRCh38
rs539494962 Chr.6: 138727322-138727322 on Build GRCh38
rs576316082 Chr.6: 138727429-138727429 on Build GRCh38
rs771386530 Chr.6: 138727424-138727424 on Build GRCh38
rs926655594 Chr.6: 138727380-138727380 on Build GRCh38
rs959795467 Chr.6: 138727469-138727469 on Build GRCh38
rs571926805 Chr.6: 138727471-138727471 on Build GRCh38
rs1035862515 Chr.6: 138727465-138727465 on Build GRCh38
rs890729265 Chr.6: 138727442-138727442 on Build GRCh38
rs749713680 Chr.6: 138727419-138727419 on Build GRCh38
rs544632333 Chr.6: 138727292-138727292 on Build GRCh38
rs768315746 Chr.6: 138727316-138727316 on Build GRCh38
rs901871867 Chr.6: 138727320-138727320 on Build GRCh38
rs945543588 Chr.6: 138727418-138727418 on Build GRCh38
rs900191701 Chr.6: 138727478-138727478 on Build GRCh38

View Full Product Details