Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47475095-47475368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELF1
Entrez Gene ID
10658
Gene Name
CUGBP, Elav-like family member 1
Gene Aliases
BRUNOL2, CUG-BP, CUGBP, CUGBP1, EDEN-BP, NAB50, NAPOR, hNab50
UniGene
Hs.595333
Gene Chromosome Location
Chr.11: 47465937-47565569 on Build GRCh38
Transcripts
NM_001025596.2 NM_001172639.1 NM_001172640.1 NM_006560.3 NM_198700.2 XM_011519848.2 XM_011519850.2 XM_011519852.2 XM_011519854.1 XM_011519855.1 XM_011519856.1 XM_011519857.1 XM_011519858.1 XM_011519859.1 XM_017017101.1 XM_017017102.1 XM_017017103.1 XM_017017104.1 XM_017017105.1 XM_017017106.1 XM_017017107.1 XM_017017108.1 XM_017017109.1 XM_017017110.1 XM_017017111.1 XM_017017112.1 XM_017017113.1 XM_017017114.1 XM_017017115.1 XM_017017116.1 XM_017017117.1 XM_017017118.1 XM_017017119.1 XM_017017120.1 XM_017017121.1 XM_017017122.1 XM_017017123.1 XM_017017124.1 XM_017017125.1 XM_017017126.1 XM_017017127.1 XM_017017128.1 XM_017017129.1 XM_017017130.1 XM_017017131.1 XM_017017132.1 XM_017017133.1 XM_017017134.1 XM_017017135.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1604596 c.1162A>G p.I388V Chr.11: 47475363-47475363 on Build GRCh38
1604595 c.1189+9A>G p.? Chr.11: 47475327-47475327 on Build GRCh38
3666341 c.1153A>G p.I385V Chr.11: 47475363-47475363 on Build GRCh38
5685906 c.1253G>A p.G418E Chr.11: 47475350-47475350 on Build GRCh38
5685907 c.1175G>A p.G392E Chr.11: 47475350-47475350 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376310745 Chr.11: 47475300-47475300 on Build GRCh38
rs777173359 Chr.11: 47475213-47475214 on Build GRCh38
rs903949698 Chr.11: 47475217-47475217 on Build GRCh38
rs555264274 Chr.11: 47475142-47475142 on Build GRCh38
rs201883554 Chr.11: 47475304-47475304 on Build GRCh38
rs966329539 Chr.11: 47475268-47475268 on Build GRCh38
rs373715557 Chr.11: 47475319-47475319 on Build GRCh38
rs999936538 Chr.11: 47475180-47475180 on Build GRCh38
rs373919936 Chr.11: 47475298-47475298 on Build GRCh38
rs752732530 Chr.11: 47475152-47475152 on Build GRCh38
rs779438154 Chr.11: 47475276-47475276 on Build GRCh38
rs548509837 Chr.11: 47475237-47475237 on Build GRCh38
rs1045606254 Chr.11: 47475210-47475210 on Build GRCh38
rs967151511 Chr.11: 47475160-47475160 on Build GRCh38
rs1056697930 Chr.11: 47475154-47475154 on Build GRCh38
rs918137242 Chr.11: 47475179-47475179 on Build GRCh38
rs1000902260 Chr.11: 47475220-47475220 on Build GRCh38
rs529625265 Chr.11: 47475125-47475125 on Build GRCh38
rs1018882713 Chr.11: 47475264-47475264 on Build GRCh38
rs559645704 Chr.11: 47475317-47475317 on Build GRCh38
rs563087295 Chr.11: 47475281-47475281 on Build GRCh38
rs191299686 Chr.11: 47475297-47475297 on Build GRCh38
rs758846283 Chr.11: 47475331-47475331 on Build GRCh38
rs745942543 Chr.11: 47475316-47475316 on Build GRCh38
rs996034153 Chr.11: 47475272-47475272 on Build GRCh38
rs764475139 Chr.11: 47475238-47475238 on Build GRCh38
rs753963919 Chr.11: 47475301-47475301 on Build GRCh38
rs991673959 Chr.11: 47475233-47475233 on Build GRCh38
rs186512472 Chr.11: 47475267-47475267 on Build GRCh38
rs7102372 Chr.11: 47475275-47475275 on Build GRCh38
rs141604102 Chr.11: 47475194-47475194 on Build GRCh38
rs571852625 Chr.11: 47475124-47475125 on Build GRCh38
rs557346578 Chr.11: 47475243-47475243 on Build GRCh38
rs367549728 Chr.11: 47475325-47475325 on Build GRCh38
rs756959024 Chr.11: 47475298-47475299 on Build GRCh38

View Full Product Details