Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 127865019-127865286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMMECR1L
Entrez Gene ID
83607
Gene Name
AMMECR1 like
Gene Aliases
-
UniGene
Hs.546449
Gene Chromosome Location
Chr.2: 127861630-127886408 on Build GRCh38
Transcripts
NM_001199140.1 NM_031445.2 XM_005263804.4 XM_005263806.4 XM_005263807.4 XM_011511952.2 XM_011511954.2 XM_011511955.2 XM_017005446.1

Gene Symbol
LOC107985803
Entrez Gene ID
107985803
Gene Name
uncharacterized LOC107985803
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.2: 127858310-127887187 on Build GRCh38
Transcripts
NM_001199140.1 NM_031445.2 XM_005263804.4 XM_005263806.4 XM_005263807.4 XM_011511952.2 XM_011511954.2 XM_011511955.2 XM_017005446.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4591034 c.908C>T p.P303L Chr.2: 127865119-127865119 on Build GRCh38
4582641 c.891C>T p.N297N Chr.2: 127865136-127865136 on Build GRCh38
7175895 c.920A>G p.N307S Chr.2: 127865107-127865107 on Build GRCh38
4390609 c.838G>A p.V280M Chr.2: 127865189-127865189 on Build GRCh38
1182681 c.822-1G>T p.? Chr.2: 127865206-127865206 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565651686 Chr.2: 127865228-127865228 on Build GRCh38
rs781017446 Chr.2: 127865061-127865061 on Build GRCh38
rs914232071 Chr.2: 127865222-127865222 on Build GRCh38
rs755590910 Chr.2: 127865062-127865063 on Build GRCh38
rs753644879 Chr.2: 127865238-127865238 on Build GRCh38
rs755938274 Chr.2: 127865232-127865232 on Build GRCh38
rs772093306 Chr.2: 127865151-127865151 on Build GRCh38
rs376088971 Chr.2: 127865065-127865065 on Build GRCh38
rs1056424070 Chr.2: 127865116-127865116 on Build GRCh38
rs370322004 Chr.2: 127865088-127865088 on Build GRCh38
rs368275829 Chr.2: 127865221-127865221 on Build GRCh38
rs759709875 Chr.2: 127865067-127865067 on Build GRCh38
rs142938193 Chr.2: 127865136-127865136 on Build GRCh38
rs766030054 Chr.2: 127865105-127865105 on Build GRCh38
rs777254370 Chr.2: 127865175-127865175 on Build GRCh38
rs746524862 Chr.2: 127865043-127865043 on Build GRCh38
rs752396665 Chr.2: 127865120-127865120 on Build GRCh38
rs191352809 Chr.2: 127865143-127865143 on Build GRCh38
rs765978994 Chr.2: 127865189-127865189 on Build GRCh38
rs533875433 Chr.2: 127865147-127865147 on Build GRCh38
rs768027205 Chr.2: 127865068-127865068 on Build GRCh38
rs375173899 Chr.2: 127865215-127865215 on Build GRCh38
rs35686022 Chr.2: 127865048-127865049 on Build GRCh38
rs751024861 Chr.2: 127865107-127865107 on Build GRCh38
rs762185100 Chr.2: 127865183-127865183 on Build GRCh38
rs544262521 Chr.2: 127865089-127865089 on Build GRCh38
rs141599954 Chr.2: 127865245-127865245 on Build GRCh38
rs751255093 Chr.2: 127865200-127865200 on Build GRCh38
rs186978997 Chr.2: 127865064-127865064 on Build GRCh38
rs754479527 Chr.2: 127865115-127865115 on Build GRCh38
rs372518720 Chr.2: 127865050-127865050 on Build GRCh38
rs769206764 Chr.2: 127865172-127865172 on Build GRCh38
rs781157197 Chr.2: 127865118-127865118 on Build GRCh38
rs761077452 Chr.2: 127865081-127865081 on Build GRCh38
rs898309614 Chr.2: 127865202-127865202 on Build GRCh38
rs371859730 Chr.2: 127865253-127865253 on Build GRCh38
rs776050768 Chr.2: 127865155-127865155 on Build GRCh38
rs200612557 Chr.2: 127865188-127865188 on Build GRCh38
rs777868138 Chr.2: 127865234-127865234 on Build GRCh38
rs761149339 Chr.2: 127865169-127865169 on Build GRCh38
rs764420793 Chr.2: 127865082-127865082 on Build GRCh38
rs755925313 Chr.2: 127865135-127865135 on Build GRCh38
rs376646470 Chr.2: 127865063-127865063 on Build GRCh38
rs757155608 Chr.2: 127865241-127865241 on Build GRCh38
rs775901771 Chr.2: 127865071-127865071 on Build GRCh38
rs374707671 Chr.2: 127865251-127865251 on Build GRCh38
rs749102350 Chr.2: 127865140-127865140 on Build GRCh38
rs886442211 Chr.2: 127865254-127865254 on Build GRCh38

View Full Product Details