Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 38394052-38394325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMPH
Entrez Gene ID
273
Gene Name
amphiphysin
Gene Aliases
AMPH1
UniGene
Hs.592182
Gene Chromosome Location
Chr.7: 38383694-38631567 on Build GRCh38
Transcripts
NM_001635.3 NM_139316.2 XM_006715689.3 XM_006715690.3 XM_011515271.2 XM_017011995.1 XM_017011996.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3638557 c.1537G>A p.G513R Chr.7: 38394076-38394076 on Build GRCh38
1698587 c.1463C>T p.P488L Chr.7: 38394150-38394150 on Build GRCh38
387751 c.1455G>C p.E485D Chr.7: 38394158-38394158 on Build GRCh38
3638553 c.1548C>T p.T516T Chr.7: 38394065-38394065 on Build GRCh38
6599460 c.1433C>T p.A478V Chr.7: 38394054-38394054 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35785367 Chr.7: 38394254-38394255 on Build GRCh38
rs1049179766 Chr.7: 38394142-38394142 on Build GRCh38
rs775314151 Chr.7: 38394220-38394220 on Build GRCh38
rs747583340 Chr.7: 38394110-38394110 on Build GRCh38
rs762652237 Chr.7: 38394116-38394116 on Build GRCh38
rs115462490 Chr.7: 38394195-38394195 on Build GRCh38
rs773471500 Chr.7: 38394071-38394071 on Build GRCh38
rs375211709 Chr.7: 38394115-38394115 on Build GRCh38
rs749883832 Chr.7: 38394254-38394254 on Build GRCh38
rs956693302 Chr.7: 38394245-38394245 on Build GRCh38
rs969901002 Chr.7: 38394211-38394211 on Build GRCh38
rs764644221 Chr.7: 38394106-38394106 on Build GRCh38
rs371720416 Chr.7: 38394151-38394151 on Build GRCh38
rs756707743 Chr.7: 38394131-38394131 on Build GRCh38
rs748496739 Chr.7: 38394158-38394158 on Build GRCh38
rs745886763 Chr.7: 38394215-38394215 on Build GRCh38
rs960856901 Chr.7: 38394193-38394193 on Build GRCh38
rs754303145 Chr.7: 38394135-38394135 on Build GRCh38
rs145479240 Chr.7: 38394074-38394074 on Build GRCh38
rs1026021595 Chr.7: 38394206-38394206 on Build GRCh38
rs755432889 Chr.7: 38394143-38394143 on Build GRCh38
rs767119143 Chr.7: 38394244-38394244 on Build GRCh38
rs369724143 Chr.7: 38394184-38394184 on Build GRCh38
rs779390439 Chr.7: 38394155-38394155 on Build GRCh38
rs769606162 Chr.7: 38394217-38394217 on Build GRCh38
rs779630927 Chr.7: 38394148-38394148 on Build GRCh38
rs760810044 Chr.7: 38394088-38394088 on Build GRCh38
rs776438912 Chr.7: 38394213-38394213 on Build GRCh38
rs574824661 Chr.7: 38394085-38394085 on Build GRCh38
rs944540649 Chr.7: 38394297-38394297 on Build GRCh38
rs77002749 Chr.7: 38394273-38394273 on Build GRCh38
rs988722931 Chr.7: 38394270-38394270 on Build GRCh38
rs766808058 Chr.7: 38394132-38394132 on Build GRCh38
rs1017716285 Chr.7: 38394268-38394268 on Build GRCh38
rs371402056 Chr.7: 38394107-38394107 on Build GRCh38
rs147689642 Chr.7: 38394118-38394118 on Build GRCh38
rs913185015 Chr.7: 38394283-38394283 on Build GRCh38
rs755555456 Chr.7: 38394257-38394257 on Build GRCh38
rs770809668 Chr.7: 38394094-38394094 on Build GRCh38
rs374462323 Chr.7: 38394241-38394241 on Build GRCh38
rs747112662 Chr.7: 38394189-38394189 on Build GRCh38
rs73348825 Chr.7: 38394163-38394163 on Build GRCh38
rs150776580 Chr.7: 38394240-38394240 on Build GRCh38
rs1023706899 Chr.7: 38394205-38394205 on Build GRCh38
rs762695209 Chr.7: 38394229-38394229 on Build GRCh38
rs759265550 Chr.7: 38394109-38394109 on Build GRCh38
rs763758442 Chr.7: 38394237-38394237 on Build GRCh38
rs35024632 Chr.7: 38394126-38394126 on Build GRCh38

View Full Product Details