Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133768519-133768744 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VAV2
Entrez Gene ID
7410
Gene Name
vav guanine nucleotide exchange factor 2
Gene Aliases
VAV-2
UniGene
Hs.369921
Gene Chromosome Location
Chr.9: 133761894-133992374 on Build GRCh38
Transcripts
NM_001134398.1 NM_003371.3 XM_005272213.1 XM_017015108.1 XM_017015109.1 XM_017015110.1 XM_017015111.1 XM_017015112.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7146570 c.2489G>A p.R830Q Chr.9: 133768542-133768542 on Build GRCh38
5541534 c.2328C>T p.P776P Chr.9: 133768586-133768586 on Build GRCh38
5541532 c.2329C>T p.R777C Chr.9: 133768585-133768585 on Build GRCh38
7199693 c.2435T>C p.V812A Chr.9: 133768596-133768596 on Build GRCh38
7146571 c.2372G>A p.R791Q Chr.9: 133768542-133768542 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs996311247 Chr.9: 133768685-133768685 on Build GRCh38
rs773663965 Chr.9: 133768628-133768628 on Build GRCh38
rs1008824444 Chr.9: 133768625-133768625 on Build GRCh38
rs375977080 Chr.9: 133768615-133768615 on Build GRCh38
rs768225551 Chr.9: 133768614-133768614 on Build GRCh38
rs762380133 Chr.9: 133768616-133768616 on Build GRCh38
rs191274326 Chr.9: 133768585-133768585 on Build GRCh38
rs879844919 Chr.9: 133768720-133768720 on Build GRCh38
rs934946882 Chr.9: 133768564-133768564 on Build GRCh38
rs1019021441 Chr.9: 133768663-133768663 on Build GRCh38
rs371403441 Chr.9: 133768605-133768605 on Build GRCh38
rs769286360 Chr.9: 133768538-133768538 on Build GRCh38
rs772000561 Chr.9: 133768595-133768595 on Build GRCh38
rs780349466 Chr.9: 133768582-133768582 on Build GRCh38
rs148033881 Chr.9: 133768576-133768576 on Build GRCh38
rs1022083449 Chr.9: 133768700-133768700 on Build GRCh38
rs146994870 Chr.9: 133768543-133768543 on Build GRCh38
rs768188499 Chr.9: 133768626-133768626 on Build GRCh38
rs201121152 Chr.9: 133768547-133768547 on Build GRCh38
rs762302158 Chr.9: 133768606-133768606 on Build GRCh38
rs1015443117 Chr.9: 133768718-133768721 on Build GRCh38
rs752109978 Chr.9: 133768574-133768574 on Build GRCh38
rs764467885 Chr.9: 133768562-133768562 on Build GRCh38
rs964449273 Chr.9: 133768675-133768675 on Build GRCh38
rs774838611 Chr.9: 133768549-133768549 on Build GRCh38
rs367561487 Chr.9: 133768598-133768598 on Build GRCh38
rs564689902 Chr.9: 133768590-133768590 on Build GRCh38
rs978997426 Chr.9: 133768557-133768557 on Build GRCh38
rs766448136 Chr.9: 133768633-133768633 on Build GRCh38
rs202055327 Chr.9: 133768584-133768584 on Build GRCh38
rs968898839 Chr.9: 133768556-133768556 on Build GRCh38
rs749792252 Chr.9: 133768672-133768672 on Build GRCh38
rs544606817 Chr.9: 133768644-133768644 on Build GRCh38
rs143397192 Chr.9: 133768583-133768583 on Build GRCh38
rs61751477 Chr.9: 133768577-133768577 on Build GRCh38
rs746931943 Chr.9: 133768588-133768588 on Build GRCh38
rs762544581 Chr.9: 133768559-133768559 on Build GRCh38
rs200140238 Chr.9: 133768589-133768589 on Build GRCh38
rs761155809 Chr.9: 133768630-133768630 on Build GRCh38
rs374240487 Chr.9: 133768542-133768542 on Build GRCh38
rs201135054 Chr.9: 133768561-133768561 on Build GRCh38
rs755209800 Chr.9: 133768645-133768645 on Build GRCh38

View Full Product Details