Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1271868-1272141 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIRBP
Entrez Gene ID
1153
Gene Name
cold inducible RNA binding protein
Gene Aliases
CIRP
UniGene
Hs.618145
Gene Chromosome Location
Chr.19: 1269266-1274810 on Build GRCh38
Transcripts
NM_001280.2 NM_001300815.1 NM_001300829.1 NR_023312.2 NR_023313.2 XM_006722637.1 XM_011527668.1 XM_017026237.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6316021 c.273-4G>T p.? Chr.19: 1271977-1271977 on Build GRCh38
6900653 c.296G>T p.S99I Chr.19: 1272004-1272004 on Build GRCh38
2816049 c.363C>T p.G121G Chr.19: 1272071-1272071 on Build GRCh38
438608 c.493G>A p.D165N Chr.19: 1272042-1272042 on Build GRCh38
6900652 c.455G>T p.S152I Chr.19: 1272004-1272004 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757038782 Chr.19: 1272094-1272094 on Build GRCh38
rs1053681916 Chr.19: 1272113-1272113 on Build GRCh38
rs756499852 Chr.19: 1271960-1271960 on Build GRCh38
rs770547263 Chr.19: 1271924-1271924 on Build GRCh38
rs757896292 Chr.19: 1272071-1272072 on Build GRCh38
rs768866584 Chr.19: 1272118-1272118 on Build GRCh38
rs750904041 Chr.19: 1272020-1272020 on Build GRCh38
rs1025747468 Chr.19: 1271927-1271927 on Build GRCh38
rs768995229 Chr.19: 1272063-1272063 on Build GRCh38
rs753456240 Chr.19: 1272092-1272092 on Build GRCh38
rs747818399 Chr.19: 1271970-1271970 on Build GRCh38
rs112823184 Chr.19: 1272052-1272052 on Build GRCh38
rs961624531 Chr.19: 1271909-1271909 on Build GRCh38
rs754193131 Chr.19: 1271963-1271963 on Build GRCh38
rs769587596 Chr.19: 1271942-1271942 on Build GRCh38
rs934857030 Chr.19: 1272109-1272109 on Build GRCh38
rs755630032 Chr.19: 1272086-1272086 on Build GRCh38
rs369487902 Chr.19: 1271954-1271954 on Build GRCh38
rs766932411 Chr.19: 1272083-1272083 on Build GRCh38
rs527705089 Chr.19: 1272071-1272071 on Build GRCh38
rs372044669 Chr.19: 1272060-1272060 on Build GRCh38
rs375230632 Chr.19: 1271949-1271949 on Build GRCh38
rs987421199 Chr.19: 1271923-1271923 on Build GRCh38
rs768338025 Chr.19: 1271932-1271932 on Build GRCh38
rs762176412 Chr.19: 1272066-1272066 on Build GRCh38
rs35810179 Chr.19: 1272063-1272064 on Build GRCh38
rs762919208 Chr.19: 1271948-1271948 on Build GRCh38
rs747014591 Chr.19: 1272117-1272117 on Build GRCh38
rs749098190 Chr.19: 1271974-1271974 on Build GRCh38
rs765705409 Chr.19: 1272019-1272019 on Build GRCh38
rs372805481 Chr.19: 1272085-1272085 on Build GRCh38
rs775812654 Chr.19: 1272011-1272011 on Build GRCh38
rs978820784 Chr.19: 1272087-1272087 on Build GRCh38
rs749046630 Chr.19: 1272042-1272042 on Build GRCh38
rs781184576 Chr.19: 1271917-1271917 on Build GRCh38
rs777525229 Chr.19: 1271973-1271973 on Build GRCh38
rs769478304 Chr.19: 1271972-1271972 on Build GRCh38
rs773096473 Chr.19: 1271943-1271943 on Build GRCh38
rs972683691 Chr.19: 1271910-1271910 on Build GRCh38
rs745826788 Chr.19: 1272051-1272051 on Build GRCh38
rs374893846 Chr.19: 1272017-1272017 on Build GRCh38
rs772302756 Chr.19: 1272009-1272009 on Build GRCh38
rs772174152 Chr.19: 1272058-1272058 on Build GRCh38
rs1022618245 Chr.19: 1271996-1271996 on Build GRCh38
rs1034525536 Chr.19: 1271888-1271888 on Build GRCh38
rs750899451 Chr.19: 1272072-1272072 on Build GRCh38
rs367647466 Chr.19: 1272080-1272080 on Build GRCh38
rs552567151 Chr.19: 1272091-1272091 on Build GRCh38
rs200903901 Chr.19: 1272062-1272062 on Build GRCh38
rs747867699 Chr.19: 1271939-1271939 on Build GRCh38
rs752257288 Chr.19: 1272035-1272035 on Build GRCh38
rs1016764557 Chr.19: 1271891-1271891 on Build GRCh38
rs764458728 Chr.19: 1272014-1272014 on Build GRCh38
rs374975169 Chr.19: 1272054-1272054 on Build GRCh38
rs192860791 Chr.19: 1271968-1271968 on Build GRCh38
rs752983394 Chr.19: 1271958-1271958 on Build GRCh38
rs79321414 Chr.19: 1272012-1272012 on Build GRCh38
rs776045173 Chr.19: 1271934-1271934 on Build GRCh38
rs529222296 Chr.19: 1272010-1272010 on Build GRCh38
rs780762874 Chr.19: 1271969-1271969 on Build GRCh38
rs780652614 Chr.19: 1272032-1272032 on Build GRCh38
rs780207074 Chr.19: 1272115-1272115 on Build GRCh38
rs774160854 Chr.19: 1271984-1271984 on Build GRCh38
rs910546982 Chr.19: 1272076-1272076 on Build GRCh38
rs755799040 Chr.19: 1271890-1271890 on Build GRCh38
rs778087610 Chr.19: 1272103-1272103 on Build GRCh38
rs754370762 Chr.19: 1271967-1271970 on Build GRCh38
rs564576371 Chr.19: 1272110-1272110 on Build GRCh38
rs778542942 Chr.19: 1272102-1272102 on Build GRCh38
rs759604030 Chr.19: 1272001-1272001 on Build GRCh38
rs776718444 Chr.19: 1272065-1272065 on Build GRCh38
rs369012420 Chr.19: 1272018-1272018 on Build GRCh38
rs757759426 Chr.19: 1271964-1271964 on Build GRCh38
rs774480816 Chr.19: 1271953-1271953 on Build GRCh38
rs943455814 Chr.19: 1271925-1271925 on Build GRCh38
rs763431767 Chr.19: 1272077-1272077 on Build GRCh38
rs12488 Chr.19: 1272041-1272041 on Build GRCh38
rs776328913 Chr.19: 1271935-1271935 on Build GRCh38
rs145902755 Chr.19: 1272050-1272050 on Build GRCh38
rs933349964 Chr.19: 1271921-1271921 on Build GRCh38
rs376153221 Chr.19: 1272021-1272021 on Build GRCh38
rs767783002 Chr.19: 1271956-1271956 on Build GRCh38
rs779346537 Chr.19: 1271965-1271965 on Build GRCh38
rs770900481 Chr.19: 1271982-1271982 on Build GRCh38
rs372289434 Chr.19: 1271930-1271930 on Build GRCh38
rs770646297 Chr.19: 1272046-1272046 on Build GRCh38

View Full Product Details