Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 31638603-31638864 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALT6
Entrez Gene ID
9331
Gene Name
beta-1,4-galactosyltransferase 6
Gene Aliases
B4Gal-T6, beta4Gal-T6
UniGene
Hs.591063
Gene Chromosome Location
Chr.18: 31622246-31724670 on Build GRCh38
Transcripts
NM_004775.3 XM_005258387.4 XM_006722578.3 XM_006722579.3 XM_017026088.1 XM_017026089.1 XM_017026090.1 XM_017026091.1 XM_017026092.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4571707 c.520T>G p.F174V Chr.18: 31638712-31638712 on Build GRCh38
1303676 c.556C>T p.R186W Chr.18: 31638676-31638676 on Build GRCh38
4840004 c.481C>A p.L161I Chr.18: 31638751-31638751 on Build GRCh38
3362490 c.570G>A p.A190A Chr.18: 31638662-31638662 on Build GRCh38
3783137 c.583G>T p.E195* Chr.18: 31638649-31638649 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017734174 Chr.18: 31638822-31638822 on Build GRCh38
rs1027486777 Chr.18: 31638786-31638789 on Build GRCh38
rs115568058 Chr.18: 31638730-31638730 on Build GRCh38
rs371621631 Chr.18: 31638739-31638739 on Build GRCh38
rs199629405 Chr.18: 31638755-31638755 on Build GRCh38
rs765525965 Chr.18: 31638691-31638691 on Build GRCh38
rs905199126 Chr.18: 31638719-31638719 on Build GRCh38
rs754031469 Chr.18: 31638677-31638677 on Build GRCh38
rs778674527 Chr.18: 31638635-31638636 on Build GRCh38
rs199540062 Chr.18: 31638697-31638697 on Build GRCh38
rs771949169 Chr.18: 31638707-31638708 on Build GRCh38
rs537591211 Chr.18: 31638814-31638814 on Build GRCh38
rs929225470 Chr.18: 31638703-31638703 on Build GRCh38
rs747837860 Chr.18: 31638641-31638642 on Build GRCh38
rs1056400957 Chr.18: 31638735-31638735 on Build GRCh38
rs368163743 Chr.18: 31638715-31638715 on Build GRCh38
rs761277184 Chr.18: 31638638-31638638 on Build GRCh38
rs758589504 Chr.18: 31638662-31638662 on Build GRCh38
rs754961111 Chr.18: 31638817-31638817 on Build GRCh38
rs768391076 Chr.18: 31638783-31638783 on Build GRCh38
rs944568897 Chr.18: 31638779-31638779 on Build GRCh38
rs368503724 Chr.18: 31638784-31638784 on Build GRCh38
rs116995051 Chr.18: 31638806-31638806 on Build GRCh38
rs755190376 Chr.18: 31638686-31638686 on Build GRCh38
rs768057247 Chr.18: 31638656-31638656 on Build GRCh38
rs777727143 Chr.18: 31638780-31638780 on Build GRCh38
rs760954653 Chr.18: 31638663-31638663 on Build GRCh38
rs180688911 Chr.18: 31638733-31638733 on Build GRCh38
rs766710771 Chr.18: 31638669-31638669 on Build GRCh38
rs920480754 Chr.18: 31638674-31638674 on Build GRCh38
rs747726560 Chr.18: 31638795-31638795 on Build GRCh38
rs748921281 Chr.18: 31638764-31638764 on Build GRCh38
rs915348123 Chr.18: 31638675-31638675 on Build GRCh38

View Full Product Details