Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 43645967-43646240 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF32
Entrez Gene ID
7580
Gene Name
zinc finger protein 32
Gene Aliases
KOX30
UniGene
Hs.522885
Gene Chromosome Location
Chr.10: 43643859-43649350 on Build GRCh38
Transcripts
NM_001005368.2 NM_001324164.1 NM_001324165.1 NM_001324166.1 NM_001324167.1 NM_001324168.1 NM_006973.2 XM_017016616.1 NR_047558.1 NR_038867.1

Gene Symbol
ZNF32-AS2
Entrez Gene ID
414208
Gene Name
ZNF32 antisense RNA 2
Gene Aliases
ZNF32OS2, b402L1.7
UniGene
Hs.608956
Gene Chromosome Location
Chr.10: 43645942-43648019 on Build GRCh38
Transcripts
NM_001005368.2 NM_001324164.1 NM_001324165.1 NM_001324166.1 NM_001324167.1 NM_001324168.1 NM_006973.2 XM_017016616.1 NR_047558.1 NR_038867.1

Gene Symbol
ZNF32-AS3
Entrez Gene ID
414201
Gene Name
ZNF32 antisense RNA 3
Gene Aliases
C10orf44, ZNF32-OT1, bA402L1.3
UniGene
Hs.666576
Gene Chromosome Location
Chr.10: 43628817-43674699 on Build GRCh38
Transcripts
NM_001005368.2 NM_001324164.1 NM_001324165.1 NM_001324166.1 NM_001324167.1 NM_001324168.1 NM_006973.2 XM_017016616.1 NR_047558.1 NR_038867.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6220531 c.70+7G>T p.? Chr.10: 43646057-43646057 on Build GRCh38
4619948 c.24C>A p.T8T Chr.10: 43646110-43646110 on Build GRCh38
918219 c.40G>T p.G14* Chr.10: 43646094-43646094 on Build GRCh38
6829322 c.62C>T p.A21V Chr.10: 43646072-43646072 on Build GRCh38
918220 c.26delT p.L9fs*12 Chr.10: 43646108-43646108 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs888528989 Chr.10: 43646195-43646195 on Build GRCh38
rs547298681 Chr.10: 43646110-43646110 on Build GRCh38
rs766395173 Chr.10: 43646032-43646032 on Build GRCh38
rs779391269 Chr.10: 43646076-43646076 on Build GRCh38
rs780715010 Chr.10: 43646096-43646096 on Build GRCh38
rs114834062 Chr.10: 43646071-43646071 on Build GRCh38
rs758705520 Chr.10: 43646095-43646095 on Build GRCh38
rs746290159 Chr.10: 43646086-43646086 on Build GRCh38
rs771058214 Chr.10: 43646139-43646139 on Build GRCh38
rs749531405 Chr.10: 43646044-43646044 on Build GRCh38
rs116698198 Chr.10: 43646035-43646035 on Build GRCh38
rs780337400 Chr.10: 43645991-43645991 on Build GRCh38
rs773056481 Chr.10: 43646019-43646019 on Build GRCh38
rs966997902 Chr.10: 43646064-43646064 on Build GRCh38
rs184512649 Chr.10: 43646036-43646036 on Build GRCh38
rs769282680 Chr.10: 43646017-43646017 on Build GRCh38
rs753842608 Chr.10: 43646068-43646068 on Build GRCh38
rs986036278 Chr.10: 43646066-43646066 on Build GRCh38
rs200854599 Chr.10: 43646171-43646171 on Build GRCh38
rs764191203 Chr.10: 43646057-43646057 on Build GRCh38
rs764444272 Chr.10: 43646175-43646175 on Build GRCh38
rs370602012 Chr.10: 43646135-43646135 on Build GRCh38
rs771924654 Chr.10: 43646173-43646173 on Build GRCh38
rs372743050 Chr.10: 43645999-43645999 on Build GRCh38
rs889580328 Chr.10: 43646007-43646007 on Build GRCh38
rs199726414 Chr.10: 43646149-43646149 on Build GRCh38
rs141643182 Chr.10: 43646034-43646034 on Build GRCh38
rs990563375 Chr.10: 43646170-43646170 on Build GRCh38
rs775603506 Chr.10: 43646185-43646185 on Build GRCh38
rs762898318 Chr.10: 43646024-43646024 on Build GRCh38
rs868283381 Chr.10: 43646192-43646192 on Build GRCh38
rs763203351 Chr.10: 43646112-43646113 on Build GRCh38
rs747797677 Chr.10: 43646108-43646108 on Build GRCh38
rs747625265 Chr.10: 43646015-43646015 on Build GRCh38
rs773824109 Chr.10: 43646131-43646131 on Build GRCh38
rs748776459 Chr.10: 43646137-43646137 on Build GRCh38
rs752838481 Chr.10: 43646053-43646054 on Build GRCh38
rs747732006 Chr.10: 43646014-43646014 on Build GRCh38

View Full Product Details