Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 5122487-5122724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL8B
Entrez Gene ID
55207
Gene Name
ADP ribosylation factor like GTPase 8B
Gene Aliases
ARL10C, Gie1
UniGene
Hs.250009
Gene Chromosome Location
Chr.3: 5122245-5180916 on Build GRCh38
Transcripts
NM_018184.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6285514 c.36C>T p.F12F Chr.3: 5122501-5122501 on Build GRCh38
2851970 c.29A>C p.D10A Chr.3: 5122494-5122494 on Build GRCh38
5974858 c.55G>A p.E19K Chr.3: 5122520-5122520 on Build GRCh38
5927272 c.64G>A p.E22K Chr.3: 5122529-5122529 on Build GRCh38
2851969 c.29A>C p.D10A Chr.3: 5122494-5122494 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770128292 Chr.3: 5122615-5122615 on Build GRCh38
rs1037472490 Chr.3: 5122668-5122668 on Build GRCh38
rs113092227 Chr.3: 5122629-5122629 on Build GRCh38
rs941967075 Chr.3: 5122654-5122654 on Build GRCh38
rs1052470352 Chr.3: 5122672-5122672 on Build GRCh38
rs941788970 Chr.3: 5122646-5122646 on Build GRCh38
rs376563141 Chr.3: 5122624-5122624 on Build GRCh38
rs776479234 Chr.3: 5122585-5122585 on Build GRCh38
rs983313513 Chr.3: 5122643-5122643 on Build GRCh38
rs781615253 Chr.3: 5122535-5122535 on Build GRCh38
rs765256373 Chr.3: 5122596-5122596 on Build GRCh38
rs763093973 Chr.3: 5122649-5122649 on Build GRCh38
rs755495217 Chr.3: 5122522-5122522 on Build GRCh38
rs755336760 Chr.3: 5122614-5122614 on Build GRCh38
rs760456879 Chr.3: 5122579-5122579 on Build GRCh38
rs894974163 Chr.3: 5122670-5122670 on Build GRCh38
rs369029104 Chr.3: 5122598-5122598 on Build GRCh38
rs761829672 Chr.3: 5122594-5122594 on Build GRCh38
rs563203518 Chr.3: 5122644-5122644 on Build GRCh38
rs980482280 Chr.3: 5122633-5122633 on Build GRCh38
rs143079086 Chr.3: 5122567-5122567 on Build GRCh38
rs543512493 Chr.3: 5122641-5122641 on Build GRCh38
rs906194658 Chr.3: 5122688-5122688 on Build GRCh38
rs574644105 Chr.3: 5122638-5122638 on Build GRCh38
rs1022230258 Chr.3: 5122558-5122558 on Build GRCh38
rs1046605328 Chr.3: 5122685-5122685 on Build GRCh38
rs745433923 Chr.3: 5122627-5122627 on Build GRCh38
rs180728253 Chr.3: 5122704-5122704 on Build GRCh38
rs763102911 Chr.3: 5122600-5122600 on Build GRCh38
rs756694097 Chr.3: 5122621-5122621 on Build GRCh38
rs990344720 Chr.3: 5122548-5122548 on Build GRCh38
rs141649171 Chr.3: 5122582-5122582 on Build GRCh38
rs748811554 Chr.3: 5122538-5122538 on Build GRCh38
rs947796098 Chr.3: 5122679-5122679 on Build GRCh38
rs758122158 Chr.3: 5122628-5122628 on Build GRCh38
rs771867776 Chr.3: 5122566-5122566 on Build GRCh38
rs999213538 Chr.3: 5122705-5122705 on Build GRCh38
rs933564815 Chr.3: 5122668-5122669 on Build GRCh38
rs372971404 Chr.3: 5122606-5122606 on Build GRCh38
rs745448271 Chr.3: 5122546-5122546 on Build GRCh38
rs776516674 Chr.3: 5122636-5122636 on Build GRCh38
rs768285246 Chr.3: 5122631-5122631 on Build GRCh38
rs975242610 Chr.3: 5122608-5122608 on Build GRCh38
rs751884170 Chr.3: 5122510-5122510 on Build GRCh38
rs147122055 Chr.3: 5122540-5122540 on Build GRCh38
rs1036500224 Chr.3: 5122663-5122663 on Build GRCh38

View Full Product Details