Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 45423960-45424231 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALOX5
Entrez Gene ID
240
Gene Name
arachidonate 5-lipoxygenase
Gene Aliases
5-LO, 5-LOX, 5LPG, LOG5
UniGene
Hs.89499
Gene Chromosome Location
Chr.10: 45374166-45446121 on Build GRCh38
Transcripts
NM_000698.4 NM_001256153.2 NM_001256154.2 NM_001320861.1 NM_001320862.1 XM_017016012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
918271 c.633C>A p.I211I Chr.10: 45424119-45424119 on Build GRCh38
7141286 c.639C>A p.V213V Chr.10: 45424125-45424125 on Build GRCh38
388924 c.619G>A p.D207N Chr.10: 45424105-45424105 on Build GRCh38
4502009 c.602C>T p.S201F Chr.10: 45424088-45424088 on Build GRCh38
6408345 c.605G>T p.W202L Chr.10: 45424091-45424091 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs188147236 Chr.10: 45423983-45423983 on Build GRCh38
rs913111764 Chr.10: 45424058-45424058 on Build GRCh38
rs201769753 Chr.10: 45424102-45424102 on Build GRCh38
rs768993639 Chr.10: 45424186-45424187 on Build GRCh38
rs754743085 Chr.10: 45424160-45424160 on Build GRCh38
rs776116374 Chr.10: 45424016-45424017 on Build GRCh38
rs535317736 Chr.10: 45424158-45424158 on Build GRCh38
rs764750532 Chr.10: 45424056-45424056 on Build GRCh38
rs41429645 Chr.10: 45424168-45424168 on Build GRCh38
rs781279595 Chr.10: 45424087-45424087 on Build GRCh38
rs962688038 Chr.10: 45424133-45424133 on Build GRCh38
rs372041946 Chr.10: 45424146-45424146 on Build GRCh38
rs143505389 Chr.10: 45424104-45424104 on Build GRCh38
rs754803013 Chr.10: 45424076-45424076 on Build GRCh38
rs766374033 Chr.10: 45424155-45424155 on Build GRCh38
rs746334217 Chr.10: 45424199-45424199 on Build GRCh38
rs748345682 Chr.10: 45424088-45424088 on Build GRCh38
rs762438808 Chr.10: 45424140-45424140 on Build GRCh38
rs774992398 Chr.10: 45424110-45424110 on Build GRCh38
rs375277942 Chr.10: 45424162-45424162 on Build GRCh38
rs761482218 Chr.10: 45424128-45424128 on Build GRCh38
rs371097552 Chr.10: 45424123-45424123 on Build GRCh38
rs757339018 Chr.10: 45424188-45424188 on Build GRCh38
rs752896072 Chr.10: 45424166-45424166 on Build GRCh38
rs749765877 Chr.10: 45424007-45424007 on Build GRCh38
rs773870077 Chr.10: 45423999-45423999 on Build GRCh38
rs751434862 Chr.10: 45424156-45424156 on Build GRCh38
rs953551723 Chr.10: 45424029-45424029 on Build GRCh38
rs1000412984 Chr.10: 45424000-45424000 on Build GRCh38
rs759299348 Chr.10: 45424050-45424050 on Build GRCh38
rs912015383 Chr.10: 45424037-45424037 on Build GRCh38
rs570065880 Chr.10: 45424051-45424051 on Build GRCh38
rs201548620 Chr.10: 45424181-45424181 on Build GRCh38
rs903552817 Chr.10: 45423986-45423986 on Build GRCh38
rs758369099 Chr.10: 45424064-45424064 on Build GRCh38
rs776279650 Chr.10: 45424047-45424047 on Build GRCh38
rs776167459 Chr.10: 45424126-45424126 on Build GRCh38
rs774904363 Chr.10: 45424012-45424012 on Build GRCh38
rs751416399 Chr.10: 45424069-45424069 on Build GRCh38
rs374707016 Chr.10: 45424010-45424010 on Build GRCh38
rs1015109299 Chr.10: 45424196-45424196 on Build GRCh38
rs568327472 Chr.10: 45424138-45424138 on Build GRCh38
rs372401060 Chr.10: 45424013-45424013 on Build GRCh38
rs899974578 Chr.10: 45424043-45424043 on Build GRCh38
rs945139859 Chr.10: 45424191-45424191 on Build GRCh38
rs943855317 Chr.10: 45424053-45424053 on Build GRCh38
rs199618529 Chr.10: 45424173-45424173 on Build GRCh38
rs372321818 Chr.10: 45424197-45424197 on Build GRCh38
rs1014421020 Chr.10: 45424185-45424185 on Build GRCh38
rs764836044 Chr.10: 45424136-45424136 on Build GRCh38
rs771622355 Chr.10: 45424105-45424105 on Build GRCh38
rs141077174 Chr.10: 45424101-45424101 on Build GRCh38
rs746223035 Chr.10: 45424122-45424122 on Build GRCh38
rs766285713 Chr.10: 45424065-45424065 on Build GRCh38
rs763720759 Chr.10: 45424036-45424036 on Build GRCh38

View Full Product Details