Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1387166-1387413 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNL2
Entrez Gene ID
81669
Gene Name
cyclin L2
Gene Aliases
ANIA-6B, CCNM, CCNS, HCLA-ISO, HLA-ISO, PCEE, SB138
UniGene
Hs.515704
Gene Chromosome Location
Chr.1: 1385711-1399338 on Build GRCh38
Transcripts
NM_001320153.1 NM_001320155.1 NM_030937.4 NR_135154.1 XM_011542216.2 XM_011542219.2 XM_011542221.2 XM_017002420.1 XM_017002421.1 XM_017002422.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4670246 c.1499C>T p.S500L Chr.1: 1387295-1387295 on Build GRCh38
3676725 c.1438C>T p.P480S Chr.1: 1387356-1387356 on Build GRCh38
894964 c.1390C>T p.R464W Chr.1: 1387404-1387404 on Build GRCh38
5682323 c.1422G>T p.R474R Chr.1: 1387372-1387372 on Build GRCh38
5088632 c.1520G>A p.R507H Chr.1: 1387274-1387274 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367605273 Chr.1: 1387310-1387310 on Build GRCh38
rs971477940 Chr.1: 1387334-1387334 on Build GRCh38
rs781642985 Chr.1: 1387308-1387308 on Build GRCh38
rs766883143 Chr.1: 1387301-1387301 on Build GRCh38
rs777021721 Chr.1: 1387338-1387339 on Build GRCh38
rs753544578 Chr.1: 1387238-1387238 on Build GRCh38
rs955471142 Chr.1: 1387371-1387371 on Build GRCh38
rs996794721 Chr.1: 1387223-1387223 on Build GRCh38
rs143282720 Chr.1: 1387364-1387364 on Build GRCh38
rs772751069 Chr.1: 1387262-1387262 on Build GRCh38
rs765003761 Chr.1: 1387351-1387351 on Build GRCh38
rs775543411 Chr.1: 1387313-1387313 on Build GRCh38
rs771424119 Chr.1: 1387268-1387268 on Build GRCh38
rs571709892 Chr.1: 1387292-1387292 on Build GRCh38
rs759465636 Chr.1: 1387214-1387214 on Build GRCh38
rs754703350 Chr.1: 1387294-1387294 on Build GRCh38
rs756441361 Chr.1: 1387362-1387362 on Build GRCh38
rs776288996 Chr.1: 1387375-1387375 on Build GRCh38
rs766061370 Chr.1: 1387345-1387345 on Build GRCh38
rs759093167 Chr.1: 1387349-1387349 on Build GRCh38
rs776657329 Chr.1: 1387269-1387269 on Build GRCh38
rs764125593 Chr.1: 1387332-1387332 on Build GRCh38
rs758966735 Chr.1: 1387300-1387300 on Build GRCh38
rs751403151 Chr.1: 1387219-1387219 on Build GRCh38
rs754399672 Chr.1: 1387239-1387239 on Build GRCh38
rs369182174 Chr.1: 1387391-1387391 on Build GRCh38
rs146506266 Chr.1: 1387355-1387355 on Build GRCh38
rs779234010 Chr.1: 1387188-1387188 on Build GRCh38
rs761101143 Chr.1: 1387244-1387244 on Build GRCh38
rs778265882 Chr.1: 1387265-1387265 on Build GRCh38
rs750929165 Chr.1: 1387204-1387204 on Build GRCh38
rs770113121 Chr.1: 1387311-1387311 on Build GRCh38
rs370956539 Chr.1: 1387275-1387275 on Build GRCh38
rs767510815 Chr.1: 1387278-1387278 on Build GRCh38
rs992014718 Chr.1: 1387210-1387210 on Build GRCh38
rs771334089 Chr.1: 1387206-1387206 on Build GRCh38
rs151152085 Chr.1: 1387274-1387274 on Build GRCh38
rs776766347 Chr.1: 1387208-1387208 on Build GRCh38
rs775235841 Chr.1: 1387392-1387392 on Build GRCh38
rs935261823 Chr.1: 1387386-1387386 on Build GRCh38
rs369853322 Chr.1: 1387335-1387335 on Build GRCh38
rs919388447 Chr.1: 1387229-1387229 on Build GRCh38
rs201094209 Chr.1: 1387221-1387221 on Build GRCh38
rs752309244 Chr.1: 1387298-1387298 on Build GRCh38
rs577309284 Chr.1: 1387198-1387198 on Build GRCh38
rs958984651 Chr.1: 1387323-1387323 on Build GRCh38
rs964591616 Chr.1: 1387338-1387338 on Build GRCh38
rs746311825 Chr.1: 1387361-1387361 on Build GRCh38
rs369070554 Chr.1: 1387368-1387368 on Build GRCh38
rs372195187 Chr.1: 1387354-1387354 on Build GRCh38
rs779508370 Chr.1: 1387228-1387228 on Build GRCh38
rs750278205 Chr.1: 1387222-1387222 on Build GRCh38
rs534775954 Chr.1: 1387286-1387286 on Build GRCh38
rs759826135 Chr.1: 1387271-1387271 on Build GRCh38
rs139865828 Chr.1: 1387393-1387393 on Build GRCh38
rs139345631 Chr.1: 1387356-1387356 on Build GRCh38
rs201055911 Chr.1: 1387385-1387385 on Build GRCh38
rs749300030 Chr.1: 1387314-1387314 on Build GRCh38
rs200276576 Chr.1: 1387374-1387374 on Build GRCh38
rs1053586852 Chr.1: 1387284-1387284 on Build GRCh38
rs778260594 Chr.1: 1387295-1387295 on Build GRCh38
rs747441825 Chr.1: 1387266-1387266 on Build GRCh38
rs768738321 Chr.1: 1387325-1387325 on Build GRCh38
rs144406967 Chr.1: 1387373-1387373 on Build GRCh38
rs374382413 Chr.1: 1387277-1387277 on Build GRCh38
rs757513204 Chr.1: 1387304-1387304 on Build GRCh38
rs780193325 Chr.1: 1387363-1387363 on Build GRCh38
rs151330631 Chr.1: 1387287-1387287 on Build GRCh38
rs879009298 Chr.1: 1387305-1387305 on Build GRCh38
rs113568283 Chr.1: 1387370-1387370 on Build GRCh38
rs773747677 Chr.1: 1387205-1387205 on Build GRCh38
rs771568690 Chr.1: 1387306-1387306 on Build GRCh38
rs967275822 Chr.1: 1387187-1387187 on Build GRCh38
rs371063310 Chr.1: 1387195-1387195 on Build GRCh38
rs951620392 Chr.1: 1387324-1387324 on Build GRCh38
rs113811513 Chr.1: 1387227-1387227 on Build GRCh38
rs748639427 Chr.1: 1387255-1387255 on Build GRCh38

View Full Product Details