Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 99850303-99850538 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMSS1
Entrez Gene ID
84319
Gene Name
cms1 ribosomal small subunit homolog (yeast)
Gene Aliases
C3orf26
UniGene
Hs.581320
Gene Chromosome Location
Chr.3: 99817834-100178632 on Build GRCh38
Transcripts
NM_032359.3 NM_001042459.2 NM_001282793.1 NM_001282794.1 NM_014890.3 NM_182909.3 XM_006713486.2 XM_011512371.2

Gene Symbol
FILIP1L
Entrez Gene ID
11259
Gene Name
filamin A interacting protein 1 like
Gene Aliases
DOC-1, DOC1, GIP130, GIP90
UniGene
Hs.104672
Gene Chromosome Location
Chr.3: 99833144-100114513 on Build GRCh38
Transcripts
NM_032359.3 NM_001042459.2 NM_001282793.1 NM_001282794.1 NM_014890.3 NM_182909.3 XM_006713486.2 XM_011512371.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
281170 c.1313A>C p.K438T Chr.3: 99850363-99850363 on Build GRCh38
1243269 c.1297G>A p.A433T Chr.3: 99850379-99850379 on Build GRCh38
3598484 c.1203G>A p.K401K Chr.3: 99850473-99850473 on Build GRCh38
4876196 c.1310G>C p.S437T Chr.3: 99850366-99850366 on Build GRCh38
3598480 c.1355G>A p.R452K Chr.3: 99850321-99850321 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747628768 Chr.3: 99850337-99850337 on Build GRCh38
rs867416827 Chr.3: 99850449-99850449 on Build GRCh38
rs985031788 Chr.3: 99850428-99850428 on Build GRCh38
rs1055606998 Chr.3: 99850435-99850435 on Build GRCh38
rs773217518 Chr.3: 99850402-99850402 on Build GRCh38
rs773071545 Chr.3: 99850496-99850496 on Build GRCh38
rs368967496 Chr.3: 99850379-99850379 on Build GRCh38
rs9828782 Chr.3: 99850344-99850344 on Build GRCh38
rs537419810 Chr.3: 99850430-99850430 on Build GRCh38
rs569046690 Chr.3: 99850340-99850340 on Build GRCh38
rs747972266 Chr.3: 99850431-99850435 on Build GRCh38
rs769884665 Chr.3: 99850376-99850376 on Build GRCh38
rs200386812 Chr.3: 99850457-99850457 on Build GRCh38
rs558704957 Chr.3: 99850437-99850437 on Build GRCh38
rs767619818 Chr.3: 99850391-99850391 on Build GRCh38
rs770746244 Chr.3: 99850501-99850501 on Build GRCh38
rs753555396 Chr.3: 99850425-99850425 on Build GRCh38
rs746186263 Chr.3: 99850468-99850468 on Build GRCh38
rs746880528 Chr.3: 99850497-99850497 on Build GRCh38
rs200354760 Chr.3: 99850456-99850456 on Build GRCh38
rs749384433 Chr.3: 99850482-99850482 on Build GRCh38
rs773046692 Chr.3: 99850490-99850490 on Build GRCh38
rs757426039 Chr.3: 99850443-99850444 on Build GRCh38
rs779155962 Chr.3: 99850484-99850484 on Build GRCh38
rs757849862 Chr.3: 99850450-99850450 on Build GRCh38
rs768817482 Chr.3: 99850483-99850483 on Build GRCh38
rs776747155 Chr.3: 99850353-99850353 on Build GRCh38
rs368410003 Chr.3: 99850469-99850469 on Build GRCh38
rs752265612 Chr.3: 99850447-99850447 on Build GRCh38
rs201901524 Chr.3: 99850477-99850477 on Build GRCh38
rs771256008 Chr.3: 99850351-99850351 on Build GRCh38
rs764868770 Chr.3: 99850444-99850444 on Build GRCh38
rs943040073 Chr.3: 99850452-99850452 on Build GRCh38
rs760419965 Chr.3: 99850405-99850405 on Build GRCh38
rs538097692 Chr.3: 99850455-99850455 on Build GRCh38
rs934398420 Chr.3: 99850475-99850475 on Build GRCh38
rs763119776 Chr.3: 99850383-99850383 on Build GRCh38
rs1037851181 Chr.3: 99850386-99850386 on Build GRCh38
rs775751047 Chr.3: 99850380-99850380 on Build GRCh38
rs746050062 Chr.3: 99850365-99850365 on Build GRCh38
rs778252258 Chr.3: 99850454-99850454 on Build GRCh38
rs747343421 Chr.3: 99850348-99850348 on Build GRCh38

View Full Product Details