Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 5034212-5034445 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC52A1
Entrez Gene ID
55065
Gene Name
solute carrier family 52 member 1
Gene Aliases
GPCR42, GPR172B, PAR2, RBFVD, RFT1, RFVT1, hRFT1
UniGene
Hs.632247
Gene Chromosome Location
Chr.17: 5032602-5035458 on Build GRCh38
Transcripts
NM_001104577.1 NM_017986.3 XM_011523951.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5563906 c.210G>A p.Q70Q Chr.17: 5034279-5034279 on Build GRCh38
5603677 c.249G>A p.V83V Chr.17: 5034240-5034240 on Build GRCh38
6870069 c.217C>T p.P73S Chr.17: 5034272-5034272 on Build GRCh38
4130409 c.209A>G p.Q70R Chr.17: 5034280-5034280 on Build GRCh38
6030767 c.239C>T p.P80L Chr.17: 5034250-5034250 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201707443 Chr.17: 5034300-5034300 on Build GRCh38
rs757872265 Chr.17: 5034273-5034273 on Build GRCh38
rs748184100 Chr.17: 5034354-5034354 on Build GRCh38
rs762541050 Chr.17: 5034345-5034345 on Build GRCh38
rs749566277 Chr.17: 5034308-5034308 on Build GRCh38
rs542144502 Chr.17: 5034348-5034348 on Build GRCh38
rs753612949 Chr.17: 5034267-5034267 on Build GRCh38
rs137937212 Chr.17: 5034271-5034271 on Build GRCh38
rs150112294 Chr.17: 5034281-5034281 on Build GRCh38
rs748471589 Chr.17: 5034232-5034232 on Build GRCh38
rs754509853 Chr.17: 5034358-5034358 on Build GRCh38
rs772259157 Chr.17: 5034245-5034246 on Build GRCh38
rs765056175 Chr.17: 5034338-5034338 on Build GRCh38
rs769190092 Chr.17: 5034404-5034413 on Build GRCh38
rs950022527 Chr.17: 5034336-5034336 on Build GRCh38
rs566207091 Chr.17: 5034321-5034321 on Build GRCh38
rs771313556 Chr.17: 5034316-5034316 on Build GRCh38
rs771187184 Chr.17: 5034243-5034243 on Build GRCh38
rs748537913 Chr.17: 5034293-5034293 on Build GRCh38
rs756520114 Chr.17: 5034382-5034382 on Build GRCh38
rs771221746 Chr.17: 5034400-5034400 on Build GRCh38
rs765958750 Chr.17: 5034346-5034346 on Build GRCh38
rs781712037 Chr.17: 5034367-5034367 on Build GRCh38
rs773631155 Chr.17: 5034239-5034239 on Build GRCh38
rs346822 Chr.17: 5034280-5034280 on Build GRCh38
rs749694465 Chr.17: 5034389-5034389 on Build GRCh38
rs770070991 Chr.17: 5034234-5034234 on Build GRCh38
rs779498689 Chr.17: 5034274-5034274 on Build GRCh38
rs751011254 Chr.17: 5034357-5034357 on Build GRCh38
rs780535507 Chr.17: 5034288-5034302 on Build GRCh38
rs376978383 Chr.17: 5034260-5034260 on Build GRCh38
rs576210341 Chr.17: 5034330-5034330 on Build GRCh38
rs781552117 Chr.17: 5034291-5034291 on Build GRCh38
rs775262570 Chr.17: 5034249-5034249 on Build GRCh38
rs776499536 Chr.17: 5034331-5034331 on Build GRCh38
rs761583604 Chr.17: 5034334-5034334 on Build GRCh38
rs745955975 Chr.17: 5034409-5034409 on Build GRCh38
rs763115742 Chr.17: 5034240-5034240 on Build GRCh38
rs750249520 Chr.17: 5034341-5034341 on Build GRCh38
rs747451336 Chr.17: 5034333-5034334 on Build GRCh38
rs138410039 Chr.17: 5034322-5034322 on Build GRCh38
rs763962389 Chr.17: 5034261-5034261 on Build GRCh38
rs753310420 Chr.17: 5034379-5034379 on Build GRCh38
rs778245551 Chr.17: 5034388-5034388 on Build GRCh38
rs779008895 Chr.17: 5034401-5034401 on Build GRCh38
rs769586204 Chr.17: 5034377-5034377 on Build GRCh38
rs201983129 Chr.17: 5034270-5034270 on Build GRCh38

View Full Product Details