Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61411716-61411987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPSF7
Entrez Gene ID
79869
Gene Name
cleavage and polyadenylation specific factor 7
Gene Aliases
CFIm59
UniGene
Hs.718984
Gene Chromosome Location
Chr.11: 61402648-61429992 on Build GRCh38
Transcripts
NM_001136040.2 NM_001142565.1 NM_024811.3 XM_005274298.4 XM_005274299.4 XM_005274303.4 XM_011545257.2 XM_011545258.2 XM_011545259.2 XM_011545260.2 XM_011545261.2 XM_011545262.2 XM_011545263.2 XM_017018345.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5838969 c.1298G>A p.R433H Chr.11: 61411853-61411853 on Build GRCh38
6371727 c.1279G>A p.A427T Chr.11: 61411872-61411872 on Build GRCh38
4464776 c.1350C>T p.S450S Chr.11: 61411801-61411801 on Build GRCh38
1202207 c.1089T>G p.D363E Chr.11: 61411933-61411933 on Build GRCh38
6476107 c.1252A>T p.R418W Chr.11: 61411770-61411770 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763248041 Chr.11: 61411799-61411799 on Build GRCh38
rs189534199 Chr.11: 61411954-61411954 on Build GRCh38
rs773569202 Chr.11: 61411895-61411895 on Build GRCh38
rs762201636 Chr.11: 61411784-61411784 on Build GRCh38
rs3019198 Chr.11: 61411813-61411813 on Build GRCh38
rs765579500 Chr.11: 61411794-61411794 on Build GRCh38
rs974235697 Chr.11: 61411925-61411925 on Build GRCh38
rs758906765 Chr.11: 61411751-61411751 on Build GRCh38
rs766443721 Chr.11: 61411802-61411802 on Build GRCh38
rs771447715 Chr.11: 61411888-61411888 on Build GRCh38
rs769241445 Chr.11: 61411753-61411753 on Build GRCh38
rs781239451 Chr.11: 61411815-61411815 on Build GRCh38
rs766655496 Chr.11: 61411909-61411909 on Build GRCh38
rs750700230 Chr.11: 61411795-61411795 on Build GRCh38
rs778393623 Chr.11: 61411877-61411877 on Build GRCh38
rs748946177 Chr.11: 61411740-61411740 on Build GRCh38
rs145145100 Chr.11: 61411920-61411920 on Build GRCh38
rs763179616 Chr.11: 61411900-61411900 on Build GRCh38
rs1046140193 Chr.11: 61411959-61411959 on Build GRCh38
rs756041085 Chr.11: 61411845-61411845 on Build GRCh38
rs749036143 Chr.11: 61411859-61411859 on Build GRCh38
rs762291543 Chr.11: 61411891-61411891 on Build GRCh38
rs185070681 Chr.11: 61411769-61411769 on Build GRCh38
rs777698856 Chr.11: 61411853-61411853 on Build GRCh38
rs777184800 Chr.11: 61411890-61411890 on Build GRCh38
rs770226551 Chr.11: 61411894-61411894 on Build GRCh38
rs767691085 Chr.11: 61411924-61411924 on Build GRCh38
rs1054948343 Chr.11: 61411868-61411868 on Build GRCh38
rs773928470 Chr.11: 61411750-61411750 on Build GRCh38
rs941970622 Chr.11: 61411866-61411866 on Build GRCh38
rs374817988 Chr.11: 61411810-61411810 on Build GRCh38
rs770624423 Chr.11: 61411876-61411876 on Build GRCh38
rs752652218 Chr.11: 61411841-61411841 on Build GRCh38
rs566417848 Chr.11: 61411743-61411743 on Build GRCh38
rs745409283 Chr.11: 61411878-61411878 on Build GRCh38
rs774406041 Chr.11: 61411912-61411912 on Build GRCh38
rs201573466 Chr.11: 61411887-61411887 on Build GRCh38

View Full Product Details