Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99342956-99343181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADH1C
Entrez Gene ID
126
Gene Name
alcohol dehydrogenase 1C (class I), gamma polypeptide
Gene Aliases
ADH3
UniGene
Hs.654537
Gene Chromosome Location
Chr.4: 99336492-99353045 on Build GRCh38
Transcripts
NM_000669.4 NR_133005.1 XM_011531588.2 XM_011531589.1

SNP Detail

SNP ID Chromosome Location
rs777878896 Chr.4: 99343056-99343056 on Build GRCh38
rs998336924 Chr.4: 99343134-99343134 on Build GRCh38
rs200810515 Chr.4: 99343052-99343052 on Build GRCh38
rs78115560 Chr.4: 99343016-99343016 on Build GRCh38
rs377645569 Chr.4: 99343024-99343024 on Build GRCh38
rs755222271 Chr.4: 99342994-99342994 on Build GRCh38
rs754312357 Chr.4: 99343044-99343044 on Build GRCh38
rs745661717 Chr.4: 99343102-99343102 on Build GRCh38
rs747056116 Chr.4: 99343064-99343064 on Build GRCh38
rs779008943 Chr.4: 99343051-99343051 on Build GRCh38
rs879132155 Chr.4: 99343058-99343058 on Build GRCh38
rs779183871 Chr.4: 99342996-99342996 on Build GRCh38
rs758427942 Chr.4: 99343055-99343055 on Build GRCh38
rs80086338 Chr.4: 99343014-99343014 on Build GRCh38
rs768891523 Chr.4: 99343087-99343089 on Build GRCh38
rs772124828 Chr.4: 99343001-99343001 on Build GRCh38
rs781094122 Chr.4: 99343084-99343084 on Build GRCh38
rs528886854 Chr.4: 99343013-99343013 on Build GRCh38
rs74561381 Chr.4: 99343010-99343010 on Build GRCh38
rs761307510 Chr.4: 99343038-99343038 on Build GRCh38
rs955883857 Chr.4: 99343009-99343009 on Build GRCh38
rs1009512338 Chr.4: 99343101-99343101 on Build GRCh38
rs892284103 Chr.4: 99343092-99343092 on Build GRCh38
rs769564379 Chr.4: 99343105-99343105 on Build GRCh38
rs776795864 Chr.4: 99343102-99343109 on Build GRCh38
rs78113489 Chr.4: 99343011-99343011 on Build GRCh38
rs762371733 Chr.4: 99343022-99343022 on Build GRCh38
rs746898978 Chr.4: 99343005-99343005 on Build GRCh38
rs905289726 Chr.4: 99343060-99343060 on Build GRCh38
rs35088274 Chr.4: 99343067-99343068 on Build GRCh38
rs777717488 Chr.4: 99343002-99343002 on Build GRCh38
rs1036723412 Chr.4: 99343113-99343113 on Build GRCh38
rs78680159 Chr.4: 99343079-99343079 on Build GRCh38
rs561697999 Chr.4: 99343008-99343008 on Build GRCh38
rs748191499 Chr.4: 99343000-99343000 on Build GRCh38
rs751095026 Chr.4: 99343037-99343037 on Build GRCh38
rs533562839 Chr.4: 99343119-99343119 on Build GRCh38
rs552031730 Chr.4: 99343145-99343145 on Build GRCh38
rs528054596 Chr.4: 99343059-99343059 on Build GRCh38
rs754228249 Chr.4: 99342978-99342978 on Build GRCh38

View Full Product Details