Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 96223606-96223843 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELK3
Entrez Gene ID
2004
Gene Name
ELK3, ETS transcription factor
Gene Aliases
ERP, NET, SAP-2, SAP2
UniGene
Hs.46523
Gene Chromosome Location
Chr.12: 96194382-96269835 on Build GRCh38
Transcripts
NM_005230.3 XM_006719275.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
287283 c.150_151insA p.T52fs*6 Chr.12: 96223716-96223717 on Build GRCh38
6445836 c.99C>G p.F33L Chr.12: 96223665-96223665 on Build GRCh38
5842068 c.136G>A p.G46R Chr.12: 96223702-96223702 on Build GRCh38
3466660 c.49G>A p.D17N Chr.12: 96223615-96223615 on Build GRCh38
2046604 c.144delA p.N50fs*5 Chr.12: 96223710-96223710 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745990061 Chr.12: 96223669-96223669 on Build GRCh38
rs776745602 Chr.12: 96223708-96223708 on Build GRCh38
rs139820919 Chr.12: 96223752-96223752 on Build GRCh38
rs118107469 Chr.12: 96223821-96223821 on Build GRCh38
rs374940996 Chr.12: 96223819-96223819 on Build GRCh38
rs754086796 Chr.12: 96223796-96223796 on Build GRCh38
rs1021438274 Chr.12: 96223794-96223794 on Build GRCh38
rs762669176 Chr.12: 96223737-96223737 on Build GRCh38
rs756511475 Chr.12: 96223649-96223649 on Build GRCh38
rs79069381 Chr.12: 96223800-96223800 on Build GRCh38
rs138151504 Chr.12: 96223650-96223650 on Build GRCh38
rs1011815122 Chr.12: 96223758-96223758 on Build GRCh38
rs772840752 Chr.12: 96223725-96223725 on Build GRCh38
rs753199489 Chr.12: 96223643-96223643 on Build GRCh38
rs755491429 Chr.12: 96223638-96223638 on Build GRCh38
rs769732719 Chr.12: 96223721-96223721 on Build GRCh38
rs756568639 Chr.12: 96223792-96223792 on Build GRCh38
rs376285118 Chr.12: 96223654-96223654 on Build GRCh38
rs778234492 Chr.12: 96223795-96223795 on Build GRCh38
rs751212381 Chr.12: 96223780-96223780 on Build GRCh38
rs146979376 Chr.12: 96223630-96223630 on Build GRCh38
rs533355356 Chr.12: 96223804-96223804 on Build GRCh38
rs753256762 Chr.12: 96223787-96223787 on Build GRCh38
rs754643863 Chr.12: 96223653-96223653 on Build GRCh38
rs757626374 Chr.12: 96223799-96223799 on Build GRCh38
rs201254610 Chr.12: 96223715-96223715 on Build GRCh38
rs746052980 Chr.12: 96223808-96223808 on Build GRCh38
rs529931556 Chr.12: 96223785-96223785 on Build GRCh38
rs766983348 Chr.12: 96223786-96223786 on Build GRCh38
rs772323404 Chr.12: 96223817-96223817 on Build GRCh38
rs550625180 Chr.12: 96223683-96223683 on Build GRCh38
rs781708033 Chr.12: 96223641-96223641 on Build GRCh38

View Full Product Details