Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1390135-1390402 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNL2
Entrez Gene ID
81669
Gene Name
cyclin L2
Gene Aliases
ANIA-6B, CCNM, CCNS, HCLA-ISO, HLA-ISO, PCEE, SB138
UniGene
Hs.515704
Gene Chromosome Location
Chr.1: 1385711-1399338 on Build GRCh38
Transcripts
NM_001320153.1 NM_001320155.1 NM_030937.4 NR_135154.1 XM_011542216.2 XM_011542219.2 XM_011542221.2 XM_017002420.1 XM_017002421.1 XM_017002422.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1200177 c.871C>A p.L291I Chr.1: 1390365-1390365 on Build GRCh38
6053960 c.999C>T p.P333P Chr.1: 1390237-1390237 on Build GRCh38
4670247 c.901_902insA p.R301fs*43 Chr.1: 1390334-1390335 on Build GRCh38
674895 c.980C>T p.S327L Chr.1: 1390256-1390256 on Build GRCh38
674896 c.991C>G p.P331A Chr.1: 1390245-1390245 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs189208343 Chr.1: 1390175-1390175 on Build GRCh38
rs372064998 Chr.1: 1390277-1390277 on Build GRCh38
rs905220040 Chr.1: 1390167-1390167 on Build GRCh38
rs767606471 Chr.1: 1390340-1390341 on Build GRCh38
rs780189508 Chr.1: 1390314-1390314 on Build GRCh38
rs752023243 Chr.1: 1390213-1390213 on Build GRCh38
rs954265879 Chr.1: 1390313-1390313 on Build GRCh38
rs769726291 Chr.1: 1390349-1390349 on Build GRCh38
rs764822403 Chr.1: 1390183-1390183 on Build GRCh38
rs866319047 Chr.1: 1390170-1390170 on Build GRCh38
rs748925471 Chr.1: 1390372-1390372 on Build GRCh38
rs767908876 Chr.1: 1390307-1390307 on Build GRCh38
rs750853732 Chr.1: 1390308-1390308 on Build GRCh38
rs373668257 Chr.1: 1390194-1390194 on Build GRCh38
rs201228864 Chr.1: 1390241-1390241 on Build GRCh38
rs762145584 Chr.1: 1390306-1390307 on Build GRCh38
rs757593691 Chr.1: 1390217-1390217 on Build GRCh38
rs752067027 Chr.1: 1390299-1390299 on Build GRCh38
rs1002225955 Chr.1: 1390168-1390168 on Build GRCh38
rs745595081 Chr.1: 1390340-1390340 on Build GRCh38
rs768182819 Chr.1: 1390378-1390378 on Build GRCh38
rs756474698 Chr.1: 1390309-1390309 on Build GRCh38
rs889385182 Chr.1: 1390178-1390178 on Build GRCh38
rs754077670 Chr.1: 1390320-1390320 on Build GRCh38
rs775233771 Chr.1: 1390354-1390354 on Build GRCh38
rs749297936 Chr.1: 1390256-1390256 on Build GRCh38
rs770724720 Chr.1: 1390280-1390280 on Build GRCh38
rs764596166 Chr.1: 1390197-1390197 on Build GRCh38
rs762494947 Chr.1: 1390298-1390298 on Build GRCh38
rs752315411 Chr.1: 1390188-1390188 on Build GRCh38
rs901887068 Chr.1: 1390201-1390201 on Build GRCh38
rs781133716 Chr.1: 1390337-1390337 on Build GRCh38
rs781441909 Chr.1: 1390221-1390221 on Build GRCh38
rs367995434 Chr.1: 1390255-1390255 on Build GRCh38
rs746945804 Chr.1: 1390326-1390326 on Build GRCh38
rs55993534 Chr.1: 1390297-1390297 on Build GRCh38
rs777712522 Chr.1: 1390273-1390273 on Build GRCh38
rs776619502 Chr.1: 1390283-1390283 on Build GRCh38
rs750621217 Chr.1: 1390239-1390239 on Build GRCh38
rs139455818 Chr.1: 1390266-1390266 on Build GRCh38
rs1009695754 Chr.1: 1390238-1390238 on Build GRCh38
rs759253906 Chr.1: 1390284-1390284 on Build GRCh38
rs1040530021 Chr.1: 1390200-1390200 on Build GRCh38
rs141435652 Chr.1: 1390304-1390304 on Build GRCh38
rs965805508 Chr.1: 1390328-1390328 on Build GRCh38
rs145603397 Chr.1: 1390327-1390327 on Build GRCh38
rs755043411 Chr.1: 1390321-1390321 on Build GRCh38
rs768628698 Chr.1: 1390265-1390265 on Build GRCh38
rs775287278 Chr.1: 1390293-1390293 on Build GRCh38
rs1056452162 Chr.1: 1390185-1390185 on Build GRCh38

View Full Product Details