Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 129846733-129846980 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM49B
Entrez Gene ID
51571
Gene Name
family with sequence similarity 49 member B
Gene Aliases
BM-009, L1
UniGene
Hs.126941
Gene Chromosome Location
Chr.8: 129839593-130016697 on Build GRCh38
Transcripts
NM_001256763.1 NM_016623.4 NR_046359.1 NR_046360.1 NR_046361.1 XM_005250966.3 XM_011517108.2 XM_011517109.2 XM_011517110.2 XM_011517111.2 XM_011517112.2 XM_011517113.2 XM_011517114.2 XM_011517115.2 XM_011517116.2 XM_011517117.2 XM_011517118.2 XM_011517120.2 XM_011517121.2 XM_011517122.2 XM_011517123.2 XM_011517124.2 XM_011517125.2 XM_017013539.1 XM_017013540.1 XM_017013541.1 XM_017013542.1 XM_017013543.1 XM_017013544.1 XM_017013545.1 XM_017013546.1 XM_017013547.1 XM_017013548.1 XM_017013549.1 XM_017013550.1 XM_017013551.1 XM_017013552.1 XM_017013553.1 XM_017013554.1 XM_017013555.1 XM_017013556.1 XM_017013557.1 XM_017013558.1 XM_017013559.1 XM_017013560.1 XM_017013561.1 XM_017013562.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6278032 c.841-6C>T p.? Chr.8: 129846880-129846880 on Build GRCh38
5460604 c.900A>T p.L300L Chr.8: 129846815-129846815 on Build GRCh38
7161081 c.899T>C p.L300P Chr.8: 129846816-129846816 on Build GRCh38
4683056 c.850_852delTGT p.C284delC Chr.8: 129846863-129846865 on Build GRCh38
4162670 c.907C>T p.L303F Chr.8: 129846808-129846808 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187178073 Chr.8: 129846784-129846784 on Build GRCh38
rs898286655 Chr.8: 129846937-129846937 on Build GRCh38
rs376275542 Chr.8: 129846815-129846815 on Build GRCh38
rs779115716 Chr.8: 129846921-129846921 on Build GRCh38
rs201901394 Chr.8: 129846881-129846881 on Build GRCh38
rs988356944 Chr.8: 129846920-129846920 on Build GRCh38
rs140233071 Chr.8: 129846898-129846900 on Build GRCh38
rs768816550 Chr.8: 129846767-129846767 on Build GRCh38
rs1025238452 Chr.8: 129846942-129846942 on Build GRCh38
rs754135339 Chr.8: 129846905-129846905 on Build GRCh38
rs769259064 Chr.8: 129846767-129846768 on Build GRCh38
rs780213892 Chr.8: 129846904-129846904 on Build GRCh38
rs369349917 Chr.8: 129846789-129846789 on Build GRCh38
rs748746611 Chr.8: 129846782-129846782 on Build GRCh38
rs928935790 Chr.8: 129846943-129846943 on Build GRCh38
rs114602465 Chr.8: 129846790-129846790 on Build GRCh38
rs751442071 Chr.8: 129846845-129846845 on Build GRCh38
rs890802624 Chr.8: 129846944-129846944 on Build GRCh38
rs111860343 Chr.8: 129846830-129846830 on Build GRCh38
rs774975100 Chr.8: 129846921-129846921 on Build GRCh38
rs779262705 Chr.8: 129846779-129846779 on Build GRCh38
rs772318112 Chr.8: 129846783-129846783 on Build GRCh38
rs750166103 Chr.8: 129846893-129846893 on Build GRCh38
rs376024139 Chr.8: 129846904-129846905 on Build GRCh38
rs754999228 Chr.8: 129846912-129846912 on Build GRCh38
rs115025292 Chr.8: 129846939-129846939 on Build GRCh38
rs763712982 Chr.8: 129846842-129846842 on Build GRCh38
rs905998256 Chr.8: 129846903-129846903 on Build GRCh38
rs755931213 Chr.8: 129846900-129846900 on Build GRCh38
rs761878742 Chr.8: 129846853-129846853 on Build GRCh38

View Full Product Details