Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4702195-4702420 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKS3
Entrez Gene ID
124401
Gene Name
ankyrin repeat and sterile alpha motif domain containing 3
Gene Aliases
-
UniGene
Hs.654859
Gene Chromosome Location
Chr.16: 4696510-4734377 on Build GRCh38
Transcripts
NM_001242929.1 NM_001308089.1 NM_001324129.1 NM_001324130.1 NM_133450.3 NR_040252.1 XM_005255119.3 XM_005255121.3 XM_011522372.1 XM_011522373.1 XM_011522374.1 XM_011522378.1 XM_017022935.1 XM_017022936.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1189171 c.880C>A p.P294T Chr.16: 4702231-4702231 on Build GRCh38
5384901 c.278C>T p.T93I Chr.16: 4702215-4702215 on Build GRCh38
5384900 c.896C>T p.T299I Chr.16: 4702215-4702215 on Build GRCh38
1708706 c.883C>T p.R295C Chr.16: 4702228-4702228 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1016902838 Chr.16: 4702352-4702352 on Build GRCh38
rs770886046 Chr.16: 4702232-4702232 on Build GRCh38
rs1009849032 Chr.16: 4702383-4702383 on Build GRCh38
rs369688646 Chr.16: 4702227-4702227 on Build GRCh38
rs962613531 Chr.16: 4702365-4702365 on Build GRCh38
rs1039839437 Chr.16: 4702276-4702276 on Build GRCh38
rs922644805 Chr.16: 4702299-4702299 on Build GRCh38
rs754655112 Chr.16: 4702221-4702221 on Build GRCh38
rs906253087 Chr.16: 4702391-4702391 on Build GRCh38
rs745606706 Chr.16: 4702245-4702245 on Build GRCh38
rs138641439 Chr.16: 4702257-4702257 on Build GRCh38
rs749232019 Chr.16: 4702231-4702231 on Build GRCh38
rs201331767 Chr.16: 4702292-4702292 on Build GRCh38
rs760865384 Chr.16: 4702259-4702259 on Build GRCh38
rs879236337 Chr.16: 4702255-4702255 on Build GRCh38
rs776042270 Chr.16: 4702375-4702375 on Build GRCh38
rs998654832 Chr.16: 4702307-4702307 on Build GRCh38
rs376883656 Chr.16: 4702252-4702252 on Build GRCh38
rs1049734062 Chr.16: 4702306-4702306 on Build GRCh38
rs116370731 Chr.16: 4702315-4702315 on Build GRCh38
rs772432689 Chr.16: 4702247-4702247 on Build GRCh38
rs889804045 Chr.16: 4702313-4702313 on Build GRCh38
rs774079337 Chr.16: 4702233-4702233 on Build GRCh38
rs755572917 Chr.16: 4702228-4702228 on Build GRCh38
rs374274866 Chr.16: 4702347-4702347 on Build GRCh38
rs575828723 Chr.16: 4702388-4702388 on Build GRCh38
rs1020752763 Chr.16: 4702387-4702387 on Build GRCh38
rs755248845 Chr.16: 4702312-4702312 on Build GRCh38
rs972804151 Chr.16: 4702291-4702291 on Build GRCh38
rs555649083 Chr.16: 4702317-4702317 on Build GRCh38
rs777233491 Chr.16: 4702230-4702230 on Build GRCh38
rs769803079 Chr.16: 4702303-4702303 on Build GRCh38
rs887074103 Chr.16: 4702327-4702327 on Build GRCh38
rs148885466 Chr.16: 4702394-4702394 on Build GRCh38
rs112546188 Chr.16: 4702376-4702376 on Build GRCh38
rs934063259 Chr.16: 4702304-4702304 on Build GRCh38
rs776752307 Chr.16: 4702284-4702284 on Build GRCh38
rs764313230 Chr.16: 4702281-4702281 on Build GRCh38

View Full Product Details