Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 109166774-109167017 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACACB
Entrez Gene ID
32
Gene Name
acetyl-CoA carboxylase beta
Gene Aliases
ACC2, ACCB, HACC275
UniGene
Hs.234898
Gene Chromosome Location
Chr.12: 109112666-109268226 on Build GRCh38
Transcripts
NM_001093.3 XM_005253876.4 XM_006719367.3 XM_011538259.2 XM_011538263.2 XM_011538264.2 XM_011538265.2 XM_017019252.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4038466 c.654-1G>T p.? Chr.12: 109166860-109166860 on Build GRCh38
6591562 c.750T>C p.F250F Chr.12: 109166957-109166957 on Build GRCh38
6591517 c.755C>T p.T252I Chr.12: 109166962-109166962 on Build GRCh38
1181495 c.730G>A p.V244M Chr.12: 109166937-109166937 on Build GRCh38
7275985 c.668G>A p.G223E Chr.12: 109166875-109166875 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144515904 Chr.12: 109166980-109166980 on Build GRCh38
rs551117282 Chr.12: 109166913-109166913 on Build GRCh38
rs775499731 Chr.12: 109166886-109166886 on Build GRCh38
rs755422374 Chr.12: 109166845-109166845 on Build GRCh38
rs998573576 Chr.12: 109166976-109166976 on Build GRCh38
rs751757015 Chr.12: 109166924-109166924 on Build GRCh38
rs374895333 Chr.12: 109166814-109166814 on Build GRCh38
rs17848820 Chr.12: 109166863-109166863 on Build GRCh38
rs768607691 Chr.12: 109166969-109166970 on Build GRCh38
rs760700219 Chr.12: 109166893-109166893 on Build GRCh38
rs751697373 Chr.12: 109166840-109166840 on Build GRCh38
rs746969049 Chr.12: 109166964-109166964 on Build GRCh38
rs781691986 Chr.12: 109166853-109166853 on Build GRCh38
rs975360803 Chr.12: 109166800-109166800 on Build GRCh38
rs146370716 Chr.12: 109166965-109166965 on Build GRCh38
rs745333353 Chr.12: 109166880-109166880 on Build GRCh38
rs528602761 Chr.12: 109166898-109166898 on Build GRCh38
rs760505301 Chr.12: 109166828-109166828 on Build GRCh38
rs748928594 Chr.12: 109166872-109166872 on Build GRCh38
rs756674820 Chr.12: 109166946-109166946 on Build GRCh38
rs150653401 Chr.12: 109166948-109166948 on Build GRCh38
rs763984374 Chr.12: 109166829-109166829 on Build GRCh38
rs146426104 Chr.12: 109166971-109166971 on Build GRCh38
rs763310968 Chr.12: 109166832-109166832 on Build GRCh38
rs149394526 Chr.12: 109166964-109166964 on Build GRCh38
rs778125313 Chr.12: 109166868-109166868 on Build GRCh38
rs769511892 Chr.12: 109166972-109166972 on Build GRCh38
rs775005793 Chr.12: 109166955-109166955 on Build GRCh38
rs776631835 Chr.12: 109166899-109166899 on Build GRCh38
rs1036336839 Chr.12: 109166859-109166859 on Build GRCh38
rs745510148 Chr.12: 109166956-109166956 on Build GRCh38
rs771552176 Chr.12: 109166962-109166962 on Build GRCh38
rs141887668 Chr.12: 109166909-109166909 on Build GRCh38
rs768555581 Chr.12: 109166894-109166894 on Build GRCh38
rs775589789 Chr.12: 109166819-109166819 on Build GRCh38
rs527512063 Chr.12: 109166878-109166878 on Build GRCh38
rs145288622 Chr.12: 109166864-109166864 on Build GRCh38
rs764635735 Chr.12: 109166990-109166990 on Build GRCh38
rs750612156 Chr.12: 109166831-109166831 on Build GRCh38
rs751921482 Chr.12: 109166936-109166936 on Build GRCh38
rs774568250 Chr.12: 109166974-109166974 on Build GRCh38
rs188913842 Chr.12: 109166987-109166987 on Build GRCh38
rs778588229 Chr.12: 109166953-109166953 on Build GRCh38
rs373813180 Chr.12: 109166873-109166873 on Build GRCh38
rs114166382 Chr.12: 109166801-109166801 on Build GRCh38
rs990499643 Chr.12: 109166914-109166914 on Build GRCh38
rs766360223 Chr.12: 109166835-109166835 on Build GRCh38
rs768750072 Chr.12: 109166968-109166968 on Build GRCh38
rs940784589 Chr.12: 109166843-109166843 on Build GRCh38
rs759694366 Chr.12: 109166979-109166979 on Build GRCh38
rs753146594 Chr.12: 109166937-109166937 on Build GRCh38
rs373292549 Chr.12: 109166875-109166875 on Build GRCh38
rs771512989 Chr.12: 109166882-109166882 on Build GRCh38
rs146179662 Chr.12: 109166988-109166988 on Build GRCh38
rs781314104 Chr.12: 109166949-109166949 on Build GRCh38

View Full Product Details