Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 154604887-154605124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGG
Entrez Gene ID
2266
Gene Name
fibrinogen gamma chain
Gene Aliases
-
UniGene
Hs.727584
Gene Chromosome Location
Chr.4: 154604134-154612808 on Build GRCh38
Transcripts
NM_000509.5 NM_021870.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3825430 c.1236A>G p.I412M Chr.4: 154604960-154604960 on Build GRCh38
7082471 c.1284G>A p.L428L Chr.4: 154604912-154604912 on Build GRCh38
4773636 c.1253T>C p.L418P Chr.4: 154604943-154604943 on Build GRCh38
1695131 c.1230G>A p.M410I Chr.4: 154604966-154604966 on Build GRCh38
6988897 c.1304G>A p.R435K Chr.4: 154604892-154604892 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201436157 Chr.4: 154605084-154605084 on Build GRCh38
rs2066878 Chr.4: 154605079-154605079 on Build GRCh38
rs1011406807 Chr.4: 154605014-154605014 on Build GRCh38
rs886059154 Chr.4: 154604959-154604959 on Build GRCh38
rs754193577 Chr.4: 154604966-154604966 on Build GRCh38
rs771478262 Chr.4: 154605090-154605090 on Build GRCh38
rs121913095 Chr.4: 154605028-154605028 on Build GRCh38
rs752621352 Chr.4: 154605056-154605056 on Build GRCh38
rs369271819 Chr.4: 154605066-154605066 on Build GRCh38
rs745574025 Chr.4: 154605089-154605089 on Build GRCh38
rs752060359 Chr.4: 154604939-154604939 on Build GRCh38
rs749095294 Chr.4: 154604930-154604930 on Build GRCh38
rs1005046731 Chr.4: 154604913-154604913 on Build GRCh38
rs35443439 Chr.4: 154604915-154604915 on Build GRCh38
rs774084996 Chr.4: 154604936-154604936 on Build GRCh38
rs201018513 Chr.4: 154604958-154604958 on Build GRCh38
rs200444079 Chr.4: 154604964-154604964 on Build GRCh38
rs778405765 Chr.4: 154605086-154605086 on Build GRCh38
rs745865758 Chr.4: 154604907-154604907 on Build GRCh38
rs773049609 Chr.4: 154604912-154604912 on Build GRCh38
rs761331562 Chr.4: 154604961-154604961 on Build GRCh38
rs587777720 Chr.4: 154604986-154604986 on Build GRCh38
rs780251516 Chr.4: 154605040-154605040 on Build GRCh38
rs770425488 Chr.4: 154604934-154604934 on Build GRCh38
rs75848804 Chr.4: 154604995-154604995 on Build GRCh38
rs776109029 Chr.4: 154604956-154604956 on Build GRCh38
rs112102623 Chr.4: 154605069-154605069 on Build GRCh38
rs758920678 Chr.4: 154604937-154604937 on Build GRCh38
rs374917187 Chr.4: 154604938-154604938 on Build GRCh38
rs199520597 Chr.4: 154605017-154605017 on Build GRCh38
rs780602722 Chr.4: 154605020-154605020 on Build GRCh38
rs746473186 Chr.4: 154604929-154604931 on Build GRCh38
rs1036477853 Chr.4: 154604923-154604923 on Build GRCh38
rs1021492173 Chr.4: 154605049-154605049 on Build GRCh38
rs6061 Chr.4: 154604968-154604968 on Build GRCh38
rs750709345 Chr.4: 154605035-154605035 on Build GRCh38
rs758380539 Chr.4: 154605036-154605036 on Build GRCh38
rs145938787 Chr.4: 154604911-154604911 on Build GRCh38
rs758530530 Chr.4: 154604910-154604910 on Build GRCh38
rs896318406 Chr.4: 154605060-154605060 on Build GRCh38
rs755482770 Chr.4: 154604998-154604998 on Build GRCh38

View Full Product Details