Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 101353793-101354064 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTK
Entrez Gene ID
695
Gene Name
Bruton tyrosine kinase
Gene Aliases
AGMX1, AT, ATK, BPK, IMD1, PSCTK1, XLA
UniGene
Hs.159494
Gene Chromosome Location
Chr.X: 101349447-101390796 on Build GRCh38
Transcripts
NM_000061.2 NM_001287344.1 NM_001287345.1

SNP Detail

SNP ID Chromosome Location
rs782358652 Chr.X: 101354003-101354003 on Build GRCh38
rs782084821 Chr.X: 101353967-101353967 on Build GRCh38
rs370315801 Chr.X: 101353848-101353848 on Build GRCh38
rs864321659 Chr.X: 101353865-101353865 on Build GRCh38
rs886039555 Chr.X: 101353990-101353990 on Build GRCh38
rs3208811 Chr.X: 101353934-101353934 on Build GRCh38
rs199950975 Chr.X: 101353928-101353928 on Build GRCh38
rs104894770 Chr.X: 101353935-101353935 on Build GRCh38
rs151072282 Chr.X: 101354044-101354044 on Build GRCh38
rs781988052 Chr.X: 101354006-101354006 on Build GRCh38
rs782187980 Chr.X: 101353840-101353840 on Build GRCh38
rs128621204 Chr.X: 101353936-101353936 on Build GRCh38
rs375718074 Chr.X: 101353838-101353838 on Build GRCh38
rs377324584 Chr.X: 101353883-101353883 on Build GRCh38
rs374131821 Chr.X: 101353860-101353860 on Build GRCh38
rs370902664 Chr.X: 101353956-101353956 on Build GRCh38
rs782458956 Chr.X: 101353843-101353843 on Build GRCh38
rs782019387 Chr.X: 101353892-101353892 on Build GRCh38
rs782141849 Chr.X: 101354008-101354008 on Build GRCh38
rs193922126 Chr.X: 101353940-101353947 on Build GRCh38
rs3208813 Chr.X: 101353915-101353915 on Build GRCh38
rs128621205 Chr.X: 101353879-101353879 on Build GRCh38
rs146681416 Chr.X: 101353919-101353919 on Build GRCh38

View Full Product Details