Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 170432325-170432586 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN11
Entrez Gene ID
5010
Gene Name
claudin 11
Gene Aliases
OSP, OTM
UniGene
Hs.31595
Gene Chromosome Location
Chr.3: 170418865-170434691 on Build GRCh38
Transcripts
NM_001185056.1 NM_005602.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4672676 c.440G>A p.R147H Chr.3: 170432572-170432572 on Build GRCh38
119880 c.451A>G p.I151V Chr.3: 170432583-170432583 on Build GRCh38
5443286 c.423C>T p.F141F Chr.3: 170432555-170432555 on Build GRCh38
6629962 c.432C>T p.C144C Chr.3: 170432564-170432564 on Build GRCh38
3427319 c.400G>A p.A134T Chr.3: 170432532-170432532 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187863554 Chr.3: 170432362-170432362 on Build GRCh38
rs761859543 Chr.3: 170432512-170432512 on Build GRCh38
rs751647141 Chr.3: 170432358-170432358 on Build GRCh38
rs756826846 Chr.3: 170432567-170432567 on Build GRCh38
rs1013268459 Chr.3: 170432454-170432454 on Build GRCh38
rs781262121 Chr.3: 170432534-170432534 on Build GRCh38
rs182147642 Chr.3: 170432353-170432353 on Build GRCh38
rs764329420 Chr.3: 170432506-170432506 on Build GRCh38
rs781040094 Chr.3: 170432506-170432507 on Build GRCh38
rs754478839 Chr.3: 170432488-170432489 on Build GRCh38
rs765340122 Chr.3: 170432516-170432516 on Build GRCh38
rs369897123 Chr.3: 170432493-170432493 on Build GRCh38
rs371975234 Chr.3: 170432504-170432504 on Build GRCh38
rs549752354 Chr.3: 170432565-170432565 on Build GRCh38
rs1035651956 Chr.3: 170432431-170432431 on Build GRCh38
rs138100932 Chr.3: 170432531-170432531 on Build GRCh38
rs564253337 Chr.3: 170432533-170432533 on Build GRCh38
rs531634544 Chr.3: 170432564-170432564 on Build GRCh38
rs34124448 Chr.3: 170432554-170432555 on Build GRCh38
rs372680458 Chr.3: 170432481-170432481 on Build GRCh38
rs149945748 Chr.3: 170432449-170432449 on Build GRCh38
rs751510772 Chr.3: 170432471-170432471 on Build GRCh38
rs776640561 Chr.3: 170432508-170432508 on Build GRCh38
rs533119827 Chr.3: 170432496-170432496 on Build GRCh38
rs139205558 Chr.3: 170432399-170432399 on Build GRCh38
rs199570340 Chr.3: 170432492-170432492 on Build GRCh38
rs201469287 Chr.3: 170432503-170432503 on Build GRCh38
rs368710254 Chr.3: 170432475-170432475 on Build GRCh38
rs887608262 Chr.3: 170432425-170432425 on Build GRCh38
rs114088900 Chr.3: 170432424-170432424 on Build GRCh38
rs143763226 Chr.3: 170432532-170432532 on Build GRCh38
rs754842359 Chr.3: 170432530-170432530 on Build GRCh38
rs115625283 Chr.3: 170432409-170432409 on Build GRCh38
rs911586804 Chr.3: 170432519-170432519 on Build GRCh38
rs767426219 Chr.3: 170432405-170432405 on Build GRCh38

View Full Product Details