Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 21416007-21416252 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNJ12
Entrez Gene ID
3768
Gene Name
potassium voltage-gated channel subfamily J member 12
Gene Aliases
IRK-2, IRK2, KCNJN1, Kir2.2, Kir2.2v, hIRK, hIRK1, hkir2.2x, kcnj12x
UniGene
Hs.200629
Gene Chromosome Location
Chr.17: 21376387-21419870 on Build GRCh38
Transcripts
NM_021012.4 XM_005256625.4 XM_011523831.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1470893 c.689C>T p.A230V Chr.17: 21416031-21416031 on Build GRCh38
312200 c.816T>C p.H272H Chr.17: 21416158-21416158 on Build GRCh38
4129718 c.889G>A p.V297I Chr.17: 21416231-21416231 on Build GRCh38
6440961 c.772G>A p.G258S Chr.17: 21416114-21416114 on Build GRCh38
5479277 c.723C>T p.G241G Chr.17: 21416065-21416065 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782310902 Chr.17: 21416026-21416026 on Build GRCh38
rs199837726 Chr.17: 21416167-21416167 on Build GRCh38
rs782229122 Chr.17: 21416204-21416204 on Build GRCh38
rs782726273 Chr.17: 21416122-21416122 on Build GRCh38
rs530111371 Chr.17: 21416233-21416233 on Build GRCh38
rs77724065 Chr.17: 21416153-21416153 on Build GRCh38
rs782653432 Chr.17: 21416046-21416046 on Build GRCh38
rs782569468 Chr.17: 21416048-21416048 on Build GRCh38
rs782345840 Chr.17: 21416187-21416187 on Build GRCh38
rs375466482 Chr.17: 21416168-21416168 on Build GRCh38
rs73979899 Chr.17: 21416080-21416080 on Build GRCh38
rs76684759 Chr.17: 21416127-21416127 on Build GRCh38
rs782250971 Chr.17: 21416209-21416209 on Build GRCh38
rs782543233 Chr.17: 21416230-21416230 on Build GRCh38
rs782530067 Chr.17: 21416199-21416199 on Build GRCh38
rs139358757 Chr.17: 21416185-21416185 on Build GRCh38
rs147653221 Chr.17: 21416047-21416047 on Build GRCh38
rs1657744 Chr.17: 21416161-21416161 on Build GRCh38
rs782779145 Chr.17: 21416125-21416125 on Build GRCh38
rs782505061 Chr.17: 21416172-21416172 on Build GRCh38
rs145578286 Chr.17: 21416096-21416096 on Build GRCh38
rs782594208 Chr.17: 21416203-21416203 on Build GRCh38
rs77048459 Chr.17: 21416057-21416057 on Build GRCh38
rs73313929 Chr.17: 21416140-21416140 on Build GRCh38
rs782443200 Chr.17: 21416205-21416205 on Build GRCh38
rs782027631 Chr.17: 21416191-21416191 on Build GRCh38
rs77270326 Chr.17: 21416124-21416124 on Build GRCh38
rs80335301 Chr.17: 21416231-21416231 on Build GRCh38
rs782260932 Chr.17: 21416061-21416061 on Build GRCh38
rs782199360 Chr.17: 21416049-21416049 on Build GRCh38
rs782335706 Chr.17: 21416190-21416190 on Build GRCh38
rs781975590 Chr.17: 21416188-21416188 on Build GRCh38
rs781897258 Chr.17: 21416043-21416043 on Build GRCh38
rs76493573 Chr.17: 21416158-21416158 on Build GRCh38
rs143946927 Chr.17: 21416195-21416195 on Build GRCh38
rs782763709 Chr.17: 21416223-21416223 on Build GRCh38
rs782450264 Chr.17: 21416197-21416197 on Build GRCh38
rs782460419 Chr.17: 21416038-21416038 on Build GRCh38
rs4985866 Chr.17: 21416087-21416087 on Build GRCh38
rs78113532 Chr.17: 21416207-21416207 on Build GRCh38
rs782590279 Chr.17: 21416206-21416206 on Build GRCh38
rs782335120 Chr.17: 21416210-21416210 on Build GRCh38
rs781966972 Chr.17: 21416117-21416117 on Build GRCh38
rs16962951 Chr.17: 21416095-21416095 on Build GRCh38
rs781790755 Chr.17: 21416083-21416083 on Build GRCh38
rs782011713 Chr.17: 21416072-21416072 on Build GRCh38
rs80022364 Chr.17: 21416215-21416215 on Build GRCh38
rs569037483 Chr.17: 21416029-21416029 on Build GRCh38
rs562361365 Chr.17: 21416228-21416228 on Build GRCh38
rs782501130 Chr.17: 21416128-21416128 on Build GRCh38
rs377513949 Chr.17: 21416031-21416031 on Build GRCh38
rs566882228 Chr.17: 21416084-21416084 on Build GRCh38
rs144590967 Chr.17: 21416139-21416139 on Build GRCh38
rs782160771 Chr.17: 21416028-21416028 on Build GRCh38
rs782564885 Chr.17: 21416178-21416178 on Build GRCh38
rs782286787 Chr.17: 21416121-21416121 on Build GRCh38
rs781994753 Chr.17: 21416194-21416194 on Build GRCh38
rs782745954 Chr.17: 21416192-21416192 on Build GRCh38
rs782016013 Chr.17: 21416216-21416216 on Build GRCh38
rs774272694 Chr.17: 21416077-21416077 on Build GRCh38
rs781823863 Chr.17: 21416227-21416227 on Build GRCh38
rs77176173 Chr.17: 21416176-21416176 on Build GRCh38
rs782159940 Chr.17: 21416074-21416074 on Build GRCh38
rs529503169 Chr.17: 21416108-21416108 on Build GRCh38
rs75316385 Chr.17: 21416086-21416086 on Build GRCh38
rs111301613 Chr.17: 21416134-21416134 on Build GRCh38
rs782327953 Chr.17: 21416232-21416232 on Build GRCh38
rs137932106 Chr.17: 21416129-21416129 on Build GRCh38
rs140875968 Chr.17: 21416076-21416076 on Build GRCh38
rs782305103 Chr.17: 21416149-21416149 on Build GRCh38
rs140687028 Chr.17: 21416146-21416146 on Build GRCh38
rs571047120 Chr.17: 21416113-21416113 on Build GRCh38
rs782078935 Chr.17: 21416078-21416078 on Build GRCh38
rs111560549 Chr.17: 21416179-21416179 on Build GRCh38
rs77987694 Chr.17: 21416211-21416211 on Build GRCh38
rs782145166 Chr.17: 21416151-21416151 on Build GRCh38
rs782780500 Chr.17: 21416039-21416039 on Build GRCh38
rs559211343 Chr.17: 21416107-21416107 on Build GRCh38
rs782044119 Chr.17: 21416118-21416118 on Build GRCh38
rs782189078 Chr.17: 21416189-21416189 on Build GRCh38
rs782287074 Chr.17: 21416104-21416104 on Build GRCh38
rs782524957 Chr.17: 21416089-21416089 on Build GRCh38
rs781932711 Chr.17: 21416150-21416150 on Build GRCh38
rs782490933 Chr.17: 21416045-21416045 on Build GRCh38
rs782343858 Chr.17: 21416064-21416064 on Build GRCh38
rs111766997 Chr.17: 21416030-21416030 on Build GRCh38
rs782165680 Chr.17: 21416217-21416217 on Build GRCh38
rs782226777 Chr.17: 21416148-21416148 on Build GRCh38
rs782611604 Chr.17: 21416090-21416090 on Build GRCh38
rs782751588 Chr.17: 21416032-21416032 on Build GRCh38
rs782069649 Chr.17: 21416164-21416164 on Build GRCh38
rs782582814 Chr.17: 21416103-21416103 on Build GRCh38
rs373382377 Chr.17: 21416027-21416027 on Build GRCh38
rs368451057 Chr.17: 21416065-21416065 on Build GRCh38
rs781886406 Chr.17: 21416123-21416123 on Build GRCh38
rs374137921 Chr.17: 21416056-21416056 on Build GRCh38
rs782257483 Chr.17: 21416186-21416186 on Build GRCh38
rs782005516 Chr.17: 21416157-21416157 on Build GRCh38
rs782794400 Chr.17: 21416198-21416198 on Build GRCh38
rs147282721 Chr.17: 21416196-21416196 on Build GRCh38
rs561450849 Chr.17: 21416099-21416100 on Build GRCh38

View Full Product Details