Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 245082102-245082327 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCAB2
Entrez Gene ID
84288
Gene Name
EF-hand calcium binding domain 2
Gene Aliases
CFAP200, DRC8
UniGene
Hs.134857
Gene Chromosome Location
Chr.1: 244969739-245125228 on Build GRCh38
Transcripts
NM_001143943.1 NM_001290327.1 NM_032328.3 NR_026587.1 XM_011544300.2 XM_011544301.1 XM_011544302.2 XM_017002538.1 XM_017002539.1 XM_017002540.1 XM_017002541.1 XM_017002542.1 XM_017002543.1 XM_017002544.1 XM_017002545.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5380316 c.637C>T p.L213F Chr.1: 245082128-245082128 on Build GRCh38
7365878 c.233C>T p.P78L Chr.1: 245082132-245082132 on Build GRCh38
2051556 c.220G>A p.E74K Chr.1: 245082119-245082119 on Build GRCh38
7136218 c.215G>T p.R72L Chr.1: 245082114-245082114 on Build GRCh38
1748277 c.256G>C p.E86Q Chr.1: 245082155-245082155 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754898871 Chr.1: 245082161-245082164 on Build GRCh38
rs747189599 Chr.1: 245082183-245082183 on Build GRCh38
rs963309705 Chr.1: 245082285-245082285 on Build GRCh38
rs767268130 Chr.1: 245082133-245082133 on Build GRCh38
rs762808379 Chr.1: 245082199-245082199 on Build GRCh38
rs775167425 Chr.1: 245082134-245082134 on Build GRCh38
rs552786395 Chr.1: 245082294-245082294 on Build GRCh38
rs777961622 Chr.1: 245082149-245082149 on Build GRCh38
rs139647250 Chr.1: 245082245-245082245 on Build GRCh38
rs772822033 Chr.1: 245082194-245082194 on Build GRCh38
rs772187066 Chr.1: 245082171-245082171 on Build GRCh38
rs35596803 Chr.1: 245082273-245082273 on Build GRCh38
rs776609813 Chr.1: 245082268-245082268 on Build GRCh38
rs745934700 Chr.1: 245082167-245082167 on Build GRCh38
rs1001548218 Chr.1: 245082189-245082189 on Build GRCh38
rs550896535 Chr.1: 245082231-245082231 on Build GRCh38
rs1055732212 Chr.1: 245082214-245082214 on Build GRCh38
rs531126702 Chr.1: 245082230-245082230 on Build GRCh38
rs768597228 Chr.1: 245082256-245082256 on Build GRCh38
rs781224893 Chr.1: 245082160-245082161 on Build GRCh38
rs761305694 Chr.1: 245082305-245082305 on Build GRCh38
rs768941322 Chr.1: 245082184-245082184 on Build GRCh38
rs7551261 Chr.1: 245082132-245082132 on Build GRCh38
rs950836298 Chr.1: 245082255-245082255 on Build GRCh38
rs571582954 Chr.1: 245082226-245082226 on Build GRCh38
rs775657519 Chr.1: 245082173-245082173 on Build GRCh38
rs757648347 Chr.1: 245082157-245082157 on Build GRCh38
rs779540752 Chr.1: 245082164-245082164 on Build GRCh38
rs756471739 Chr.1: 245082136-245082136 on Build GRCh38
rs536901040 Chr.1: 245082271-245082271 on Build GRCh38
rs749686955 Chr.1: 245082150-245082150 on Build GRCh38

View Full Product Details