Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 122303789-122304024 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FEZF1
Entrez Gene ID
389549
Gene Name
FEZ family zinc finger 1
Gene Aliases
FEZ, HH22, ZNF312B
UniGene
Hs.553970
Gene Chromosome Location
Chr.7: 122300939-122311855 on Build GRCh38
Transcripts
NM_001024613.3 NM_001160264.2 XM_005250337.3 XM_011516202.2 NR_036484.1

Gene Symbol
FEZF1-AS1
Entrez Gene ID
154860
Gene Name
FEZF1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.664204
Gene Chromosome Location
Chr.7: 122303658-122310077 on Build GRCh38
Transcripts
NM_001024613.3 NM_001160264.2 XM_005250337.3 XM_011516202.2 NR_036484.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6211828 c.585A>C p.K195N Chr.7: 122303853-122303853 on Build GRCh38
1330129 c.573C>A p.H191Q Chr.7: 122303865-122303865 on Build GRCh38
320408 c.598G>T p.E200* Chr.7: 122303840-122303840 on Build GRCh38
7111840 c.628G>C p.E210Q Chr.7: 122303810-122303810 on Build GRCh38
3877657 c.603G>A p.R201R Chr.7: 122303835-122303835 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771460949 Chr.7: 122303989-122303989 on Build GRCh38
rs780783637 Chr.7: 122303841-122303841 on Build GRCh38
rs747228982 Chr.7: 122303975-122303975 on Build GRCh38
rs771407843 Chr.7: 122303913-122303913 on Build GRCh38
rs370467502 Chr.7: 122303885-122303885 on Build GRCh38
rs755972168 Chr.7: 122303944-122303944 on Build GRCh38
rs570196942 Chr.7: 122303915-122303915 on Build GRCh38
rs761248281 Chr.7: 122303935-122303935 on Build GRCh38
rs759906335 Chr.7: 122303994-122303994 on Build GRCh38
rs757983995 Chr.7: 122303823-122303823 on Build GRCh38
rs893670572 Chr.7: 122303912-122303912 on Build GRCh38
rs781242932 Chr.7: 122303863-122303864 on Build GRCh38
rs754892083 Chr.7: 122303956-122303956 on Build GRCh38
rs751275983 Chr.7: 122303836-122303836 on Build GRCh38
rs749180825 Chr.7: 122303877-122303877 on Build GRCh38
rs778198481 Chr.7: 122303973-122303973 on Build GRCh38
rs772221066 Chr.7: 122303972-122303972 on Build GRCh38
rs1025604327 Chr.7: 122303963-122303963 on Build GRCh38
rs780100491 Chr.7: 122303851-122303851 on Build GRCh38
rs778866185 Chr.7: 122303965-122303965 on Build GRCh38
rs761133760 Chr.7: 122303897-122303897 on Build GRCh38
rs753860361 Chr.7: 122303952-122303952 on Build GRCh38
rs751063817 Chr.7: 122303925-122303925 on Build GRCh38
rs1054042595 Chr.7: 122303886-122303886 on Build GRCh38
rs763505250 Chr.7: 122303825-122303825 on Build GRCh38
rs748239139 Chr.7: 122303968-122303968 on Build GRCh38
rs374950795 Chr.7: 122304000-122304000 on Build GRCh38
rs866141957 Chr.7: 122303998-122303998 on Build GRCh38
rs754240132 Chr.7: 122303816-122303816 on Build GRCh38
rs779686674 Chr.7: 122303946-122303946 on Build GRCh38
rs776884375 Chr.7: 122303991-122303991 on Build GRCh38
rs773497993 Chr.7: 122303894-122303894 on Build GRCh38
rs938528614 Chr.7: 122303848-122303848 on Build GRCh38
rs1045001044 Chr.7: 122303932-122303932 on Build GRCh38
rs867706113 Chr.7: 122303967-122303967 on Build GRCh38
rs538115706 Chr.7: 122303887-122303887 on Build GRCh38
rs74961049 Chr.7: 122303842-122303842 on Build GRCh38
rs750065914 Chr.7: 122303937-122303937 on Build GRCh38
rs371371960 Chr.7: 122303943-122303943 on Build GRCh38
rs772451169 Chr.7: 122303890-122303890 on Build GRCh38
rs534580940 Chr.7: 122303923-122303923 on Build GRCh38
rs768824348 Chr.7: 122303878-122303878 on Build GRCh38
rs750619742 Chr.7: 122303888-122303890 on Build GRCh38
rs759927643 Chr.7: 122303922-122303922 on Build GRCh38
rs756926183 Chr.7: 122303840-122303840 on Build GRCh38
rs1048562563 Chr.7: 122303958-122303958 on Build GRCh38

View Full Product Details