Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 31605934-31606207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ID1
Entrez Gene ID
3397
Gene Name
inhibitor of DNA binding 1, HLH protein
Gene Aliases
ID, bHLHb24
UniGene
Hs.504609
Gene Chromosome Location
Chr.20: 31605283-31606515 on Build GRCh38
Transcripts
NM_002165.3 NM_181353.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1713314 c.448G>A p.D150N Chr.20: 31606074-31606074 on Build GRCh38
6558644 c.444G>T p.A148A Chr.20: 31606070-31606070 on Build GRCh38
3405002 c.427-1G>A p.? Chr.20: 31606052-31606052 on Build GRCh38
4774238 c.464G>A p.R155H Chr.20: 31606090-31606090 on Build GRCh38
6283538 c.431C>T p.A144V Chr.20: 31606057-31606057 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs375753998 Chr.20: 31606042-31606042 on Build GRCh38
rs577102752 Chr.20: 31606020-31606020 on Build GRCh38
rs558742055 Chr.20: 31606002-31606002 on Build GRCh38
rs775744042 Chr.20: 31606024-31606024 on Build GRCh38
rs761909488 Chr.20: 31606141-31606141 on Build GRCh38
rs944424815 Chr.20: 31606105-31606105 on Build GRCh38
rs921562569 Chr.20: 31606109-31606109 on Build GRCh38
rs202034102 Chr.20: 31606013-31606013 on Build GRCh38
rs771424453 Chr.20: 31606014-31606014 on Build GRCh38
rs567050825 Chr.20: 31605975-31605975 on Build GRCh38
rs767431877 Chr.20: 31606027-31606027 on Build GRCh38
rs776583398 Chr.20: 31606018-31606018 on Build GRCh38
rs931877363 Chr.20: 31605999-31605999 on Build GRCh38
rs763329754 Chr.20: 31606053-31606053 on Build GRCh38
rs8116155 Chr.20: 31606151-31606151 on Build GRCh38
rs754611809 Chr.20: 31606034-31606034 on Build GRCh38
rs781382541 Chr.20: 31606111-31606111 on Build GRCh38
rs115136221 Chr.20: 31606152-31606152 on Build GRCh38
rs774673437 Chr.20: 31606136-31606136 on Build GRCh38
rs201531919 Chr.20: 31606030-31606030 on Build GRCh38
rs749322712 Chr.20: 31606131-31606131 on Build GRCh38
rs375037749 Chr.20: 31606154-31606154 on Build GRCh38
rs755045570 Chr.20: 31606123-31606124 on Build GRCh38
rs769021844 Chr.20: 31606134-31606134 on Build GRCh38
rs373127064 Chr.20: 31606007-31606007 on Build GRCh38
rs773544843 Chr.20: 31606011-31606011 on Build GRCh38
rs753341526 Chr.20: 31606071-31606071 on Build GRCh38
rs780281435 Chr.20: 31606130-31606130 on Build GRCh38
rs1012250246 Chr.20: 31606180-31606180 on Build GRCh38
rs200627717 Chr.20: 31606065-31606065 on Build GRCh38
rs559514698 Chr.20: 31606121-31606121 on Build GRCh38
rs538900346 Chr.20: 31605998-31605998 on Build GRCh38
rs760508233 Chr.20: 31606068-31606068 on Build GRCh38
rs757604081 Chr.20: 31606091-31606091 on Build GRCh38
rs746442620 Chr.20: 31606127-31606127 on Build GRCh38
rs750414039 Chr.20: 31606066-31606066 on Build GRCh38
rs896261355 Chr.20: 31606043-31606043 on Build GRCh38
rs570073683 Chr.20: 31606171-31606171 on Build GRCh38
rs562697633 Chr.20: 31606077-31606077 on Build GRCh38
rs138737768 Chr.20: 31605964-31605964 on Build GRCh38
rs369242443 Chr.20: 31605960-31605960 on Build GRCh38
rs116636122 Chr.20: 31606166-31606166 on Build GRCh38
rs760732929 Chr.20: 31606146-31606146 on Build GRCh38
rs778178353 Chr.20: 31606035-31606035 on Build GRCh38
rs200894288 Chr.20: 31606100-31606100 on Build GRCh38
rs192792677 Chr.20: 31605955-31605955 on Build GRCh38
rs771359656 Chr.20: 31606040-31606040 on Build GRCh38
rs748991966 Chr.20: 31606032-31606032 on Build GRCh38
rs768929678 Chr.20: 31606054-31606054 on Build GRCh38
rs751106947 Chr.20: 31606165-31606166 on Build GRCh38
rs554749102 Chr.20: 31606023-31606023 on Build GRCh38
rs765973693 Chr.20: 31606069-31606069 on Build GRCh38
rs370401552 Chr.20: 31606055-31606055 on Build GRCh38
rs531514924 Chr.20: 31606143-31606143 on Build GRCh38
rs931658353 Chr.20: 31606113-31606113 on Build GRCh38
rs61731022 Chr.20: 31605967-31605967 on Build GRCh38
rs138944353 Chr.20: 31606035-31606037 on Build GRCh38
rs915372159 Chr.20: 31605958-31605958 on Build GRCh38
rs760486496 Chr.20: 31606037-31606037 on Build GRCh38
rs978646170 Chr.20: 31606082-31606082 on Build GRCh38
rs532455191 Chr.20: 31606051-31606051 on Build GRCh38
rs749766178 Chr.20: 31606083-31606085 on Build GRCh38
rs745969071 Chr.20: 31606047-31606047 on Build GRCh38
rs12480444 Chr.20: 31605982-31605982 on Build GRCh38
rs775696039 Chr.20: 31606052-31606052 on Build GRCh38
rs976081359 Chr.20: 31605992-31605992 on Build GRCh38
rs975875418 Chr.20: 31606108-31606108 on Build GRCh38
rs988823967 Chr.20: 31605965-31605965 on Build GRCh38
rs778483048 Chr.20: 31606079-31606079 on Build GRCh38
rs374966633 Chr.20: 31606144-31606144 on Build GRCh38
rs770194701 Chr.20: 31606128-31606128 on Build GRCh38
rs574815536 Chr.20: 31606026-31606026 on Build GRCh38
rs575404897 Chr.20: 31606009-31606009 on Build GRCh38
rs1022545309 Chr.20: 31606057-31606057 on Build GRCh38
rs372956084 Chr.20: 31606086-31606086 on Build GRCh38
rs749922542 Chr.20: 31606028-31606028 on Build GRCh38
rs767383808 Chr.20: 31606059-31606059 on Build GRCh38

View Full Product Details