Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 74501378-74501651 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LTBP2
Entrez Gene ID
4053
Gene Name
latent transforming growth factor beta binding protein 2
Gene Aliases
C14orf141, GLC3D, LTBP3, MSPKA, MSTP031, WMS3
UniGene
Hs.512776
Gene Chromosome Location
Chr.14: 74498183-74612331 on Build GRCh38
Transcripts
NM_000428.2 XM_011536765.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4052380 c.5251G>A p.V1751M Chr.14: 74501510-74501510 on Build GRCh38
3497931 c.5261G>A p.G1754D Chr.14: 74501500-74501500 on Build GRCh38
6704883 c.5202G>A p.A1734A Chr.14: 74501559-74501559 on Build GRCh38
5040746 c.5235T>G p.N1745K Chr.14: 74501526-74501526 on Build GRCh38
5848518 c.5248C>T p.R1750C Chr.14: 74501513-74501513 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759870336 Chr.14: 74501579-74501579 on Build GRCh38
rs201736724 Chr.14: 74501511-74501511 on Build GRCh38
rs1016201705 Chr.14: 74501439-74501439 on Build GRCh38
rs149242944 Chr.14: 74501510-74501510 on Build GRCh38
rs777633134 Chr.14: 74501424-74501424 on Build GRCh38
rs972060587 Chr.14: 74501497-74501497 on Build GRCh38
rs780081960 Chr.14: 74501513-74501513 on Build GRCh38
rs1024045703 Chr.14: 74501415-74501417 on Build GRCh38
rs921853911 Chr.14: 74501564-74501564 on Build GRCh38
rs138556118 Chr.14: 74501461-74501461 on Build GRCh38
rs1051538171 Chr.14: 74501589-74501589 on Build GRCh38
rs200772274 Chr.14: 74501538-74501538 on Build GRCh38
rs181190434 Chr.14: 74501620-74501620 on Build GRCh38
rs777241198 Chr.14: 74501550-74501550 on Build GRCh38
rs755989532 Chr.14: 74501512-74501512 on Build GRCh38
rs1041406215 Chr.14: 74501576-74501576 on Build GRCh38
rs766970271 Chr.14: 74501476-74501476 on Build GRCh38
rs771546236 Chr.14: 74501608-74501609 on Build GRCh38
rs374869558 Chr.14: 74501506-74501506 on Build GRCh38
rs779067555 Chr.14: 74501531-74501531 on Build GRCh38
rs201787635 Chr.14: 74501496-74501496 on Build GRCh38
rs758548129 Chr.14: 74501588-74501588 on Build GRCh38
rs749061798 Chr.14: 74501522-74501522 on Build GRCh38
rs377616549 Chr.14: 74501582-74501582 on Build GRCh38
rs549957928 Chr.14: 74501460-74501460 on Build GRCh38
rs200392077 Chr.14: 74501489-74501489 on Build GRCh38
rs945674645 Chr.14: 74501587-74501587 on Build GRCh38
rs772174775 Chr.14: 74501465-74501465 on Build GRCh38
rs761570723 Chr.14: 74501561-74501561 on Build GRCh38
rs754146202 Chr.14: 74501483-74501483 on Build GRCh38
rs765723436 Chr.14: 74501559-74501559 on Build GRCh38
rs745522540 Chr.14: 74501553-74501555 on Build GRCh38
rs758112209 Chr.14: 74501417-74501417 on Build GRCh38
rs749894277 Chr.14: 74501573-74501573 on Build GRCh38
rs886050755 Chr.14: 74501492-74501492 on Build GRCh38
rs767256407 Chr.14: 74501571-74501571 on Build GRCh38
rs773705689 Chr.14: 74501468-74501468 on Build GRCh38
rs370987753 Chr.14: 74501606-74501606 on Build GRCh38
rs528254230 Chr.14: 74501526-74501526 on Build GRCh38
rs761102550 Chr.14: 74501469-74501469 on Build GRCh38
rs563278459 Chr.14: 74501412-74501412 on Build GRCh38
rs774080243 Chr.14: 74501560-74501560 on Build GRCh38
rs747998553 Chr.14: 74501586-74501586 on Build GRCh38
rs376019974 Chr.14: 74501619-74501619 on Build GRCh38
rs761832980 Chr.14: 74501463-74501463 on Build GRCh38
rs769564522 Chr.14: 74501459-74501459 on Build GRCh38
rs751080389 Chr.14: 74501431-74501431 on Build GRCh38
rs886050754 Chr.14: 74501491-74501491 on Build GRCh38
rs781130604 Chr.14: 74501445-74501445 on Build GRCh38
rs951375544 Chr.14: 74501521-74501521 on Build GRCh38
rs759927089 Chr.14: 74501552-74501552 on Build GRCh38
rs756787943 Chr.14: 74501507-74501507 on Build GRCh38
rs906921321 Chr.14: 74501600-74501600 on Build GRCh38
rs144298924 Chr.14: 74501607-74501607 on Build GRCh38
rs745823064 Chr.14: 74501453-74501453 on Build GRCh38
rs372981316 Chr.14: 74501580-74501580 on Build GRCh38
rs371338219 Chr.14: 74501444-74501444 on Build GRCh38
rs752338952 Chr.14: 74501414-74501414 on Build GRCh38
rs1001677999 Chr.14: 74501454-74501454 on Build GRCh38
rs149587738 Chr.14: 74501574-74501574 on Build GRCh38
rs773154880 Chr.14: 74501540-74501540 on Build GRCh38
rs748378774 Chr.14: 74501537-74501537 on Build GRCh38
rs199739772 Chr.14: 74501627-74501627 on Build GRCh38
rs760878019 Chr.14: 74501549-74501549 on Build GRCh38

View Full Product Details