Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 109565656-109565917 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RANBP2
Entrez Gene ID
5903
Gene Name
RAN binding protein 2
Gene Aliases
ADANE, ANE1, IIAE3, NUP358, TRP1, TRP2
UniGene
Hs.199561
Gene Chromosome Location
Chr.2: 108719446-109842301 on Build GRCh38
Transcripts
XM_017004623.1 NM_001321496.1 NM_001321498.1 NM_001321499.1 NM_001321500.1 NM_001321501.1 NM_001321502.1 NM_001321503.1 NM_001321504.1 NM_001321505.1 NM_001321506.1 NM_001321507.1 NM_001321508.1 NM_001321509.1 NM_001321510.1 NM_001321511.1 NM_001321512.1 NM_001321513.1 NM_001321514.1 NM_001321515.1 NM_144710.4 NM_178584.3 XM_006712317.1 XM_011510698.2 XM_011510699.2 XM_011510700.2 XM_011510701.2 XM_011510702.2 XM_011510703.2 XM_011510704.2

Gene Symbol
SEPT10
Entrez Gene ID
151011
Gene Name
septin 10
Gene Aliases
-
UniGene
Hs.469615
Gene Chromosome Location
Chr.2: 109542797-109614247 on Build GRCh38
Transcripts
XM_017004623.1 NM_001321496.1 NM_001321498.1 NM_001321499.1 NM_001321500.1 NM_001321501.1 NM_001321502.1 NM_001321503.1 NM_001321504.1 NM_001321505.1 NM_001321506.1 NM_001321507.1 NM_001321508.1 NM_001321509.1 NM_001321510.1 NM_001321511.1 NM_001321512.1 NM_001321513.1 NM_001321514.1 NM_001321515.1 NM_144710.4 NM_178584.3 XM_006712317.1 XM_011510698.2 XM_011510699.2 XM_011510700.2 XM_011510701.2 XM_011510702.2 XM_011510703.2 XM_011510704.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4128101 c.779G>T p.G260V Chr.2: 109565774-109565774 on Build GRCh38
4084137 c.767A>C p.Q256P Chr.2: 109565855-109565855 on Build GRCh38
1005331 c.782T>G p.V261G Chr.2: 109565771-109565771 on Build GRCh38
6385430 c.754G>T p.V252F Chr.2: 109565799-109565799 on Build GRCh38
714786 c.756C>G p.V252V Chr.2: 109565797-109565797 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200735743 Chr.2: 109565798-109565798 on Build GRCh38
rs986328394 Chr.2: 109565687-109565687 on Build GRCh38
rs1011377843 Chr.2: 109565838-109565838 on Build GRCh38
rs932204365 Chr.2: 109565800-109565800 on Build GRCh38
rs187997163 Chr.2: 109565714-109565714 on Build GRCh38
rs751689147 Chr.2: 109565777-109565777 on Build GRCh38
rs778686257 Chr.2: 109565875-109565875 on Build GRCh38
rs776016761 Chr.2: 109565842-109565842 on Build GRCh38
rs192464207 Chr.2: 109565803-109565803 on Build GRCh38
rs116719735 Chr.2: 109565733-109565733 on Build GRCh38
rs749478292 Chr.2: 109565848-109565848 on Build GRCh38
rs766275277 Chr.2: 109565804-109565805 on Build GRCh38
rs756070264 Chr.2: 109565861-109565861 on Build GRCh38
rs754707962 Chr.2: 109565874-109565874 on Build GRCh38
rs201356578 Chr.2: 109565811-109565811 on Build GRCh38
rs907560008 Chr.2: 109565769-109565769 on Build GRCh38
rs771470210 Chr.2: 109565815-109565815 on Build GRCh38
rs767502369 Chr.2: 109565850-109565850 on Build GRCh38
rs766949890 Chr.2: 109565729-109565729 on Build GRCh38
rs199570779 Chr.2: 109565847-109565847 on Build GRCh38
rs376369966 Chr.2: 109565849-109565849 on Build GRCh38
rs549803354 Chr.2: 109565870-109565870 on Build GRCh38
rs753042647 Chr.2: 109565771-109565771 on Build GRCh38
rs368621147 Chr.2: 109565790-109565790 on Build GRCh38
rs929419681 Chr.2: 109565784-109565784 on Build GRCh38
rs371694319 Chr.2: 109565791-109565791 on Build GRCh38
rs753730583 Chr.2: 109565765-109565765 on Build GRCh38
rs369518984 Chr.2: 109565812-109565812 on Build GRCh38
rs377202328 Chr.2: 109565789-109565789 on Build GRCh38
rs763185491 Chr.2: 109565845-109565845 on Build GRCh38
rs762002165 Chr.2: 109565722-109565725 on Build GRCh38
rs183258808 Chr.2: 109565885-109565885 on Build GRCh38
rs61999358 Chr.2: 109565853-109565853 on Build GRCh38
rs536588521 Chr.2: 109565837-109565837 on Build GRCh38
rs759350516 Chr.2: 109565845-109565845 on Build GRCh38
rs760123201 Chr.2: 109565832-109565832 on Build GRCh38
rs768387357 Chr.2: 109565807-109565807 on Build GRCh38
rs758706146 Chr.2: 109565774-109565774 on Build GRCh38
rs1007214829 Chr.2: 109565855-109565855 on Build GRCh38
rs757248499 Chr.2: 109565781-109565781 on Build GRCh38
rs773936474 Chr.2: 109565715-109565715 on Build GRCh38
rs1046944748 Chr.2: 109565785-109565785 on Build GRCh38
rs369310997 Chr.2: 109565802-109565802 on Build GRCh38
rs938030882 Chr.2: 109565805-109565805 on Build GRCh38
rs375032818 Chr.2: 109565726-109565726 on Build GRCh38
rs777344796 Chr.2: 109565829-109565829 on Build GRCh38
rs372556461 Chr.2: 109565748-109565748 on Build GRCh38
rs765692504 Chr.2: 109565770-109565770 on Build GRCh38
rs747751240 Chr.2: 109565880-109565880 on Build GRCh38
rs372952684 Chr.2: 109565871-109565871 on Build GRCh38
rs983308452 Chr.2: 109565763-109565763 on Build GRCh38
rs774189615 Chr.2: 109565831-109565831 on Build GRCh38
rs762601193 Chr.2: 109565712-109565712 on Build GRCh38
rs766495465 Chr.2: 109565692-109565692 on Build GRCh38
rs777805950 Chr.2: 109565775-109565775 on Build GRCh38

View Full Product Details