Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 19380059-19380320 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS6
Entrez Gene ID
6194
Gene Name
ribosomal protein S6
Gene Aliases
S6
UniGene
Hs.408073
Gene Chromosome Location
Chr.9: 19376256-19380237 on Build GRCh38
Transcripts
NM_001010.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4814505 c.3G>A p.M1I Chr.9: 19380193-19380193 on Build GRCh38
455748 c.3G>A p.M1I Chr.9: 19380193-19380193 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1002969188 Chr.9: 19380219-19380219 on Build GRCh38
rs201852243 Chr.9: 19380208-19380208 on Build GRCh38
rs938811233 Chr.9: 19380294-19380294 on Build GRCh38
rs993580840 Chr.9: 19380287-19380287 on Build GRCh38
rs371399724 Chr.9: 19380160-19380160 on Build GRCh38
rs909803493 Chr.9: 19380264-19380264 on Build GRCh38
rs565220729 Chr.9: 19380212-19380212 on Build GRCh38
rs1037699624 Chr.9: 19380280-19380280 on Build GRCh38
rs746750178 Chr.9: 19380159-19380159 on Build GRCh38
rs539124899 Chr.9: 19380246-19380246 on Build GRCh38
rs756983352 Chr.9: 19380183-19380183 on Build GRCh38
rs777671286 Chr.9: 19380155-19380155 on Build GRCh38
rs12342100 Chr.9: 19380240-19380240 on Build GRCh38
rs769224636 Chr.9: 19380166-19380166 on Build GRCh38
rs113302866 Chr.9: 19380189-19380189 on Build GRCh38
rs184998286 Chr.9: 19380202-19380202 on Build GRCh38
rs149565972 Chr.9: 19380211-19380211 on Build GRCh38
rs541296826 Chr.9: 19380181-19380181 on Build GRCh38
rs376914338 Chr.9: 19380235-19380235 on Build GRCh38
rs771826675 Chr.9: 19380228-19380228 on Build GRCh38
rs745326144 Chr.9: 19380247-19380247 on Build GRCh38
rs878869189 Chr.9: 19380136-19380136 on Build GRCh38
rs1055014827 Chr.9: 19380261-19380261 on Build GRCh38
rs200083793 Chr.9: 19380197-19380197 on Build GRCh38
rs753861014 Chr.9: 19380215-19380216 on Build GRCh38
rs374125503 Chr.9: 19380131-19380131 on Build GRCh38
rs551080353 Chr.9: 19380248-19380248 on Build GRCh38
rs776619444 Chr.9: 19380097-19380097 on Build GRCh38
rs369511403 Chr.9: 19380206-19380206 on Build GRCh38
rs542046649 Chr.9: 19380204-19380204 on Build GRCh38
rs758149120 Chr.9: 19380145-19380145 on Build GRCh38
rs757317851 Chr.9: 19380169-19380169 on Build GRCh38
rs1057274019 Chr.9: 19380302-19380302 on Build GRCh38
rs746229844 Chr.9: 19380230-19380230 on Build GRCh38
rs945217462 Chr.9: 19380103-19380103 on Build GRCh38
rs543861356 Chr.9: 19380248-19380248 on Build GRCh38
rs41270111 Chr.9: 19380207-19380207 on Build GRCh38
rs35050537 Chr.9: 19380274-19380275 on Build GRCh38
rs371429328 Chr.9: 19380173-19380173 on Build GRCh38
rs554845311 Chr.9: 19380227-19380227 on Build GRCh38
rs764427303 Chr.9: 19380245-19380245 on Build GRCh38
rs752898834 Chr.9: 19380143-19380143 on Build GRCh38
rs755646427 Chr.9: 19380187-19380187 on Build GRCh38
rs750198792 Chr.9: 19380216-19380216 on Build GRCh38
rs779071874 Chr.9: 19380233-19380234 on Build GRCh38
rs770389372 Chr.9: 19380231-19380231 on Build GRCh38
rs897852263 Chr.9: 19380283-19380283 on Build GRCh38
rs566082136 Chr.9: 19380270-19380270 on Build GRCh38
rs773862636 Chr.9: 19380198-19380198 on Build GRCh38
rs781718074 Chr.9: 19380229-19380229 on Build GRCh38
rs748924235 Chr.9: 19380170-19380170 on Build GRCh38
rs17523533 Chr.9: 19380234-19380234 on Build GRCh38
rs897910858 Chr.9: 19380222-19380222 on Build GRCh38
rs758686587 Chr.9: 19380180-19380180 on Build GRCh38
rs41270113 Chr.9: 19380241-19380241 on Build GRCh38
rs754267688 Chr.9: 19380301-19380301 on Build GRCh38
rs755753380 Chr.9: 19380112-19380112 on Build GRCh38
rs756121963 Chr.9: 19380218-19380218 on Build GRCh38
rs755240117 Chr.9: 19380230-19380231 on Build GRCh38
rs376100121 Chr.9: 19380164-19380164 on Build GRCh38
rs745393679 Chr.9: 19380165-19380165 on Build GRCh38
rs972279633 Chr.9: 19380265-19380265 on Build GRCh38
rs748089597 Chr.9: 19380125-19380125 on Build GRCh38
rs752223014 Chr.9: 19380244-19380244 on Build GRCh38
rs892268406 Chr.9: 19380087-19380087 on Build GRCh38
rs909459280 Chr.9: 19380203-19380203 on Build GRCh38
rs144218100 Chr.9: 19380221-19380221 on Build GRCh38
rs373625038 Chr.9: 19380118-19380118 on Build GRCh38
rs942315184 Chr.9: 19380262-19380262 on Build GRCh38
rs368693979 Chr.9: 19380172-19380172 on Build GRCh38
rs200133667 Chr.9: 19380243-19380243 on Build GRCh38
rs745762926 Chr.9: 19380184-19380184 on Build GRCh38
rs897971347 Chr.9: 19380214-19380214 on Build GRCh38
rs369300231 Chr.9: 19380236-19380236 on Build GRCh38
rs759695423 Chr.9: 19380175-19380175 on Build GRCh38
rs367798566 Chr.9: 19380199-19380199 on Build GRCh38
rs751410275 Chr.9: 19380182-19380182 on Build GRCh38
rs780190215 Chr.9: 19380169-19380170 on Build GRCh38
rs747764206 Chr.9: 19380223-19380223 on Build GRCh38
rs753446607 Chr.9: 19380220-19380220 on Build GRCh38
rs138274707 Chr.9: 19380093-19380093 on Build GRCh38
rs12001628 Chr.9: 19380174-19380174 on Build GRCh38
rs779666077 Chr.9: 19380196-19380196 on Build GRCh38
rs760377421 Chr.9: 19380239-19380239 on Build GRCh38
rs1036481503 Chr.9: 19380109-19380109 on Build GRCh38
rs765628443 Chr.9: 19380205-19380205 on Build GRCh38
rs199963241 Chr.9: 19380237-19380237 on Build GRCh38
rs201066924 Chr.9: 19380238-19380238 on Build GRCh38
rs769690365 Chr.9: 19380176-19380176 on Build GRCh38
rs775039096 Chr.9: 19380167-19380167 on Build GRCh38

View Full Product Details