Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 22573372-22573609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIP4K2A
Entrez Gene ID
5305
Gene Name
phosphatidylinositol-5-phosphate 4-kinase type 2 alpha
Gene Aliases
PI5P4KA, PIP5K2A, PIP5KII-alpha, PIP5KIIA, PIPK
UniGene
Hs.57079
Gene Chromosome Location
Chr.10: 22534837-22714574 on Build GRCh38
Transcripts
NM_005028.4 XM_006717450.2 XM_011519496.1 XM_017016330.1 XM_017016331.1 XM_017016332.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4864636 c.421G>T p.V141F Chr.10: 22573385-22573385 on Build GRCh38
917059 c.565G>T p.V189F Chr.10: 22573385-22573385 on Build GRCh38
4807975 c.403C>T p.R135W Chr.10: 22573403-22573403 on Build GRCh38
6530783 c.534C>G p.F178L Chr.10: 22573416-22573416 on Build GRCh38
1262224 c.534C>T p.F178F Chr.10: 22573416-22573416 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs974102162 Chr.10: 22573561-22573561 on Build GRCh38
rs760086351 Chr.10: 22573485-22573485 on Build GRCh38
rs1031679279 Chr.10: 22573504-22573504 on Build GRCh38
rs571857152 Chr.10: 22573475-22573475 on Build GRCh38
rs749030397 Chr.10: 22573463-22573463 on Build GRCh38
rs757358809 Chr.10: 22573508-22573508 on Build GRCh38
rs751409044 Chr.10: 22573572-22573572 on Build GRCh38
rs781130866 Chr.10: 22573403-22573403 on Build GRCh38
rs563593537 Chr.10: 22573517-22573517 on Build GRCh38
rs11013052 Chr.10: 22573558-22573558 on Build GRCh38
rs773991583 Chr.10: 22573470-22573470 on Build GRCh38
rs775128778 Chr.10: 22573454-22573454 on Build GRCh38
rs745767301 Chr.10: 22573408-22573408 on Build GRCh38
rs201077800 Chr.10: 22573409-22573409 on Build GRCh38
rs375709819 Chr.10: 22573515-22573515 on Build GRCh38
rs757271233 Chr.10: 22573402-22573402 on Build GRCh38
rs200441361 Chr.10: 22573502-22573502 on Build GRCh38
rs61847466 Chr.10: 22573573-22573573 on Build GRCh38
rs566255938 Chr.10: 22573539-22573539 on Build GRCh38
rs149851162 Chr.10: 22573540-22573540 on Build GRCh38
rs765681053 Chr.10: 22573486-22573486 on Build GRCh38
rs535087618 Chr.10: 22573530-22573530 on Build GRCh38
rs768193534 Chr.10: 22573465-22573465 on Build GRCh38
rs1023250774 Chr.10: 22573531-22573531 on Build GRCh38
rs762464468 Chr.10: 22573482-22573482 on Build GRCh38
rs752988288 Chr.10: 22573489-22573489 on Build GRCh38
rs186810454 Chr.10: 22573498-22573498 on Build GRCh38
rs12258324 Chr.10: 22573477-22573477 on Build GRCh38
rs763513174 Chr.10: 22573481-22573482 on Build GRCh38

View Full Product Details