Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 43025887-43026124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RND2
Entrez Gene ID
8153
Gene Name
Rho family GTPase 2
Gene Aliases
ARHN, RHO7, RhoN
UniGene
Hs.603111
Gene Chromosome Location
Chr.17: 43025241-43032041 on Build GRCh38
Transcripts
NM_005440.4 XM_011525316.1 XM_011525317.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4842330 c.188C>G p.S63* Chr.17: 43026045-43026045 on Build GRCh38
3517872 c.124G>A p.E42K Chr.17: 43025981-43025981 on Build GRCh38
1383495 c.157C>T p.R53C Chr.17: 43026014-43026014 on Build GRCh38
4721915 c.161G>A p.R54H Chr.17: 43026018-43026018 on Build GRCh38
223881 c.134C>T p.T45I Chr.17: 43025991-43025991 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763552235 Chr.17: 43026059-43026059 on Build GRCh38
rs780616312 Chr.17: 43025942-43025942 on Build GRCh38
rs933623795 Chr.17: 43025960-43025960 on Build GRCh38
rs762520087 Chr.17: 43026042-43026042 on Build GRCh38
rs201422969 Chr.17: 43025916-43025916 on Build GRCh38
rs765574637 Chr.17: 43025958-43025959 on Build GRCh38
rs780497902 Chr.17: 43026085-43026085 on Build GRCh38
rs769208841 Chr.17: 43025956-43025956 on Build GRCh38
rs756994280 Chr.17: 43026071-43026071 on Build GRCh38
rs751236062 Chr.17: 43026066-43026066 on Build GRCh38
rs559466260 Chr.17: 43025952-43025952 on Build GRCh38
rs749543833 Chr.17: 43025912-43025912 on Build GRCh38
rs778800457 Chr.17: 43025959-43025959 on Build GRCh38
rs773448266 Chr.17: 43025990-43025990 on Build GRCh38
rs776444727 Chr.17: 43026015-43026015 on Build GRCh38
rs866202135 Chr.17: 43025972-43025972 on Build GRCh38
rs777975287 Chr.17: 43025945-43025946 on Build GRCh38
rs746726283 Chr.17: 43025930-43025930 on Build GRCh38
rs752638563 Chr.17: 43026032-43026032 on Build GRCh38
rs370206545 Chr.17: 43025917-43025917 on Build GRCh38
rs756518252 Chr.17: 43025933-43025933 on Build GRCh38
rs765237092 Chr.17: 43026026-43026026 on Build GRCh38
rs11553384 Chr.17: 43025994-43025994 on Build GRCh38
rs777469311 Chr.17: 43025927-43025927 on Build GRCh38
rs866917036 Chr.17: 43025965-43025965 on Build GRCh38
rs754927507 Chr.17: 43025910-43025910 on Build GRCh38
rs759404812 Chr.17: 43026025-43026025 on Build GRCh38
rs749771815 Chr.17: 43025947-43025947 on Build GRCh38
rs537385290 Chr.17: 43026094-43026094 on Build GRCh38
rs374133872 Chr.17: 43025946-43025946 on Build GRCh38
rs772107613 Chr.17: 43025979-43025979 on Build GRCh38
rs748245588 Chr.17: 43025975-43025975 on Build GRCh38
rs770660339 Chr.17: 43025997-43025997 on Build GRCh38
rs754265423 Chr.17: 43026086-43026086 on Build GRCh38

View Full Product Details