Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 191034453-191034686 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STAT4
Entrez Gene ID
6775
Gene Name
signal transducer and activator of transcription 4
Gene Aliases
SLEB11
UniGene
Hs.735572
Gene Chromosome Location
Chr.2: 191029576-191172684 on Build GRCh38
Transcripts
NM_001243835.1 NM_003151.3 XM_006712719.3 XM_011511705.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
110501 c.1620+1G>A p.? Chr.2: 191034547-191034547 on Build GRCh38
5663730 c.1586G>C p.S529T Chr.2: 191034582-191034582 on Build GRCh38
718789 c.1611G>C p.K537N Chr.2: 191034557-191034557 on Build GRCh38
3575610 c.1591G>A p.G531S Chr.2: 191034577-191034577 on Build GRCh38
7279312 c.1620+9G>A p.? Chr.2: 191034539-191034539 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147850919 Chr.2: 191034572-191034572 on Build GRCh38
rs745523665 Chr.2: 191034518-191034518 on Build GRCh38
rs748620825 Chr.2: 191034634-191034634 on Build GRCh38
rs745844883 Chr.2: 191034506-191034507 on Build GRCh38
rs775485683 Chr.2: 191034532-191034532 on Build GRCh38
rs896917147 Chr.2: 191034493-191034493 on Build GRCh38
rs751680956 Chr.2: 191034497-191034497 on Build GRCh38
rs756723247 Chr.2: 191034638-191034638 on Build GRCh38
rs774187563 Chr.2: 191034552-191034552 on Build GRCh38
rs369929926 Chr.2: 191034535-191034535 on Build GRCh38
rs955798457 Chr.2: 191034628-191034628 on Build GRCh38
rs768445696 Chr.2: 191034584-191034584 on Build GRCh38
rs200772241 Chr.2: 191034539-191034539 on Build GRCh38
rs551924843 Chr.2: 191034514-191034514 on Build GRCh38
rs776369911 Chr.2: 191034517-191034517 on Build GRCh38
rs780464694 Chr.2: 191034510-191034510 on Build GRCh38
rs746565637 Chr.2: 191034516-191034516 on Build GRCh38
rs750692459 Chr.2: 191034507-191034507 on Build GRCh38
rs754970873 Chr.2: 191034610-191034610 on Build GRCh38
rs369477828 Chr.2: 191034482-191034482 on Build GRCh38
rs754247387 Chr.2: 191034615-191034615 on Build GRCh38
rs368894302 Chr.2: 191034642-191034642 on Build GRCh38
rs545292550 Chr.2: 191034600-191034600 on Build GRCh38
rs3024889 Chr.2: 191034620-191034620 on Build GRCh38
rs190854635 Chr.2: 191034536-191034536 on Build GRCh38
rs116809401 Chr.2: 191034631-191034631 on Build GRCh38
rs73981214 Chr.2: 191034509-191034509 on Build GRCh38
rs757380228 Chr.2: 191034499-191034499 on Build GRCh38
rs767596903 Chr.2: 191034571-191034571 on Build GRCh38
rs781444658 Chr.2: 191034504-191034504 on Build GRCh38
rs769711196 Chr.2: 191034529-191034529 on Build GRCh38
rs760335389 Chr.2: 191034582-191034582 on Build GRCh38
rs992959125 Chr.2: 191034593-191034593 on Build GRCh38
rs749789837 Chr.2: 191034513-191034513 on Build GRCh38
rs148688926 Chr.2: 191034547-191034547 on Build GRCh38
rs761904937 Chr.2: 191034562-191034562 on Build GRCh38
rs997523285 Chr.2: 191034614-191034614 on Build GRCh38

View Full Product Details