Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154764798-154765067 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKC1
Entrez Gene ID
1736
Gene Name
dyskerin pseudouridine synthase 1
Gene Aliases
CBF5, DKC, DKCX, NAP57, NOLA4, XAP101
UniGene
Hs.4747
Gene Chromosome Location
Chr.X: 154762742-154777689 on Build GRCh38
Transcripts
NM_001142463.2 NM_001288747.1 NM_001363.4 NR_110021.1 NR_110022.1 NR_110023.1

SNP Detail

SNP ID Chromosome Location
rs782231492 Chr.X: 154764885-154764885 on Build GRCh38
rs199422242 Chr.X: 154764911-154764911 on Build GRCh38
rs782010351 Chr.X: 154764937-154764937 on Build GRCh38
rs782408824 Chr.X: 154764862-154764862 on Build GRCh38
rs1015273682 Chr.X: 154765025-154765025 on Build GRCh38
rs781840691 Chr.X: 154764889-154764890 on Build GRCh38
rs961071971 Chr.X: 154765030-154765030 on Build GRCh38
rs186546647 Chr.X: 154764844-154764844 on Build GRCh38
rs781794417 Chr.X: 154764979-154764979 on Build GRCh38
rs782017760 Chr.X: 154765008-154765008 on Build GRCh38
rs369105166 Chr.X: 154765004-154765004 on Build GRCh38
rs782389383 Chr.X: 154764944-154764944 on Build GRCh38
rs782005369 Chr.X: 154764867-154764867 on Build GRCh38
rs142410621 Chr.X: 154764829-154764829 on Build GRCh38
rs782718798 Chr.X: 154764976-154764976 on Build GRCh38
rs781849751 Chr.X: 154764946-154764946 on Build GRCh38
rs782259079 Chr.X: 154764999-154764999 on Build GRCh38
rs782623843 Chr.X: 154764827-154764827 on Build GRCh38
rs782121373 Chr.X: 154764938-154764938 on Build GRCh38
rs903674446 Chr.X: 154764846-154764846 on Build GRCh38
rs199975650 Chr.X: 154764985-154764985 on Build GRCh38
rs1011619375 Chr.X: 154764833-154764833 on Build GRCh38
rs782678759 Chr.X: 154764991-154764991 on Build GRCh38
rs782044075 Chr.X: 154764921-154764921 on Build GRCh38
rs782343800 Chr.X: 154764902-154764902 on Build GRCh38
rs376632263 Chr.X: 154764923-154764923 on Build GRCh38
rs782293840 Chr.X: 154764860-154764860 on Build GRCh38
rs781936419 Chr.X: 154764918-154764918 on Build GRCh38
rs142474971 Chr.X: 154764966-154764966 on Build GRCh38

View Full Product Details