Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 5862881-5863140 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C20orf196
Entrez Gene ID
149840
Gene Name
chromosome 20 open reading frame 196
Gene Aliases
-
UniGene
Hs.529340
Gene Chromosome Location
Chr.20: 5750286-5864407 on Build GRCh38
Transcripts
NM_001303477.1 NM_001303478.1 NM_152504.3 XM_006723549.3 XM_011529177.2 XM_017027684.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3423765 c.199G>A p.E67K Chr.20: 5863044-5863044 on Build GRCh38
3758666 c.219C>T p.T73T Chr.20: 5863064-5863064 on Build GRCh38
3548387 c.267G>A p.K89K Chr.20: 5863112-5863112 on Build GRCh38
1028595 c.184A>C p.S62R Chr.20: 5863029-5863029 on Build GRCh38
4929446 c.220G>A p.A74T Chr.20: 5863065-5863065 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778421677 Chr.20: 5863042-5863042 on Build GRCh38
rs770939329 Chr.20: 5862987-5862987 on Build GRCh38
rs768862384 Chr.20: 5863010-5863010 on Build GRCh38
rs774532609 Chr.20: 5863017-5863017 on Build GRCh38
rs750842218 Chr.20: 5863029-5863029 on Build GRCh38
rs766972128 Chr.20: 5863038-5863038 on Build GRCh38
rs1503896 Chr.20: 5862931-5862931 on Build GRCh38
rs761955136 Chr.20: 5863120-5863120 on Build GRCh38
rs745824977 Chr.20: 5863100-5863100 on Build GRCh38
rs145271595 Chr.20: 5863081-5863081 on Build GRCh38
rs771372649 Chr.20: 5863010-5863011 on Build GRCh38
rs758092845 Chr.20: 5863065-5863065 on Build GRCh38
rs777372428 Chr.20: 5863067-5863067 on Build GRCh38
rs981060257 Chr.20: 5863089-5863089 on Build GRCh38
rs114118253 Chr.20: 5863026-5863026 on Build GRCh38
rs147402348 Chr.20: 5863035-5863035 on Build GRCh38
rs201976741 Chr.20: 5863115-5863115 on Build GRCh38
rs41282136 Chr.20: 5863064-5863064 on Build GRCh38
rs151170855 Chr.20: 5862984-5862984 on Build GRCh38
rs777564276 Chr.20: 5862976-5862976 on Build GRCh38
rs776426706 Chr.20: 5863004-5863004 on Build GRCh38
rs767826318 Chr.20: 5863027-5863027 on Build GRCh38
rs376695335 Chr.20: 5863099-5863099 on Build GRCh38
rs748962924 Chr.20: 5862969-5862969 on Build GRCh38
rs746886404 Chr.20: 5863075-5863075 on Build GRCh38
rs763149662 Chr.20: 5863009-5863009 on Build GRCh38
rs1037100048 Chr.20: 5863030-5863030 on Build GRCh38
rs755442162 Chr.20: 5863041-5863041 on Build GRCh38
rs888298088 Chr.20: 5862928-5862928 on Build GRCh38
rs776970446 Chr.20: 5863073-5863075 on Build GRCh38
rs746342498 Chr.20: 5863072-5863073 on Build GRCh38

View Full Product Details