Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 36399424-36399697 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHAF1B
Entrez Gene ID
8208
Gene Name
chromatin assembly factor 1 subunit B
Gene Aliases
CAF-1, CAF-IP60, CAF1, CAF1A, CAF1P60, MPHOSPH7, MPP7
UniGene
Hs.75238
Gene Chromosome Location
Chr.21: 36380145-36416827 on Build GRCh38
Transcripts
NM_005441.2 XM_011529755.2 XM_017028477.1 XM_017028478.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1483929 c.591A>G p.V197V Chr.21: 36399533-36399533 on Build GRCh38
224075 c.652G>A p.A218T Chr.21: 36399594-36399594 on Build GRCh38
6627700 c.586C>T p.R196* Chr.21: 36399528-36399528 on Build GRCh38
6627701 c.587G>A p.R196Q Chr.21: 36399529-36399529 on Build GRCh38
3550542 c.587G>T p.R196L Chr.21: 36399529-36399529 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745473711 Chr.21: 36399522-36399522 on Build GRCh38
rs374984835 Chr.21: 36399482-36399482 on Build GRCh38
rs1051925992 Chr.21: 36399576-36399576 on Build GRCh38
rs759798573 Chr.21: 36399558-36399558 on Build GRCh38
rs778650359 Chr.21: 36399590-36399590 on Build GRCh38
rs768015697 Chr.21: 36399626-36399626 on Build GRCh38
rs375539352 Chr.21: 36399524-36399524 on Build GRCh38
rs752862735 Chr.21: 36399473-36399473 on Build GRCh38
rs569287285 Chr.21: 36399543-36399543 on Build GRCh38
rs1016562330 Chr.21: 36399654-36399654 on Build GRCh38
rs778656976 Chr.21: 36399648-36399649 on Build GRCh38
rs755002708 Chr.21: 36399626-36399627 on Build GRCh38
rs756935581 Chr.21: 36399506-36399506 on Build GRCh38
rs988120393 Chr.21: 36399633-36399633 on Build GRCh38
rs967102352 Chr.21: 36399483-36399483 on Build GRCh38
rs368254916 Chr.21: 36399656-36399656 on Build GRCh38
rs776230418 Chr.21: 36399637-36399637 on Build GRCh38
rs866842640 Chr.21: 36399591-36399591 on Build GRCh38
rs760033114 Chr.21: 36399651-36399651 on Build GRCh38
rs756088596 Chr.21: 36399571-36399571 on Build GRCh38
rs774908427 Chr.21: 36399557-36399557 on Build GRCh38
rs764238425 Chr.21: 36399572-36399572 on Build GRCh38
rs753676556 Chr.21: 36399581-36399581 on Build GRCh38
rs958204278 Chr.21: 36399508-36399508 on Build GRCh38
rs934576838 Chr.21: 36399481-36399481 on Build GRCh38
rs548768762 Chr.21: 36399501-36399501 on Build GRCh38
rs149193400 Chr.21: 36399537-36399537 on Build GRCh38
rs899426108 Chr.21: 36399668-36399668 on Build GRCh38
rs377510748 Chr.21: 36399469-36399469 on Build GRCh38
rs774849385 Chr.21: 36399644-36399644 on Build GRCh38
rs528675863 Chr.21: 36399489-36399489 on Build GRCh38
rs377720200 Chr.21: 36399487-36399487 on Build GRCh38
rs555711299 Chr.21: 36399541-36399541 on Build GRCh38
rs558217922 Chr.21: 36399657-36399657 on Build GRCh38
rs758033735 Chr.21: 36399602-36399602 on Build GRCh38
rs757207825 Chr.21: 36399588-36399588 on Build GRCh38
rs774978522 Chr.21: 36399527-36399527 on Build GRCh38
rs771753643 Chr.21: 36399525-36399525 on Build GRCh38
rs537762285 Chr.21: 36399539-36399539 on Build GRCh38
rs201339256 Chr.21: 36399552-36399552 on Build GRCh38
rs757938706 Chr.21: 36399533-36399533 on Build GRCh38
rs200157692 Chr.21: 36399507-36399507 on Build GRCh38
rs745732322 Chr.21: 36399596-36399596 on Build GRCh38
rs779625542 Chr.21: 36399605-36399605 on Build GRCh38
rs767914372 Chr.21: 36399563-36399563 on Build GRCh38
rs749739766 Chr.21: 36399638-36399638 on Build GRCh38
rs771603711 Chr.21: 36399639-36399639 on Build GRCh38
rs746682092 Chr.21: 36399610-36399610 on Build GRCh38
rs1014586566 Chr.21: 36399464-36399464 on Build GRCh38
rs752804794 Chr.21: 36399565-36399565 on Build GRCh38

View Full Product Details