Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 15827220-15827489 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1S2
Entrez Gene ID
8905
Gene Name
adaptor related protein complex 1 sigma 2 subunit
Gene Aliases
DC22, MRX59, MRXS21, MRXS5, MRXSF, PGS, SIGMA1B
UniGene
Hs.121592
Gene Chromosome Location
Chr.X: 15825806-15855014 on Build GRCh38
Transcripts
NM_001272071.1 NM_003916.4

SNP Detail

SNP ID Chromosome Location
rs771024691 Chr.X: 15827382-15827382 on Build GRCh38
rs775923167 Chr.X: 15827424-15827424 on Build GRCh38
rs749881573 Chr.X: 15827297-15827297 on Build GRCh38
rs944853404 Chr.X: 15827450-15827450 on Build GRCh38
rs143031717 Chr.X: 15827336-15827336 on Build GRCh38
rs377210898 Chr.X: 15827395-15827395 on Build GRCh38
rs930892856 Chr.X: 15827311-15827311 on Build GRCh38
rs370388468 Chr.X: 15827437-15827437 on Build GRCh38
rs778700579 Chr.X: 15827286-15827286 on Build GRCh38
rs772166461 Chr.X: 15827419-15827419 on Build GRCh38
rs1016898604 Chr.X: 15827324-15827324 on Build GRCh38
rs779964886 Chr.X: 15827283-15827284 on Build GRCh38
rs745884033 Chr.X: 15827403-15827403 on Build GRCh38
rs764983427 Chr.X: 15827292-15827292 on Build GRCh38
rs1028938120 Chr.X: 15827260-15827260 on Build GRCh38
rs1050648419 Chr.X: 15827390-15827391 on Build GRCh38
rs748041023 Chr.X: 15827370-15827370 on Build GRCh38
rs200662353 Chr.X: 15827301-15827301 on Build GRCh38
rs200146767 Chr.X: 15827354-15827354 on Build GRCh38
rs752727083 Chr.X: 15827325-15827325 on Build GRCh38
rs374565433 Chr.X: 15827282-15827282 on Build GRCh38
rs11559235 Chr.X: 15827333-15827333 on Build GRCh38
rs780101139 Chr.X: 15827312-15827312 on Build GRCh38
rs751546908 Chr.X: 15827353-15827353 on Build GRCh38
rs944895474 Chr.X: 15827316-15827316 on Build GRCh38
rs368784561 Chr.X: 15827279-15827279 on Build GRCh38
rs754761928 Chr.X: 15827357-15827357 on Build GRCh38
rs376685147 Chr.X: 15827278-15827278 on Build GRCh38

View Full Product Details