Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16989710-16989935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP13A2
Entrez Gene ID
23400
Gene Name
ATPase 13A2
Gene Aliases
CLN12, HSA9947, KRPPD, PARK9
UniGene
Hs.128866
Gene Chromosome Location
Chr.1: 16985958-17011972 on Build GRCh38
Transcripts
NM_001141973.2 NM_001141974.2 NM_022089.3 XM_005245810.1 XM_005245811.1 XM_005245812.1 XM_005245815.1 XM_006710512.1 XM_006710513.1 XM_011541128.1 XM_011541129.1 XM_017000844.1 XM_017000845.1 XM_017000846.1 XM_017000847.1 XM_017000848.1 XM_017000849.1 XM_017000850.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7356921 c.2437G>T p.E813* Chr.1: 16989731-16989731 on Build GRCh38
7356920 c.2569G>T p.E857* Chr.1: 16989731-16989731 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371083479 Chr.1: 16989860-16989860 on Build GRCh38
rs201321612 Chr.1: 16989825-16989825 on Build GRCh38
rs747248855 Chr.1: 16989856-16989856 on Build GRCh38
rs566172222 Chr.1: 16989752-16989752 on Build GRCh38
rs886045579 Chr.1: 16989737-16989737 on Build GRCh38
rs986226126 Chr.1: 16989819-16989819 on Build GRCh38
rs55929813 Chr.1: 16989826-16989826 on Build GRCh38
rs374008623 Chr.1: 16989779-16989779 on Build GRCh38
rs747242869 Chr.1: 16989746-16989746 on Build GRCh38
rs535209471 Chr.1: 16989821-16989821 on Build GRCh38
rs897837480 Chr.1: 16989848-16989848 on Build GRCh38
rs763659305 Chr.1: 16989884-16989884 on Build GRCh38
rs587776890 Chr.1: 16989747-16989748 on Build GRCh38
rs745954026 Chr.1: 16989742-16989742 on Build GRCh38
rs759142662 Chr.1: 16989784-16989784 on Build GRCh38
rs770434494 Chr.1: 16989776-16989776 on Build GRCh38
rs752367474 Chr.1: 16989795-16989795 on Build GRCh38
rs537112220 Chr.1: 16989871-16989871 on Build GRCh38
rs954971480 Chr.1: 16989800-16989800 on Build GRCh38
rs868725919 Chr.1: 16989764-16989764 on Build GRCh38
rs778906228 Chr.1: 16989740-16989740 on Build GRCh38
rs771708290 Chr.1: 16989879-16989879 on Build GRCh38
rs949240567 Chr.1: 16989810-16989810 on Build GRCh38
rs770381718 Chr.1: 16989876-16989876 on Build GRCh38
rs760318841 Chr.1: 16989882-16989882 on Build GRCh38
rs587777053 Chr.1: 16989739-16989739 on Build GRCh38
rs757153866 Chr.1: 16989841-16989841 on Build GRCh38
rs778854959 Chr.1: 16989846-16989846 on Build GRCh38
rs776925680 Chr.1: 16989765-16989765 on Build GRCh38
rs774924482 Chr.1: 16989881-16989881 on Build GRCh38
rs772243999 Chr.1: 16989743-16989743 on Build GRCh38
rs377073826 Chr.1: 16989851-16989851 on Build GRCh38
rs776448394 Chr.1: 16989886-16989886 on Build GRCh38
rs780146972 Chr.1: 16989854-16989854 on Build GRCh38
rs55889979 Chr.1: 16989744-16989744 on Build GRCh38
rs986327732 Chr.1: 16989868-16989868 on Build GRCh38
rs760315801 Chr.1: 16989816-16989816 on Build GRCh38
rs200809857 Chr.1: 16989878-16989878 on Build GRCh38
rs769033175 Chr.1: 16989756-16989756 on Build GRCh38
rs917176878 Chr.1: 16989891-16989891 on Build GRCh38
rs763937012 Chr.1: 16989820-16989820 on Build GRCh38
rs935118855 Chr.1: 16989782-16989782 on Build GRCh38
rs781557299 Chr.1: 16989861-16989861 on Build GRCh38
rs375894941 Chr.1: 16989773-16989773 on Build GRCh38
rs748463334 Chr.1: 16989869-16989869 on Build GRCh38
rs758449038 Chr.1: 16989853-16989853 on Build GRCh38
rs774420241 Chr.1: 16989736-16989736 on Build GRCh38
rs773837281 Chr.1: 16989877-16989877 on Build GRCh38

View Full Product Details