Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33237633-33237888 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPIFA1
Entrez Gene ID
51297
Gene Name
BPI fold containing family A member 1
Gene Aliases
LUNX, NASG, PLUNC, SPLUNC1, SPURT, bA49G10.5
UniGene
Hs.211092
Gene Chromosome Location
Chr.20: 33235996-33243309 on Build GRCh38
Transcripts
NM_001243193.1 NM_016583.3 NM_130852.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6612325 c.155C>T p.T52I Chr.20: 33237866-33237866 on Build GRCh38
5643398 c.84G>T p.L28L Chr.20: 33237795-33237795 on Build GRCh38
1726602 c.68G>A p.G23D Chr.20: 33237779-33237779 on Build GRCh38
6612323 c.58C>A p.Q20K Chr.20: 33237769-33237769 on Build GRCh38
5527391 c.35G>A p.G12E Chr.20: 33237746-33237746 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749691685 Chr.20: 33237757-33237757 on Build GRCh38
rs768913833 Chr.20: 33237711-33237711 on Build GRCh38
rs754657733 Chr.20: 33237728-33237728 on Build GRCh38
rs760652134 Chr.20: 33237773-33237773 on Build GRCh38
rs770138104 Chr.20: 33237697-33237697 on Build GRCh38
rs775898825 Chr.20: 33237698-33237698 on Build GRCh38
rs185078840 Chr.20: 33237860-33237860 on Build GRCh38
rs374193470 Chr.20: 33237727-33237727 on Build GRCh38
rs529335344 Chr.20: 33237802-33237802 on Build GRCh38
rs757991996 Chr.20: 33237799-33237799 on Build GRCh38
rs754747808 Chr.20: 33237667-33237667 on Build GRCh38
rs895842993 Chr.20: 33237659-33237659 on Build GRCh38
rs761950576 Chr.20: 33237763-33237763 on Build GRCh38
rs767514343 Chr.20: 33237714-33237714 on Build GRCh38
rs771170398 Chr.20: 33237864-33237864 on Build GRCh38
rs201375171 Chr.20: 33237785-33237785 on Build GRCh38
rs780374736 Chr.20: 33237754-33237754 on Build GRCh38
rs773236309 Chr.20: 33237770-33237770 on Build GRCh38
rs773321578 Chr.20: 33237846-33237846 on Build GRCh38
rs1032065146 Chr.20: 33237692-33237692 on Build GRCh38
rs369658054 Chr.20: 33237696-33237696 on Build GRCh38
rs561854574 Chr.20: 33237790-33237790 on Build GRCh38
rs778752160 Chr.20: 33237733-33237733 on Build GRCh38
rs761909415 Chr.20: 33237713-33237713 on Build GRCh38
rs754698570 Chr.20: 33237720-33237720 on Build GRCh38
rs750312331 Chr.20: 33237723-33237723 on Build GRCh38
rs779148150 Chr.20: 33237834-33237834 on Build GRCh38
rs200268997 Chr.20: 33237748-33237748 on Build GRCh38
rs770343234 Chr.20: 33237747-33237747 on Build GRCh38
rs749814431 Chr.20: 33237832-33237832 on Build GRCh38
rs377416490 Chr.20: 33237804-33237804 on Build GRCh38
rs200585255 Chr.20: 33237787-33237787 on Build GRCh38
rs774504523 Chr.20: 33237762-33237762 on Build GRCh38
rs778556002 Chr.20: 33237668-33237668 on Build GRCh38
rs866818565 Chr.20: 33237780-33237780 on Build GRCh38
rs140030379 Chr.20: 33237833-33237833 on Build GRCh38
rs111799330 Chr.20: 33237678-33237678 on Build GRCh38
rs373386646 Chr.20: 33237699-33237699 on Build GRCh38
rs748886798 Chr.20: 33237666-33237666 on Build GRCh38
rs1047880651 Chr.20: 33237857-33237857 on Build GRCh38
rs748394815 Chr.20: 33237837-33237837 on Build GRCh38
rs777162544 Chr.20: 33237745-33237745 on Build GRCh38
rs550742438 Chr.20: 33237827-33237827 on Build GRCh38
rs772244544 Chr.20: 33237838-33237838 on Build GRCh38
rs752336724 Chr.20: 33237793-33237793 on Build GRCh38
rs371166466 Chr.20: 33237707-33237707 on Build GRCh38
rs775887890 Chr.20: 33237798-33237799 on Build GRCh38
rs1010921621 Chr.20: 33237796-33237796 on Build GRCh38
rs150581925 Chr.20: 33237671-33237671 on Build GRCh38
rs760530605 Chr.20: 33237724-33237724 on Build GRCh38
rs868857531 Chr.20: 33237729-33237729 on Build GRCh38
rs144532776 Chr.20: 33237765-33237765 on Build GRCh38
rs769038767 Chr.20: 33237761-33237761 on Build GRCh38
rs575497639 Chr.20: 33237734-33237734 on Build GRCh38
rs770083139 Chr.20: 33237790-33237790 on Build GRCh38
rs6120177 Chr.20: 33237744-33237744 on Build GRCh38
rs745964213 Chr.20: 33237754-33237754 on Build GRCh38
rs2752908 Chr.20: 33237687-33237687 on Build GRCh38

View Full Product Details