Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 31577823-31578064 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC129
Entrez Gene ID
223075
Gene Name
coiled-coil domain containing 129
Gene Aliases
-
UniGene
Hs.224269
Gene Chromosome Location
Chr.7: 31514071-31658720 on Build GRCh38
Transcripts
NM_001257967.1 NM_001257968.1 NM_194300.3 NR_047565.1 XM_011515213.1 XM_017011871.1 XM_017011872.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7359725 c.754C>A p.P252T Chr.7: 31577940-31577940 on Build GRCh38
6882906 c.786G>A p.E262E Chr.7: 31577972-31577972 on Build GRCh38
4556786 c.702G>A p.L234L Chr.7: 31577888-31577888 on Build GRCh38
3394716 c.748C>T p.R250* Chr.7: 31578012-31578012 on Build GRCh38
4556785 c.624G>A p.L208L Chr.7: 31577888-31577888 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs928730438 Chr.7: 31577987-31577987 on Build GRCh38
rs748758260 Chr.7: 31577915-31577915 on Build GRCh38
rs373956174 Chr.7: 31577983-31577983 on Build GRCh38
rs201901788 Chr.7: 31578013-31578013 on Build GRCh38
rs756099710 Chr.7: 31578007-31578007 on Build GRCh38
rs757044575 Chr.7: 31577872-31577872 on Build GRCh38
rs750607089 Chr.7: 31577871-31577871 on Build GRCh38
rs989789321 Chr.7: 31577876-31577876 on Build GRCh38
rs568878337 Chr.7: 31578040-31578040 on Build GRCh38
rs758532403 Chr.7: 31577906-31577906 on Build GRCh38
rs756001751 Chr.7: 31578030-31578030 on Build GRCh38
rs192648448 Chr.7: 31577859-31577859 on Build GRCh38
rs745835615 Chr.7: 31577888-31577888 on Build GRCh38
rs752868206 Chr.7: 31577950-31577950 on Build GRCh38
rs779836058 Chr.7: 31578009-31578009 on Build GRCh38
rs766861413 Chr.7: 31577927-31577927 on Build GRCh38
rs768474435 Chr.7: 31577921-31577921 on Build GRCh38
rs757498755 Chr.7: 31577876-31577876 on Build GRCh38
rs768082867 Chr.7: 31577880-31577881 on Build GRCh38
rs761178359 Chr.7: 31577926-31577926 on Build GRCh38
rs760116621 Chr.7: 31577944-31577944 on Build GRCh38
rs745610581 Chr.7: 31578006-31578006 on Build GRCh38
rs376757677 Chr.7: 31578000-31578000 on Build GRCh38
rs373138016 Chr.7: 31578043-31578043 on Build GRCh38
rs769555214 Chr.7: 31577894-31577894 on Build GRCh38
rs781052167 Chr.7: 31577884-31577884 on Build GRCh38
rs868039425 Chr.7: 31577998-31577998 on Build GRCh38
rs200848834 Chr.7: 31577892-31577892 on Build GRCh38
rs151254328 Chr.7: 31577866-31577866 on Build GRCh38
rs140428776 Chr.7: 31577922-31577922 on Build GRCh38
rs766314963 Chr.7: 31577979-31577979 on Build GRCh38
rs748849900 Chr.7: 31578012-31578012 on Build GRCh38
rs750949457 Chr.7: 31577946-31577947 on Build GRCh38
rs977112487 Chr.7: 31577970-31577970 on Build GRCh38
rs757417828 Chr.7: 31577999-31577999 on Build GRCh38
rs546906392 Chr.7: 31577931-31577931 on Build GRCh38
rs775097558 Chr.7: 31577905-31577905 on Build GRCh38
rs200031625 Chr.7: 31577955-31577955 on Build GRCh38
rs765608598 Chr.7: 31577948-31577948 on Build GRCh38
rs184653370 Chr.7: 31578025-31578025 on Build GRCh38

View Full Product Details