Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73914631-73914904 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTG2
Entrez Gene ID
72
Gene Name
actin, gamma 2, smooth muscle, enteric
Gene Aliases
ACT, ACTA3, ACTE, ACTL3, ACTSG, VSCM
UniGene
Hs.516105
Gene Chromosome Location
Chr.2: 73892966-73919653 on Build GRCh38
Transcripts
NM_001199893.1 NM_001615.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6542727 c.738G>T p.G246G Chr.2: 73914804-73914804 on Build GRCh38
4095689 c.681G>C p.E227D Chr.2: 73914747-73914747 on Build GRCh38
6592881 c.727C>T p.L243L Chr.2: 73914793-73914793 on Build GRCh38
4555153 c.643G>A p.E215K Chr.2: 73914709-73914709 on Build GRCh38
3840016 c.645G>C p.E215D Chr.2: 73914711-73914711 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377258138 Chr.2: 73914676-73914676 on Build GRCh38
rs760189166 Chr.2: 73914779-73914779 on Build GRCh38
rs757370414 Chr.2: 73914723-73914723 on Build GRCh38
rs765884368 Chr.2: 73914829-73914829 on Build GRCh38
rs964154418 Chr.2: 73914708-73914708 on Build GRCh38
rs3190470 Chr.2: 73914763-73914763 on Build GRCh38
rs202002293 Chr.2: 73914729-73914729 on Build GRCh38
rs922673109 Chr.2: 73914744-73914744 on Build GRCh38
rs760302494 Chr.2: 73914659-73914659 on Build GRCh38
rs587777387 Chr.2: 73914835-73914835 on Build GRCh38
rs748032661 Chr.2: 73914715-73914715 on Build GRCh38
rs201215277 Chr.2: 73914720-73914720 on Build GRCh38
rs765920521 Chr.2: 73914783-73914783 on Build GRCh38
rs936010213 Chr.2: 73914843-73914843 on Build GRCh38
rs778124051 Chr.2: 73914699-73914699 on Build GRCh38
rs111892371 Chr.2: 73914873-73914873 on Build GRCh38
rs755478171 Chr.2: 73914677-73914677 on Build GRCh38
rs760143686 Chr.2: 73914824-73914824 on Build GRCh38
rs764306389 Chr.2: 73914849-73914849 on Build GRCh38
rs765796117 Chr.2: 73914671-73914671 on Build GRCh38
rs747183647 Chr.2: 73914722-73914722 on Build GRCh38
rs3209900 Chr.2: 73914821-73914821 on Build GRCh38
rs768527884 Chr.2: 73914758-73914758 on Build GRCh38
rs201286699 Chr.2: 73914818-73914818 on Build GRCh38
rs797044959 Chr.2: 73914836-73914836 on Build GRCh38
rs113252317 Chr.2: 73914874-73914874 on Build GRCh38
rs146517993 Chr.2: 73914690-73914690 on Build GRCh38
rs781084881 Chr.2: 73914870-73914870 on Build GRCh38
rs370526448 Chr.2: 73914774-73914774 on Build GRCh38
rs199962888 Chr.2: 73914768-73914768 on Build GRCh38
rs758804225 Chr.2: 73914697-73914697 on Build GRCh38
rs867923618 Chr.2: 73914848-73914848 on Build GRCh38
rs200875518 Chr.2: 73914704-73914704 on Build GRCh38
rs758889276 Chr.2: 73914840-73914840 on Build GRCh38
rs765727614 Chr.2: 73914688-73914688 on Build GRCh38
rs753251409 Chr.2: 73914830-73914830 on Build GRCh38
rs747982211 Chr.2: 73914767-73914767 on Build GRCh38
rs749252651 Chr.2: 73914750-73914750 on Build GRCh38
rs775462646 Chr.2: 73914682-73914685 on Build GRCh38
rs1041839772 Chr.2: 73914773-73914773 on Build GRCh38
rs751807957 Chr.2: 73914858-73914858 on Build GRCh38

View Full Product Details