Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 41404649-41404910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHD6
Entrez Gene ID
84181
Gene Name
chromodomain helicase DNA binding protein 6
Gene Aliases
CHD-6, CHD5, RIGB
UniGene
Hs.740645
Gene Chromosome Location
Chr.20: 41402101-41618494 on Build GRCh38
Transcripts
NM_032221.4 XM_005260573.2 XM_005260576.4 XM_011529080.2 XM_011529082.2 XM_017028099.1 XM_017028100.1 XM_017028101.1 XM_017028103.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5528632 c.8000C>T p.S2667F Chr.20: 41404741-41404741 on Build GRCh38
4098433 c.7857T>G p.P2619P Chr.20: 41404884-41404884 on Build GRCh38
4098432 c.8091A>G p.E2697E Chr.20: 41404650-41404650 on Build GRCh38
6319241 c.8052C>A p.N2684K Chr.20: 41404689-41404689 on Build GRCh38
3291624 c.8066G>A p.S2689N Chr.20: 41404675-41404675 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756085841 Chr.20: 41404765-41404765 on Build GRCh38
rs1012444740 Chr.20: 41404802-41404802 on Build GRCh38
rs146425509 Chr.20: 41404763-41404763 on Build GRCh38
rs906292841 Chr.20: 41404857-41404857 on Build GRCh38
rs770764198 Chr.20: 41404794-41404794 on Build GRCh38
rs572418298 Chr.20: 41404763-41404765 on Build GRCh38
rs751303308 Chr.20: 41404754-41404754 on Build GRCh38
rs749105703 Chr.20: 41404791-41404791 on Build GRCh38
rs899403144 Chr.20: 41404878-41404878 on Build GRCh38
rs982520060 Chr.20: 41404819-41404819 on Build GRCh38
rs774367979 Chr.20: 41404676-41404676 on Build GRCh38
rs374665904 Chr.20: 41404682-41404682 on Build GRCh38
rs866463844 Chr.20: 41404694-41404694 on Build GRCh38
rs759287274 Chr.20: 41404865-41404865 on Build GRCh38
rs1038067600 Chr.20: 41404863-41404863 on Build GRCh38
rs754411813 Chr.20: 41404758-41404758 on Build GRCh38
rs779077688 Chr.20: 41404796-41404796 on Build GRCh38
rs201674871 Chr.20: 41404807-41404807 on Build GRCh38
rs922436097 Chr.20: 41404786-41404786 on Build GRCh38
rs573605078 Chr.20: 41404764-41404764 on Build GRCh38
rs908324328 Chr.20: 41404852-41404852 on Build GRCh38
rs762651945 Chr.20: 41404736-41404736 on Build GRCh38
rs142643615 Chr.20: 41404683-41404683 on Build GRCh38
rs79519789 Chr.20: 41404806-41404806 on Build GRCh38
rs762243573 Chr.20: 41404850-41404850 on Build GRCh38
rs777394866 Chr.20: 41404781-41404781 on Build GRCh38
rs761302167 Chr.20: 41404688-41404688 on Build GRCh38
rs766149109 Chr.20: 41404748-41404748 on Build GRCh38
rs775652248 Chr.20: 41404812-41404812 on Build GRCh38
rs201618483 Chr.20: 41404869-41404869 on Build GRCh38
rs575778947 Chr.20: 41404853-41404853 on Build GRCh38
rs377255552 Chr.20: 41404859-41404859 on Build GRCh38
rs770605015 Chr.20: 41404700-41404701 on Build GRCh38
rs764306469 Chr.20: 41404700-41404700 on Build GRCh38
rs769235551 Chr.20: 41404841-41404841 on Build GRCh38
rs745580314 Chr.20: 41404822-41404822 on Build GRCh38
rs368641098 Chr.20: 41404881-41404881 on Build GRCh38
rs372231144 Chr.20: 41404847-41404847 on Build GRCh38
rs766874842 Chr.20: 41404868-41404868 on Build GRCh38
rs368334964 Chr.20: 41404701-41404701 on Build GRCh38

View Full Product Details