Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 38417742-38418003 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMPH
Entrez Gene ID
273
Gene Name
amphiphysin
Gene Aliases
AMPH1
UniGene
Hs.592182
Gene Chromosome Location
Chr.7: 38383694-38631567 on Build GRCh38
Transcripts
NM_001635.3 NM_139316.2 XM_006715689.3 XM_006715690.3 XM_011515271.2 XM_017011995.1 XM_017011996.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1450611 c.1376A>G p.E459G Chr.7: 38417847-38417847 on Build GRCh38
4777437 c.1375G>A p.E459K Chr.7: 38417848-38417848 on Build GRCh38
7420874 c.1391A>T p.E464V Chr.7: 38417832-38417832 on Build GRCh38
3923591 c.1282G>A p.E428K Chr.7: 38417941-38417941 on Build GRCh38
1089476 c.1391A>G p.E464G Chr.7: 38417832-38417832 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750537808 Chr.7: 38417901-38417901 on Build GRCh38
rs756088309 Chr.7: 38417904-38417904 on Build GRCh38
rs765805153 Chr.7: 38417777-38417777 on Build GRCh38
rs779971440 Chr.7: 38417913-38417913 on Build GRCh38
rs906260088 Chr.7: 38417831-38417831 on Build GRCh38
rs199697871 Chr.7: 38417888-38417888 on Build GRCh38
rs201059599 Chr.7: 38417854-38417854 on Build GRCh38
rs749289574 Chr.7: 38417918-38417918 on Build GRCh38
rs768373657 Chr.7: 38417830-38417830 on Build GRCh38
rs747862983 Chr.7: 38417935-38417935 on Build GRCh38
rs745915061 Chr.7: 38417804-38417804 on Build GRCh38
rs771596953 Chr.7: 38417853-38417853 on Build GRCh38
rs778159398 Chr.7: 38417784-38417784 on Build GRCh38
rs781391518 Chr.7: 38417797-38417797 on Build GRCh38
rs766656638 Chr.7: 38417764-38417764 on Build GRCh38
rs891387766 Chr.7: 38417937-38417937 on Build GRCh38
rs760249336 Chr.7: 38417949-38417949 on Build GRCh38
rs377750480 Chr.7: 38417788-38417788 on Build GRCh38
rs202236524 Chr.7: 38417809-38417809 on Build GRCh38
rs373071104 Chr.7: 38417881-38417881 on Build GRCh38
rs757550847 Chr.7: 38417890-38417890 on Build GRCh38
rs759911669 Chr.7: 38417832-38417832 on Build GRCh38
rs1033343259 Chr.7: 38417921-38417921 on Build GRCh38
rs201353173 Chr.7: 38417781-38417781 on Build GRCh38
rs200519832 Chr.7: 38417891-38417891 on Build GRCh38
rs1005453871 Chr.7: 38417938-38417938 on Build GRCh38
rs771785238 Chr.7: 38417940-38417940 on Build GRCh38
rs150091535 Chr.7: 38417934-38417934 on Build GRCh38
rs761580305 Chr.7: 38417845-38417845 on Build GRCh38
rs772695203 Chr.7: 38417943-38417943 on Build GRCh38
rs774126913 Chr.7: 38417843-38417843 on Build GRCh38
rs368760883 Chr.7: 38417814-38417814 on Build GRCh38
rs140860940 Chr.7: 38417865-38417865 on Build GRCh38
rs925119015 Chr.7: 38417919-38417919 on Build GRCh38
rs763418401 Chr.7: 38417882-38417882 on Build GRCh38
rs770504090 Chr.7: 38417969-38417969 on Build GRCh38
rs199995985 Chr.7: 38417789-38417789 on Build GRCh38
rs558887615 Chr.7: 38417771-38417771 on Build GRCh38
rs776280981 Chr.7: 38417971-38417971 on Build GRCh38
rs749093263 Chr.7: 38417824-38417824 on Build GRCh38
rs768564392 Chr.7: 38417928-38417928 on Build GRCh38
rs1008368912 Chr.7: 38417958-38417958 on Build GRCh38
rs752059132 Chr.7: 38417935-38417935 on Build GRCh38
rs760060401 Chr.7: 38417855-38417855 on Build GRCh38

View Full Product Details