Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 159323065-159323290 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL12B
Entrez Gene ID
3593
Gene Name
interleukin 12B
Gene Aliases
CLMF, CLMF2, IL-12B, IMD28, IMD29, NKSF, NKSF2
UniGene
Hs.674
Gene Chromosome Location
Chr.5: 159314783-159330473 on Build GRCh38
Transcripts
NM_002187.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3613384 c.223C>T p.L75L Chr.5: 159323195-159323195 on Build GRCh38
1435656 c.204C>T p.V68V Chr.5: 159323214-159323214 on Build GRCh38
5895768 c.241G>T p.E81* Chr.5: 159323177-159323177 on Build GRCh38
6836646 c.269G>C p.C90S Chr.5: 159323149-159323149 on Build GRCh38
5109673 c.294C>T p.S98S Chr.5: 159323124-159323124 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140131034 Chr.5: 159323263-159323263 on Build GRCh38
rs753044197 Chr.5: 159323152-159323152 on Build GRCh38
rs756537897 Chr.5: 159323171-159323171 on Build GRCh38
rs765124116 Chr.5: 159323242-159323242 on Build GRCh38
rs751689646 Chr.5: 159323133-159323133 on Build GRCh38
rs1010779225 Chr.5: 159323122-159323122 on Build GRCh38
rs778518679 Chr.5: 159323189-159323189 on Build GRCh38
rs563992321 Chr.5: 159323187-159323187 on Build GRCh38
rs267600520 Chr.5: 159323140-159323140 on Build GRCh38
rs56287471 Chr.5: 159323135-159323135 on Build GRCh38
rs761837865 Chr.5: 159323237-159323237 on Build GRCh38
rs768578364 Chr.5: 159323223-159323223 on Build GRCh38
rs142998256 Chr.5: 159323118-159323118 on Build GRCh38
rs776655957 Chr.5: 159323229-159323229 on Build GRCh38
rs587776807 Chr.5: 159323098-159323099 on Build GRCh38
rs786201006 Chr.5: 159323113-159323120 on Build GRCh38
rs1015688626 Chr.5: 159323216-159323216 on Build GRCh38
rs986782158 Chr.5: 159323217-159323217 on Build GRCh38
rs142017503 Chr.5: 159323136-159323136 on Build GRCh38
rs150659914 Chr.5: 159323227-159323227 on Build GRCh38
rs183111978 Chr.5: 159323261-159323261 on Build GRCh38
rs955389274 Chr.5: 159323162-159323162 on Build GRCh38
rs763062544 Chr.5: 159323125-159323125 on Build GRCh38
rs749808063 Chr.5: 159323178-159323178 on Build GRCh38
rs201684769 Chr.5: 159323210-159323210 on Build GRCh38
rs868595228 Chr.5: 159323158-159323158 on Build GRCh38
rs974315484 Chr.5: 159323266-159323266 on Build GRCh38
rs745410494 Chr.5: 159323197-159323197 on Build GRCh38
rs200722574 Chr.5: 159323117-159323117 on Build GRCh38
rs771734221 Chr.5: 159323208-159323208 on Build GRCh38
rs144694601 Chr.5: 159323119-159323119 on Build GRCh38
rs775325033 Chr.5: 159323096-159323096 on Build GRCh38
rs746799998 Chr.5: 159323221-159323221 on Build GRCh38
rs540842370 Chr.5: 159323176-159323176 on Build GRCh38
rs368468349 Chr.5: 159323132-159323132 on Build GRCh38

View Full Product Details