Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 247531685-247531956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCSAML
Entrez Gene ID
148823
Gene Name
germinal center associated signaling and motility like
Gene Aliases
C1orf150
UniGene
Hs.732113
Gene Chromosome Location
Chr.1: 247507058-247577690 on Build GRCh38
Transcripts
NM_001281834.1 NM_001281835.1 NM_001281836.1 NM_001281837.1 NM_001281838.1 NM_001281853.1 XM_017000399.1 NM_198074.4

Gene Symbol
OR2C3
Entrez Gene ID
81472
Gene Name
olfactory receptor family 2 subfamily C member 3
Gene Aliases
OR2C4, OR2C5P, OST742
UniGene
Hs.730218
Gene Chromosome Location
Chr.1: 247530132-247533839 on Build GRCh38
Transcripts
NM_001281834.1 NM_001281835.1 NM_001281836.1 NM_001281837.1 NM_001281838.1 NM_001281853.1 XM_017000399.1 NM_198074.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
365539 c.803C>A p.S268Y Chr.1: 247531709-247531709 on Build GRCh38
7237848 c.695C>T p.A232V Chr.1: 247531817-247531817 on Build GRCh38
273048 c.659G>A p.G220D Chr.1: 247531853-247531853 on Build GRCh38
7121252 c.805C>A p.H269N Chr.1: 247531707-247531707 on Build GRCh38
5482418 c.667G>T p.A223S Chr.1: 247531845-247531845 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747543947 Chr.1: 247531850-247531850 on Build GRCh38
rs146484773 Chr.1: 247531921-247531921 on Build GRCh38
rs889439338 Chr.1: 247531819-247531819 on Build GRCh38
rs1005233087 Chr.1: 247531820-247531820 on Build GRCh38
rs925600424 Chr.1: 247531857-247531857 on Build GRCh38
rs746274604 Chr.1: 247531742-247531742 on Build GRCh38
rs747444615 Chr.1: 247531926-247531926 on Build GRCh38
rs1033622417 Chr.1: 247531733-247531734 on Build GRCh38
rs756690486 Chr.1: 247531814-247531814 on Build GRCh38
rs777909338 Chr.1: 247531839-247531839 on Build GRCh38
rs376920492 Chr.1: 247531766-247531766 on Build GRCh38
rs1053300184 Chr.1: 247531793-247531793 on Build GRCh38
rs149975458 Chr.1: 247531837-247531837 on Build GRCh38
rs777271986 Chr.1: 247531726-247531726 on Build GRCh38
rs758240248 Chr.1: 247531934-247531934 on Build GRCh38
rs776976495 Chr.1: 247531854-247531854 on Build GRCh38
rs770566998 Chr.1: 247531736-247531736 on Build GRCh38
rs780497675 Chr.1: 247531844-247531844 on Build GRCh38
rs755636356 Chr.1: 247531722-247531722 on Build GRCh38
rs868813772 Chr.1: 247531900-247531900 on Build GRCh38
rs759386191 Chr.1: 247531783-247531783 on Build GRCh38
rs763096579 Chr.1: 247531873-247531873 on Build GRCh38
rs756092890 Chr.1: 247531764-247531765 on Build GRCh38
rs781174311 Chr.1: 247531720-247531720 on Build GRCh38
rs767333048 Chr.1: 247531875-247531875 on Build GRCh38
rs753715148 Chr.1: 247531894-247531894 on Build GRCh38
rs779833070 Chr.1: 247531925-247531925 on Build GRCh38
rs778407855 Chr.1: 247531740-247531740 on Build GRCh38
rs866695097 Chr.1: 247531859-247531859 on Build GRCh38
rs748923172 Chr.1: 247531732-247531732 on Build GRCh38
rs529863004 Chr.1: 247531753-247531753 on Build GRCh38
rs762023698 Chr.1: 247531855-247531855 on Build GRCh38
rs767243257 Chr.1: 247531784-247531784 on Build GRCh38
rs374074268 Chr.1: 247531904-247531904 on Build GRCh38
rs768620569 Chr.1: 247531755-247531755 on Build GRCh38
rs146068024 Chr.1: 247531841-247531841 on Build GRCh38
rs764131898 Chr.1: 247531884-247531884 on Build GRCh38
rs781752495 Chr.1: 247531933-247531933 on Build GRCh38
rs369513053 Chr.1: 247531881-247531881 on Build GRCh38
rs961158096 Chr.1: 247531836-247531836 on Build GRCh38
rs1026535095 Chr.1: 247531804-247531804 on Build GRCh38
rs769258016 Chr.1: 247531852-247531852 on Build GRCh38
rs773436526 Chr.1: 247531872-247531872 on Build GRCh38
rs538753683 Chr.1: 247531808-247531808 on Build GRCh38
rs200221174 Chr.1: 247531772-247531772 on Build GRCh38
rs752637692 Chr.1: 247531800-247531800 on Build GRCh38
rs754740400 Chr.1: 247531719-247531719 on Build GRCh38
rs752454845 Chr.1: 247531760-247531761 on Build GRCh38
rs778690362 Chr.1: 247531821-247531821 on Build GRCh38
rs769749914 Chr.1: 247531869-247531869 on Build GRCh38
rs756844910 Chr.1: 247531903-247531903 on Build GRCh38
rs139293334 Chr.1: 247531779-247531779 on Build GRCh38
rs570164676 Chr.1: 247531855-247531856 on Build GRCh38
rs202039549 Chr.1: 247531752-247531752 on Build GRCh38
rs753436393 Chr.1: 247531810-247531810 on Build GRCh38
rs750101190 Chr.1: 247531917-247531917 on Build GRCh38
rs755297878 Chr.1: 247531928-247531928 on Build GRCh38
rs745498893 Chr.1: 247531840-247531840 on Build GRCh38
rs141524952 Chr.1: 247531809-247531809 on Build GRCh38

View Full Product Details