Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 23322065-23322338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOXL2
Entrez Gene ID
4017
Gene Name
lysyl oxidase like 2
Gene Aliases
LOR2, WS9-14
UniGene
Hs.626637
Gene Chromosome Location
Chr.8: 23296897-23404209 on Build GRCh38
Transcripts
NM_002318.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6423362 c.1174G>T p.E392* Chr.8: 23322258-23322258 on Build GRCh38
5746852 c.1276A>G p.T426A Chr.8: 23322156-23322156 on Build GRCh38
1456160 c.1221C>A p.F407L Chr.8: 23322211-23322211 on Build GRCh38
1699822 c.1195G>C p.E399Q Chr.8: 23322237-23322237 on Build GRCh38
1098252 c.1199_1200insA p.S401fs*149 Chr.8: 23322232-23322233 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780056153 Chr.8: 23322182-23322182 on Build GRCh38
rs199848147 Chr.8: 23322277-23322277 on Build GRCh38
rs772478913 Chr.8: 23322280-23322280 on Build GRCh38
rs201086267 Chr.8: 23322276-23322276 on Build GRCh38
rs867777268 Chr.8: 23322211-23322214 on Build GRCh38
rs774478852 Chr.8: 23322228-23322228 on Build GRCh38
rs963804189 Chr.8: 23322113-23322113 on Build GRCh38
rs753979228 Chr.8: 23322157-23322157 on Build GRCh38
rs761040157 Chr.8: 23322285-23322285 on Build GRCh38
rs374072510 Chr.8: 23322294-23322294 on Build GRCh38
rs756084529 Chr.8: 23322130-23322130 on Build GRCh38
rs576573847 Chr.8: 23322245-23322245 on Build GRCh38
rs757328199 Chr.8: 23322224-23322224 on Build GRCh38
rs762551423 Chr.8: 23322304-23322304 on Build GRCh38
rs757837599 Chr.8: 23322175-23322175 on Build GRCh38
rs754196114 Chr.8: 23322303-23322303 on Build GRCh38
rs201295841 Chr.8: 23322271-23322271 on Build GRCh38
rs768039441 Chr.8: 23322235-23322235 on Build GRCh38
rs764499044 Chr.8: 23322291-23322291 on Build GRCh38
rs779494470 Chr.8: 23322253-23322253 on Build GRCh38
rs754918494 Chr.8: 23322259-23322259 on Build GRCh38
rs773524538 Chr.8: 23322208-23322208 on Build GRCh38
rs746493999 Chr.8: 23322258-23322258 on Build GRCh38
rs144194678 Chr.8: 23322184-23322184 on Build GRCh38
rs145891351 Chr.8: 23322209-23322209 on Build GRCh38
rs559347716 Chr.8: 23322310-23322310 on Build GRCh38
rs577223373 Chr.8: 23322313-23322313 on Build GRCh38
rs753054497 Chr.8: 23322136-23322136 on Build GRCh38
rs558284783 Chr.8: 23322204-23322204 on Build GRCh38
rs760029868 Chr.8: 23322230-23322230 on Build GRCh38
rs756942388 Chr.8: 23322148-23322148 on Build GRCh38
rs201330878 Chr.8: 23322109-23322109 on Build GRCh38
rs766315101 Chr.8: 23322223-23322223 on Build GRCh38
rs140255048 Chr.8: 23322164-23322164 on Build GRCh38
rs754764095 Chr.8: 23322205-23322205 on Build GRCh38
rs539983434 Chr.8: 23322183-23322183 on Build GRCh38
rs960602563 Chr.8: 23322137-23322137 on Build GRCh38
rs979517532 Chr.8: 23322181-23322181 on Build GRCh38
rs148580652 Chr.8: 23322238-23322238 on Build GRCh38
rs745414107 Chr.8: 23322171-23322171 on Build GRCh38
rs776333766 Chr.8: 23322193-23322193 on Build GRCh38
rs758053870 Chr.8: 23322249-23322249 on Build GRCh38
rs751051285 Chr.8: 23322312-23322312 on Build GRCh38
rs759640733 Chr.8: 23322278-23322278 on Build GRCh38
rs201646081 Chr.8: 23322247-23322247 on Build GRCh38
rs759832597 Chr.8: 23322130-23322131 on Build GRCh38
rs142942245 Chr.8: 23322239-23322239 on Build GRCh38
rs746347445 Chr.8: 23322284-23322284 on Build GRCh38

View Full Product Details